| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015764.hg.1 |
ENST00000360507.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015764.hg.1 |
ENST00000360507.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015764.hg.1 |
ENST00000360507.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015764.hg.1 |
ENST00000360507.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015764.hg.1 |
ENST00000360507.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015764.hg.1 |
ENST00000360507.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015764.hg.1 |
ENST00000360507.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015764.hg.1 |
ENST00000360507.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015764.hg.1 |
ENST00000360507.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015764.hg.1 |
ENST00000360507.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015764.hg.1 |
ENST00000360507.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015764.hg.1 |
ENST00000360507.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015764.hg.1 |
ENST00000360507.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015764.hg.1 |
ENST00000360507.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015764.hg.1 |
ENST00000409789.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015764.hg.1 |
ENST00000409789.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015764.hg.1 |
ENST00000409789.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015764.hg.1 |
ENST00000409789.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015764.hg.1 |
ENST00000409789.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015764.hg.1 |
ENST00000409789.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015764.hg.1 |
ENST00000409789.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015764.hg.1 |
ENST00000409789.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015764.hg.1 |
ENST00000409789.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015764.hg.1 |
ENST00000409789.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015764.hg.1 |
ENST00000409789.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015764.hg.1 |
ENST00000409789.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015764.hg.1 |
ENST00000409789.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015764.hg.1 |
ENST00000451647.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015764.hg.1 |
ENST00000451647.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015764.hg.1 |
ENST00000451647.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015764.hg.1 |
ENST00000451647.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015764.hg.1 |
ENST00000451647.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015764.hg.1 |
ENST00000451647.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015764.hg.1 |
ENST00000451647.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015764.hg.1 |
ENST00000451647.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015764.hg.1 |
ENST00000451647.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015764.hg.1 |
ENST00000451647.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015764.hg.1 |
ENST00000453038.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015764.hg.1 |
ENST00000453038.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015764.hg.1 |
ENST00000453038.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015764.hg.1 |
ENST00000453038.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015764.hg.1 |
ENST00000453038.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015764.hg.1 |
ENST00000453038.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015764.hg.1 |
ENST00000453038.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015764.hg.1 |
ENST00000453038.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015764.hg.1 |
ENST00000453038.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015764.hg.1 |
ENST00000453038.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015764.hg.1 |
NM_015680.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015764.hg.1 |
NM_015680.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015764.hg.1 |
NM_015680.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015764.hg.1 |
NM_015680.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015764.hg.1 |
NM_015680.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015764.hg.1 |
NM_015680.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015764.hg.1 |
NM_015680.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015764.hg.1 |
NM_015680.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015764.hg.1 |
NM_015680.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015764.hg.1 |
NM_015680.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015764.hg.1 |
NM_015680.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015764.hg.1 |
NM_015680.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015764.hg.1 |
NM_015680.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015764.hg.1 |
NM_015680.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200015764.hg.1 |
ENST00000360507.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200015764.hg.1 |
ENST00000360507.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200015764.hg.1 |
ENST00000360507.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200015764.hg.1 |
ENST00000360507.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200015764.hg.1 |
ENST00000360507.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200015764.hg.1 |
ENST00000360507.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200015764.hg.1 |
ENST00000360507.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200015764.hg.1 |
ENST00000360507.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200015764.hg.1 |
ENST00000360507.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200015764.hg.1 |
ENST00000360507.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200015764.hg.1 |
ENST00000360507.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200015764.hg.1 |
ENST00000360507.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200015764.hg.1 |
ENST00000360507.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200015764.hg.1 |
ENST00000360507.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200015764.hg.1 |
ENST00000409789.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200015764.hg.1 |
ENST00000409789.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200015764.hg.1 |
ENST00000409789.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200015764.hg.1 |
ENST00000409789.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200015764.hg.1 |
ENST00000409789.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200015764.hg.1 |
ENST00000409789.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200015764.hg.1 |
ENST00000409789.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200015764.hg.1 |
ENST00000409789.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200015764.hg.1 |
ENST00000409789.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200015764.hg.1 |
ENST00000409789.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200015764.hg.1 |
ENST00000409789.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200015764.hg.1 |
ENST00000409789.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200015764.hg.1 |
ENST00000409789.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200015764.hg.1 |
ENST00000451647.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200015764.hg.1 |
ENST00000451647.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200015764.hg.1 |
ENST00000451647.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200015764.hg.1 |
ENST00000451647.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200015764.hg.1 |
ENST00000451647.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200015764.hg.1 |
ENST00000451647.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200015764.hg.1 |
ENST00000451647.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200015764.hg.1 |
ENST00000451647.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200015764.hg.1 |
ENST00000451647.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200015764.hg.1 |
ENST00000451647.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200015764.hg.1 |
ENST00000453038.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200015764.hg.1 |
ENST00000453038.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200015764.hg.1 |
ENST00000453038.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200015764.hg.1 |
ENST00000453038.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200015764.hg.1 |
ENST00000453038.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200015764.hg.1 |
ENST00000453038.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200015764.hg.1 |
ENST00000453038.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200015764.hg.1 |
ENST00000453038.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200015764.hg.1 |
ENST00000453038.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200015764.hg.1 |
ENST00000453038.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200015764.hg.1 |
NM_015680.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200015764.hg.1 |
NM_015680.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200015764.hg.1 |
NM_015680.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200015764.hg.1 |
NM_015680.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200015764.hg.1 |
NM_015680.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200015764.hg.1 |
NM_015680.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200015764.hg.1 |
NM_015680.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200015764.hg.1 |
NM_015680.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200015764.hg.1 |
NM_015680.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200015764.hg.1 |
NM_015680.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200015764.hg.1 |
NM_015680.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200015764.hg.1 |
NM_015680.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200015764.hg.1 |
NM_015680.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200015764.hg.1 |
NM_015680.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200215853.hg.1 |
ENST00000360507.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200215853.hg.1 |
ENST00000409789.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200215853.hg.1 |
ENST00000451647.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200215853.hg.1 |
ENST00000453038.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200215853.hg.1 |
NM_015680.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200215853.hg.1 |
ENST00000360507.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200215853.hg.1 |
ENST00000409789.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200215853.hg.1 |
ENST00000451647.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200215853.hg.1 |
ENST00000453038.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200215853.hg.1 |
NM_015680.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200215853.hg.1 |
TC0200015764.hg.1 |
Clariom D TC |