Assay Details
| Gene Symbol: LAMA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011525655.2 | XP_011523957.1 | 56 - 57 | 8211 | 76 | |
| XM_011525656.2 | XP_011523958.1 | 46 - 47 | 6632 | 76 | ||
More Information
Set Membership: |
|

LAMA1
Genomic Map