Assay Details
| Gene Symbol: CCDC169-SOHLH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198910.1 | NP_001185839.1 | 13 - 14 | 1427 | 79 | |
| GenBank mRNA | AK301863.1 | - | 13 - 14 | 1414 | 79 | |
| Gene Symbol: SOHLH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017826.2 | NP_060296.2 | 8 - 9 | 970 | 79 | |
| GenBank mRNA | AK000456.1 | - | 8 - 9 | 894 | 79 | |
| AY884305.1 | - | 7 - 8 | 812 | 79 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC13001724.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
transcription from RNA polymerase II promoter
multicellular organism development
spermatogenesis
cell differentiation
positive regulation of transcription from RNA polymerase II promoter
oogenesis
core promoter sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding
protein dimerization activity
transcription from RNA polymerase II promoter
multicellular organism development
spermatogenesis
cell differentiation
positive regulation of transcription from RNA polymerase II promoter
oogenesis
core promoter sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding
protein dimerization activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1300010032.hg.1 | ENST00000317764.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300010032.hg.1 | ENST00000317764.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300010032.hg.1 | ENST00000317764.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300010032.hg.1 | ENST00000317764.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300010032.hg.1 | ENST00000317764.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300010032.hg.1 | ENST00000317764.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300010032.hg.1 | ENST00000317764.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300010032.hg.1 | ENST00000317764.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300010032.hg.1 | ENST00000317764.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300010032.hg.1 | ENST00000379881.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300010032.hg.1 | ENST00000379881.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300010032.hg.1 | ENST00000379881.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300010032.hg.1 | ENST00000379881.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300010032.hg.1 | ENST00000379881.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300010032.hg.1 | ENST00000379881.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300010032.hg.1 | ENST00000379881.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300010032.hg.1 | ENST00000379881.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300010032.hg.1 | ENST00000379881.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300010032.hg.1 | ENST00000379881.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300010032.hg.1 | ENST00000379881.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300010032.hg.1 | ENST00000379881.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300010032.hg.1 | ENST00000379881.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300010032.hg.1 | NM_001282147.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300010032.hg.1 | NM_001282147.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300010032.hg.1 | NM_001282147.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300010032.hg.1 | NM_001282147.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300010032.hg.1 | NM_001282147.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300010032.hg.1 | NM_001282147.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300010032.hg.1 | NM_001282147.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300010032.hg.1 | NM_001282147.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300010032.hg.1 | NM_001282147.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300010032.hg.1 | NM_001282147.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300010032.hg.1 | NM_017826.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300010032.hg.1 | NM_017826.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300010032.hg.1 | NM_017826.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300010032.hg.1 | NM_017826.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300010032.hg.1 | NM_017826.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300010032.hg.1 | NM_017826.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300010032.hg.1 | NM_017826.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300010032.hg.1 | NM_017826.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300010032.hg.1 | NM_017826.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300010032.hg.1 | NM_017826.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300010032.hg.1 | NM_017826.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300010032.hg.1 | NM_017826.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300010032.hg.1 | NM_017826.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR13006360.hg.1 | TC13001724.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR13006360.hg.1 | ENST00000379881.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR13006360.hg.1 | NM_017826.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300010031.hg.1 | ENST00000511166.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300010031.hg.1 | ENST00000511166.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300010031.hg.1 | ENST00000511166.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300010031.hg.1 | ENST00000511166.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300010031.hg.1 | ENST00000511166.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300010031.hg.1 | ENST00000511166.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300010031.hg.1 | ENST00000511166.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300010031.hg.1 | ENST00000511166.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300010031.hg.1 | ENST00000511166.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300010031.hg.1 | ENST00000511166.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300010031.hg.1 | ENST00000511166.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300010031.hg.1 | ENST00000511166.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300010031.hg.1 | ENST00000511166.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300010031.hg.1 | ENST00000511166.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300010031.hg.1 | ENST00000511166.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300010031.hg.1 | ENST00000511166.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300010031.hg.1 | NM_001198910.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300010031.hg.1 | NM_001198910.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300010031.hg.1 | NM_001198910.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300010031.hg.1 | NM_001198910.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300010031.hg.1 | NM_001198910.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300010031.hg.1 | NM_001198910.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300010031.hg.1 | NM_001198910.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300010031.hg.1 | NM_001198910.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300010031.hg.1 | NM_001198910.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300010031.hg.1 | NM_001198910.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300010031.hg.1 | NM_001198910.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300010031.hg.1 | NM_001198910.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300010031.hg.1 | NM_001198910.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300010031.hg.1 | NM_001198910.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300010031.hg.1 | NM_001198910.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300010031.hg.1 | NM_001198910.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300010031.hg.1 | NM_001198910.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300010031.hg.1 | ENST00000511166.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300010031.hg.1 | ENST00000511166.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300010031.hg.1 | ENST00000511166.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300010031.hg.1 | ENST00000511166.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300010031.hg.1 | ENST00000511166.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300010031.hg.1 | ENST00000511166.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300010031.hg.1 | ENST00000511166.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300010031.hg.1 | ENST00000511166.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300010031.hg.1 | ENST00000511166.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300010031.hg.1 | ENST00000511166.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300010031.hg.1 | ENST00000511166.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300010031.hg.1 | ENST00000511166.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300010031.hg.1 | ENST00000511166.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300010031.hg.1 | ENST00000511166.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300010031.hg.1 | ENST00000511166.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300010031.hg.1 | ENST00000511166.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300010031.hg.1 | NM_001198910.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300010031.hg.1 | NM_001198910.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300010031.hg.1 | NM_001198910.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300010031.hg.1 | NM_001198910.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300010031.hg.1 | NM_001198910.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300010031.hg.1 | NM_001198910.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300010031.hg.1 | NM_001198910.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300010031.hg.1 | NM_001198910.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300010031.hg.1 | NM_001198910.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300010031.hg.1 | NM_001198910.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300010031.hg.1 | NM_001198910.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300010031.hg.1 | NM_001198910.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300010031.hg.1 | NM_001198910.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300010031.hg.1 | NM_001198910.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300010031.hg.1 | NM_001198910.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300010031.hg.1 | NM_001198910.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300010031.hg.1 | NM_001198910.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300167570.hg.1 | ENST00000511166.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300167570.hg.1 | NM_001198910.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300167570.hg.1 | ENST00000511166.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300167570.hg.1 | NM_001198910.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300167570.hg.1 | TC1300010031.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1300167589.hg.1 | ENST00000379881.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300167589.hg.1 | NM_017826.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300167589.hg.1 | ENST00000379881.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300167589.hg.1 | NM_017826.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300167589.hg.1 | TC1300010032.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1300010032.hg.1 | ENST00000317764.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300010032.hg.1 | ENST00000317764.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300010032.hg.1 | ENST00000317764.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300010032.hg.1 | ENST00000317764.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300010032.hg.1 | ENST00000317764.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300010032.hg.1 | ENST00000317764.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300010032.hg.1 | ENST00000317764.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300010032.hg.1 | ENST00000317764.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300010032.hg.1 | ENST00000317764.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300010032.hg.1 | ENST00000379881.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300010032.hg.1 | ENST00000379881.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300010032.hg.1 | ENST00000379881.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300010032.hg.1 | ENST00000379881.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300010032.hg.1 | ENST00000379881.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300010032.hg.1 | ENST00000379881.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300010032.hg.1 | ENST00000379881.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300010032.hg.1 | ENST00000379881.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300010032.hg.1 | ENST00000379881.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300010032.hg.1 | ENST00000379881.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300010032.hg.1 | ENST00000379881.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300010032.hg.1 | ENST00000379881.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300010032.hg.1 | ENST00000379881.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300010032.hg.1 | NM_001282147.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300010032.hg.1 | NM_001282147.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300010032.hg.1 | NM_001282147.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300010032.hg.1 | NM_001282147.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300010032.hg.1 | NM_001282147.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300010032.hg.1 | NM_001282147.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300010032.hg.1 | NM_001282147.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300010032.hg.1 | NM_001282147.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300010032.hg.1 | NM_001282147.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300010032.hg.1 | NM_001282147.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300010032.hg.1 | NM_017826.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300010032.hg.1 | NM_017826.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300010032.hg.1 | NM_017826.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300010032.hg.1 | NM_017826.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300010032.hg.1 | NM_017826.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300010032.hg.1 | NM_017826.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300010032.hg.1 | NM_017826.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300010032.hg.1 | NM_017826.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300010032.hg.1 | NM_017826.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300010032.hg.1 | NM_017826.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300010032.hg.1 | NM_017826.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300010032.hg.1 | NM_017826.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300010032.hg.1 | NM_017826.2 | Refseq |

CCDC169-SOHLH2
Genomic Map