Assay Details
| Gene Symbol: BNC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001717.3 | NP_001708.3 | 1 - 2 | 184 | 146 | |
| GenBank mRNA | L03427.1 | - | 180 | 143 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC15001767.hg.1
Additional Information:
This assay is in the 5'UTR region of the L03427.1,NM_001717.3 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1500010261.hg.1 | ENST00000345382.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010261.hg.1 | ENST00000345382.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010261.hg.1 | ENST00000345382.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010261.hg.1 | ENST00000345382.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010261.hg.1 | ENST00000345382.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010261.hg.1 | ENST00000345382.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010261.hg.1 | ENST00000345382.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010261.hg.1 | ENST00000345382.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010261.hg.1 | ENST00000569704.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010261.hg.1 | ENST00000569704.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010261.hg.1 | ENST00000569704.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010261.hg.1 | ENST00000569704.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010261.hg.1 | ENST00000569704.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010261.hg.1 | ENST00000569704.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010261.hg.1 | ENST00000569704.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010261.hg.1 | ENST00000618086.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010261.hg.1 | ENST00000618086.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010261.hg.1 | ENST00000618086.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010261.hg.1 | ENST00000618086.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010261.hg.1 | ENST00000618086.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010261.hg.1 | ENST00000618086.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010261.hg.1 | ENST00000618086.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010261.hg.1 | ENST00000618086.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010261.hg.1 | NM_001301206.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010261.hg.1 | NM_001301206.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010261.hg.1 | NM_001301206.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010261.hg.1 | NM_001301206.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010261.hg.1 | NM_001301206.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010261.hg.1 | NM_001301206.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010261.hg.1 | NM_001301206.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010261.hg.1 | NM_001301206.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010261.hg.1 | NM_001301206.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010261.hg.1 | NM_001717.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010261.hg.1 | NM_001717.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010261.hg.1 | NM_001717.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010261.hg.1 | NM_001717.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010261.hg.1 | NM_001717.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010261.hg.1 | NM_001717.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010261.hg.1 | NM_001717.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010261.hg.1 | NM_001717.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR15017344.hg.1 | ENST00000345382.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15017344.hg.1 | NM_001717.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR15017344.hg.1 | TC15001767.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR15017339.hg.1 | ENST00000345382.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15017339.hg.1 | ENST00000541809.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15017339.hg.1 | NM_001717.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR15017339.hg.1 | TC15001767.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR15017343.hg.1 | ENST00000345382.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15017343.hg.1 | NM_001717.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR15017343.hg.1 | TC15001767.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1500177259.hg.1 | ENST00000345382.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500177259.hg.1 | ENST00000618086.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500177259.hg.1 | NM_001717.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500177259.hg.1 | ENST00000345382.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500177259.hg.1 | ENST00000618086.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500177259.hg.1 | NM_001717.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500177259.hg.1 | TC1500010261.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1500177255.hg.1 | ENST00000345382.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500177255.hg.1 | ENST00000569704.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500177255.hg.1 | ENST00000618086.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500177255.hg.1 | NM_001301206.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500177255.hg.1 | NM_001717.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500177255.hg.1 | ENST00000345382.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500177255.hg.1 | ENST00000569704.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500177255.hg.1 | ENST00000618086.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500177255.hg.1 | NM_001301206.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500177255.hg.1 | NM_001717.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500177255.hg.1 | TC1500010261.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1500010261.hg.1 | ENST00000345382.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010261.hg.1 | ENST00000345382.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010261.hg.1 | ENST00000345382.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010261.hg.1 | ENST00000345382.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010261.hg.1 | ENST00000345382.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010261.hg.1 | ENST00000345382.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010261.hg.1 | ENST00000345382.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010261.hg.1 | ENST00000345382.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010261.hg.1 | ENST00000569704.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010261.hg.1 | ENST00000569704.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010261.hg.1 | ENST00000569704.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010261.hg.1 | ENST00000569704.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010261.hg.1 | ENST00000569704.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010261.hg.1 | ENST00000569704.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010261.hg.1 | ENST00000569704.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010261.hg.1 | ENST00000618086.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010261.hg.1 | ENST00000618086.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010261.hg.1 | ENST00000618086.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010261.hg.1 | ENST00000618086.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010261.hg.1 | ENST00000618086.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010261.hg.1 | ENST00000618086.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010261.hg.1 | ENST00000618086.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010261.hg.1 | ENST00000618086.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010261.hg.1 | NM_001301206.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010261.hg.1 | NM_001301206.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010261.hg.1 | NM_001301206.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010261.hg.1 | NM_001301206.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010261.hg.1 | NM_001301206.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010261.hg.1 | NM_001301206.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010261.hg.1 | NM_001301206.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010261.hg.1 | NM_001301206.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010261.hg.1 | NM_001301206.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010261.hg.1 | NM_001717.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010261.hg.1 | NM_001717.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010261.hg.1 | NM_001717.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010261.hg.1 | NM_001717.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010261.hg.1 | NM_001717.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010261.hg.1 | NM_001717.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010261.hg.1 | NM_001717.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010261.hg.1 | NM_001717.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500177260.hg.1 | ENST00000345382.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500177260.hg.1 | ENST00000618086.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500177260.hg.1 | NM_001717.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500177260.hg.1 | ENST00000345382.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500177260.hg.1 | ENST00000618086.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500177260.hg.1 | NM_001717.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500177260.hg.1 | TC1500010261.hg.1 | Clariom D TC |

BNC1
Genomic Map