Assay Details
| Gene Symbol: MUC15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001135091.1 | NP_001128563.1 | 3 - 4 | 1048 | 111 | |
| NM_145650.3 | NP_663625.2 | 2 - 3 | 960 | 111 | ||
| XM_017017235.1 | XP_016872724.1 | 3 - 4 | 1048 | 111 | ||
| GenBank mRNA | AJ417818.1 | - | 3 - 4 | 916 | 111 | |
| AK128337.1 | - | 2 - 3 | 813 | 111 | ||
| AY358668.1 | - | 3 - 4 | 1013 | 111 | ||
| BC020912.1 | - | 3 - 4 | 1040 | 4733074 | 111 | |
| BC058007.1 | - | 3 - 4 | 926 | 4719841 | 111 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC11001505.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1100010391.hg.1 | ENST00000281268.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010391.hg.1 | ENST00000281268.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010391.hg.1 | ENST00000281268.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010391.hg.1 | ENST00000281268.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010391.hg.1 | ENST00000281268.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010391.hg.1 | ENST00000281268.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010391.hg.1 | ENST00000281268.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010391.hg.1 | ENST00000281268.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010391.hg.1 | ENST00000436318.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010391.hg.1 | ENST00000436318.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010391.hg.1 | ENST00000436318.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010391.hg.1 | ENST00000436318.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010391.hg.1 | ENST00000436318.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010391.hg.1 | ENST00000436318.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010391.hg.1 | ENST00000436318.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010391.hg.1 | ENST00000436318.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010391.hg.1 | ENST00000436318.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010391.hg.1 | ENST00000455601.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010391.hg.1 | ENST00000455601.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010391.hg.1 | ENST00000455601.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010391.hg.1 | ENST00000455601.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010391.hg.1 | ENST00000455601.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010391.hg.1 | ENST00000455601.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010391.hg.1 | ENST00000455601.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010391.hg.1 | ENST00000455601.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010391.hg.1 | ENST00000455601.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010391.hg.1 | ENST00000527569.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010391.hg.1 | ENST00000527569.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010391.hg.1 | ENST00000527569.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010391.hg.1 | ENST00000527569.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010391.hg.1 | ENST00000527569.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010391.hg.1 | ENST00000527569.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010391.hg.1 | ENST00000527569.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010391.hg.1 | ENST00000529533.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010391.hg.1 | ENST00000529533.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010391.hg.1 | ENST00000529533.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010391.hg.1 | ENST00000529533.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010391.hg.1 | ENST00000529533.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010391.hg.1 | ENST00000529533.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010391.hg.1 | ENST00000529533.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010391.hg.1 | ENST00000529533.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010391.hg.1 | ENST00000529533.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010391.hg.1 | ENST00000529533.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010391.hg.1 | ENST00000529533.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010391.hg.1 | NM_001135091.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010391.hg.1 | NM_001135091.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010391.hg.1 | NM_001135091.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010391.hg.1 | NM_001135091.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010391.hg.1 | NM_001135091.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010391.hg.1 | NM_001135091.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010391.hg.1 | NM_001135091.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010391.hg.1 | NM_001135091.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010391.hg.1 | NM_001135091.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010391.hg.1 | NM_001135091.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010391.hg.1 | NM_001135091.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010391.hg.1 | NM_001135091.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010391.hg.1 | NM_001135091.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010391.hg.1 | NM_001135092.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010391.hg.1 | NM_001135092.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010391.hg.1 | NM_001135092.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010391.hg.1 | NM_001135092.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010391.hg.1 | NM_001135092.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010391.hg.1 | NM_001135092.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010391.hg.1 | NM_001135092.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010391.hg.1 | NM_001135092.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010391.hg.1 | NM_001135092.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010391.hg.1 | NM_001135092.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010391.hg.1 | NM_001135092.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010391.hg.1 | NM_001135092.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010391.hg.1 | NM_145650.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010391.hg.1 | NM_145650.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010391.hg.1 | NM_145650.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010391.hg.1 | NM_145650.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010391.hg.1 | NM_145650.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010391.hg.1 | NM_145650.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010391.hg.1 | NM_145650.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010391.hg.1 | NM_145650.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010391.hg.1 | NM_145650.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010391.hg.1 | NM_145650.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010391.hg.1 | NM_145650.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11018826.hg.1 | ENST00000436318.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11018826.hg.1 | ENST00000455601.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11018826.hg.1 | ENST00000529533.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11018826.hg.1 | NM_001135091.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11018826.hg.1 | NM_145650.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11018826.hg.1 | TC11001505.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1100178742.hg.1 | ENST00000436318.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100178742.hg.1 | ENST00000455601.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100178742.hg.1 | ENST00000529533.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100178742.hg.1 | NM_001135091.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100178742.hg.1 | NM_145650.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100178742.hg.1 | ENST00000436318.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100178742.hg.1 | ENST00000455601.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100178742.hg.1 | ENST00000529533.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100178742.hg.1 | NM_001135091.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100178742.hg.1 | NM_145650.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100178742.hg.1 | TC1100010391.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1100010391.hg.1 | ENST00000281268.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010391.hg.1 | ENST00000281268.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010391.hg.1 | ENST00000281268.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010391.hg.1 | ENST00000281268.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010391.hg.1 | ENST00000281268.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010391.hg.1 | ENST00000281268.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010391.hg.1 | ENST00000281268.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010391.hg.1 | ENST00000281268.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010391.hg.1 | ENST00000436318.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010391.hg.1 | ENST00000436318.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010391.hg.1 | ENST00000436318.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010391.hg.1 | ENST00000436318.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010391.hg.1 | ENST00000436318.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010391.hg.1 | ENST00000436318.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010391.hg.1 | ENST00000436318.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010391.hg.1 | ENST00000436318.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010391.hg.1 | ENST00000436318.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010391.hg.1 | ENST00000455601.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010391.hg.1 | ENST00000455601.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010391.hg.1 | ENST00000455601.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010391.hg.1 | ENST00000455601.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010391.hg.1 | ENST00000455601.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010391.hg.1 | ENST00000455601.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010391.hg.1 | ENST00000455601.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010391.hg.1 | ENST00000455601.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010391.hg.1 | ENST00000455601.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010391.hg.1 | ENST00000527569.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010391.hg.1 | ENST00000527569.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010391.hg.1 | ENST00000527569.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010391.hg.1 | ENST00000527569.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010391.hg.1 | ENST00000527569.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010391.hg.1 | ENST00000527569.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010391.hg.1 | ENST00000527569.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010391.hg.1 | ENST00000529533.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010391.hg.1 | ENST00000529533.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010391.hg.1 | ENST00000529533.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010391.hg.1 | ENST00000529533.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010391.hg.1 | ENST00000529533.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010391.hg.1 | ENST00000529533.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010391.hg.1 | ENST00000529533.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010391.hg.1 | ENST00000529533.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010391.hg.1 | ENST00000529533.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010391.hg.1 | ENST00000529533.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010391.hg.1 | ENST00000529533.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010391.hg.1 | NM_001135091.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010391.hg.1 | NM_001135091.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010391.hg.1 | NM_001135091.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010391.hg.1 | NM_001135091.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010391.hg.1 | NM_001135091.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010391.hg.1 | NM_001135091.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010391.hg.1 | NM_001135091.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010391.hg.1 | NM_001135091.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010391.hg.1 | NM_001135091.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010391.hg.1 | NM_001135091.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010391.hg.1 | NM_001135091.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010391.hg.1 | NM_001135091.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010391.hg.1 | NM_001135091.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010391.hg.1 | NM_001135092.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010391.hg.1 | NM_001135092.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010391.hg.1 | NM_001135092.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010391.hg.1 | NM_001135092.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010391.hg.1 | NM_001135092.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010391.hg.1 | NM_001135092.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010391.hg.1 | NM_001135092.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010391.hg.1 | NM_001135092.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010391.hg.1 | NM_001135092.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010391.hg.1 | NM_001135092.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010391.hg.1 | NM_001135092.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010391.hg.1 | NM_001135092.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010391.hg.1 | NM_145650.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010391.hg.1 | NM_145650.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010391.hg.1 | NM_145650.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010391.hg.1 | NM_145650.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010391.hg.1 | NM_145650.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010391.hg.1 | NM_145650.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010391.hg.1 | NM_145650.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010391.hg.1 | NM_145650.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010391.hg.1 | NM_145650.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010391.hg.1 | NM_145650.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010391.hg.1 | NM_145650.3 | Refseq |

MUC15
Genomic Map