Assay Details
| Gene Symbol: RBM12B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005250915.4 | XP_005250972.1 | 3 - 4 | 246 | 148 | |
| XM_017013406.1 | XP_016868895.1 | 2 - 3 | 429 | 148 | ||
| XM_017013407.1 | XP_016868896.1 | 4 - 5 | 301 | 210 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005250915.4, XM_017013406.1, XM_017013407.1 transcripts.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0800011116.hg.1 | ENST00000399300.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011116.hg.1 | ENST00000399300.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011116.hg.1 | ENST00000399300.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011116.hg.1 | ENST00000399300.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011116.hg.1 | ENST00000399300.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011116.hg.1 | ENST00000399300.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011116.hg.1 | ENST00000399300.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011116.hg.1 | ENST00000399300.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011116.hg.1 | ENST00000399300.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011116.hg.1 | ENST00000399300.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011116.hg.1 | ENST00000399300.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011116.hg.1 | ENST00000399300.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011116.hg.1 | ENST00000399300.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011116.hg.1 | ENST00000399300.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011116.hg.1 | ENST00000399300.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011116.hg.1 | ENST00000517700.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011116.hg.1 | ENST00000517700.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011116.hg.1 | ENST00000517700.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011116.hg.1 | ENST00000517700.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011116.hg.1 | ENST00000517700.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011116.hg.1 | ENST00000517700.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011116.hg.1 | ENST00000517700.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011116.hg.1 | ENST00000517700.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011116.hg.1 | ENST00000517700.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011116.hg.1 | ENST00000517700.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011116.hg.1 | ENST00000517700.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011116.hg.1 | ENST00000517700.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011116.hg.1 | ENST00000517700.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011116.hg.1 | ENST00000518597.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011116.hg.1 | ENST00000518597.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011116.hg.1 | ENST00000518597.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011116.hg.1 | ENST00000518597.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011116.hg.1 | ENST00000518597.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011116.hg.1 | ENST00000518597.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011116.hg.1 | ENST00000519109.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011116.hg.1 | ENST00000519109.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011116.hg.1 | ENST00000519109.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011116.hg.1 | ENST00000519109.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011116.hg.1 | ENST00000519109.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011116.hg.1 | ENST00000519109.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011116.hg.1 | ENST00000520560.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011116.hg.1 | ENST00000520560.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011116.hg.1 | ENST00000520560.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011116.hg.1 | ENST00000520560.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011116.hg.1 | ENST00000520560.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011116.hg.1 | ENST00000520560.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011116.hg.1 | ENST00000520961.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011116.hg.1 | ENST00000520961.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011116.hg.1 | ENST00000521947.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011116.hg.1 | ENST00000521947.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011116.hg.1 | ENST00000521947.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011116.hg.1 | ENST00000521947.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011116.hg.1 | ENST00000521947.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011116.hg.1 | ENST00000627241.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011116.hg.1 | ENST00000627241.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011116.hg.1 | ENST00000627241.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011116.hg.1 | ENST00000627241.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011116.hg.1 | ENST00000627241.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011116.hg.1 | NM_203390.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011116.hg.1 | NM_203390.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011116.hg.1 | NM_203390.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011116.hg.1 | NM_203390.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011116.hg.1 | NM_203390.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011116.hg.1 | NM_203390.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011116.hg.1 | NM_203390.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011116.hg.1 | NM_203390.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011116.hg.1 | NM_203390.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011116.hg.1 | NM_203390.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011116.hg.1 | NM_203390.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011116.hg.1 | ENST00000399300.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011116.hg.1 | ENST00000399300.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011116.hg.1 | ENST00000399300.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011116.hg.1 | ENST00000399300.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011116.hg.1 | ENST00000399300.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011116.hg.1 | ENST00000399300.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011116.hg.1 | ENST00000399300.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011116.hg.1 | ENST00000399300.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011116.hg.1 | ENST00000399300.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011116.hg.1 | ENST00000399300.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011116.hg.1 | ENST00000399300.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011116.hg.1 | ENST00000399300.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011116.hg.1 | ENST00000399300.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011116.hg.1 | ENST00000399300.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011116.hg.1 | ENST00000399300.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011116.hg.1 | ENST00000517700.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011116.hg.1 | ENST00000517700.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011116.hg.1 | ENST00000517700.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011116.hg.1 | ENST00000517700.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011116.hg.1 | ENST00000517700.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011116.hg.1 | ENST00000517700.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011116.hg.1 | ENST00000517700.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011116.hg.1 | ENST00000517700.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011116.hg.1 | ENST00000517700.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011116.hg.1 | ENST00000517700.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011116.hg.1 | ENST00000517700.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011116.hg.1 | ENST00000517700.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011116.hg.1 | ENST00000517700.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011116.hg.1 | ENST00000518597.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011116.hg.1 | ENST00000518597.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011116.hg.1 | ENST00000518597.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011116.hg.1 | ENST00000518597.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011116.hg.1 | ENST00000518597.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011116.hg.1 | ENST00000518597.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011116.hg.1 | ENST00000519109.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011116.hg.1 | ENST00000519109.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011116.hg.1 | ENST00000519109.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011116.hg.1 | ENST00000519109.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011116.hg.1 | ENST00000519109.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011116.hg.1 | ENST00000519109.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011116.hg.1 | ENST00000520560.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011116.hg.1 | ENST00000520560.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011116.hg.1 | ENST00000520560.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011116.hg.1 | ENST00000520560.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011116.hg.1 | ENST00000520560.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011116.hg.1 | ENST00000520560.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011116.hg.1 | ENST00000520961.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011116.hg.1 | ENST00000520961.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011116.hg.1 | ENST00000521947.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011116.hg.1 | ENST00000521947.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011116.hg.1 | ENST00000521947.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011116.hg.1 | ENST00000521947.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011116.hg.1 | ENST00000521947.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011116.hg.1 | ENST00000627241.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011116.hg.1 | ENST00000627241.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011116.hg.1 | ENST00000627241.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011116.hg.1 | ENST00000627241.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011116.hg.1 | ENST00000627241.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011116.hg.1 | NM_203390.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011116.hg.1 | NM_203390.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011116.hg.1 | NM_203390.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011116.hg.1 | NM_203390.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011116.hg.1 | NM_203390.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011116.hg.1 | NM_203390.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011116.hg.1 | NM_203390.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011116.hg.1 | NM_203390.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011116.hg.1 | NM_203390.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011116.hg.1 | NM_203390.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011116.hg.1 | NM_203390.2 | Refseq |

RBM12B
Genomic Map