Assay Details
| Gene Symbol: KLRA1P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_028045.1 | 1 - 2 | 312 | 147 | ||
| GenBank mRNA | AK304677.1 | - | 1 - 2 | 98 | 147 | |
| AK310151.1 | - | 1 - 2 | 146 | 147 | ||
| DA776936.1 | - | 1 - 2 | 312 | 147 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC12001224.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK304677.1 transcripts.
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR12016151.hg.1 | ENST00000510134.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12016151.hg.1 | NR_028045.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12016151.hg.1 | TC12001224.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR12016152.hg.1 | ENST00000510134.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12016152.hg.1 | NR_028045.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12016152.hg.1 | TC12001224.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR12016149.hg.1 | ENST00000510134.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12016149.hg.1 | NR_028045.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12016149.hg.1 | TC12001224.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR12016150.hg.1 | NR_028045.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12016150.hg.1 | TC12001224.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009910.hg.1 | ENST00000503499.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009910.hg.1 | ENST00000503499.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009910.hg.1 | ENST00000503499.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009910.hg.1 | ENST00000503499.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009910.hg.1 | ENST00000503499.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009910.hg.1 | ENST00000503499.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009910.hg.1 | ENST00000503499.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009910.hg.1 | ENST00000510134.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009910.hg.1 | ENST00000510134.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009910.hg.1 | ENST00000510134.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009910.hg.1 | ENST00000510134.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009910.hg.1 | ENST00000510134.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009910.hg.1 | ENST00000510134.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009910.hg.1 | ENST00000510134.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009910.hg.1 | ENST00000510134.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009910.hg.1 | ENST00000510134.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009910.hg.1 | ENST00000535939.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009910.hg.1 | ENST00000535939.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009910.hg.1 | ENST00000535939.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009910.hg.1 | ENST00000535939.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009910.hg.1 | ENST00000535939.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009910.hg.1 | ENST00000535939.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009910.hg.1 | ENST00000535939.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009910.hg.1 | ENST00000535939.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009910.hg.1 | ENST00000535939.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009910.hg.1 | ENST00000535939.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009910.hg.1 | ENST00000535939.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009910.hg.1 | NR_028045.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009910.hg.1 | NR_028045.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009910.hg.1 | NR_028045.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009910.hg.1 | NR_028045.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009910.hg.1 | NR_028045.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009910.hg.1 | NR_028045.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009910.hg.1 | NR_028045.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009910.hg.1 | NR_028045.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009910.hg.1 | NR_028045.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009910.hg.1 | NR_028045.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009910.hg.1 | NR_028045.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009910.hg.1 | NR_028045.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009910.hg.1 | NR_028045.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12016145.hg.1 | ENST00000510134.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12016145.hg.1 | NR_028045.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12016145.hg.1 | TC12001224.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1200174765.hg.1 | ENST00000503499.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200174765.hg.1 | ENST00000510134.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200174765.hg.1 | ENST00000535939.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200174765.hg.1 | NR_028045.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200174765.hg.1 | ENST00000503499.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200174765.hg.1 | ENST00000510134.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200174765.hg.1 | ENST00000535939.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200174765.hg.1 | NR_028045.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200174765.hg.1 | TC1200009910.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1200174772.hg.1 | ENST00000510134.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200174772.hg.1 | NR_028045.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200174772.hg.1 | ENST00000510134.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200174772.hg.1 | NR_028045.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200174772.hg.1 | TC1200009910.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1200174774.hg.1 | NR_028045.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200174774.hg.1 | NR_028045.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200174774.hg.1 | TC1200009910.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1200174773.hg.1 | ENST00000510134.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200174773.hg.1 | NR_028045.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200174773.hg.1 | ENST00000510134.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200174773.hg.1 | NR_028045.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200174773.hg.1 | TC1200009910.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1200009910.hg.1 | ENST00000503499.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009910.hg.1 | ENST00000503499.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009910.hg.1 | ENST00000503499.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009910.hg.1 | ENST00000503499.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009910.hg.1 | ENST00000503499.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009910.hg.1 | ENST00000503499.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009910.hg.1 | ENST00000503499.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009910.hg.1 | ENST00000510134.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009910.hg.1 | ENST00000510134.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009910.hg.1 | ENST00000510134.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009910.hg.1 | ENST00000510134.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009910.hg.1 | ENST00000510134.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009910.hg.1 | ENST00000510134.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009910.hg.1 | ENST00000510134.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009910.hg.1 | ENST00000510134.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009910.hg.1 | ENST00000510134.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009910.hg.1 | ENST00000535939.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009910.hg.1 | ENST00000535939.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009910.hg.1 | ENST00000535939.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009910.hg.1 | ENST00000535939.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009910.hg.1 | ENST00000535939.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009910.hg.1 | ENST00000535939.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009910.hg.1 | ENST00000535939.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009910.hg.1 | ENST00000535939.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009910.hg.1 | ENST00000535939.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009910.hg.1 | ENST00000535939.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009910.hg.1 | ENST00000535939.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009910.hg.1 | NR_028045.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009910.hg.1 | NR_028045.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009910.hg.1 | NR_028045.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009910.hg.1 | NR_028045.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009910.hg.1 | NR_028045.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009910.hg.1 | NR_028045.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009910.hg.1 | NR_028045.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009910.hg.1 | NR_028045.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009910.hg.1 | NR_028045.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009910.hg.1 | NR_028045.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009910.hg.1 | NR_028045.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009910.hg.1 | NR_028045.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009910.hg.1 | NR_028045.1 | Refseq |

KLRA1P
Genomic Map