Assay Details
| Gene Symbol: PKIA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181839.2 | NP_862822.1 | 1 - 2 | 370 | 70 | |
| GenBank mRNA | DB480496.1 | - | 1 - 2 | 152 | 70 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC08000505.hg.1
Additional Information:
This assay is in the 5'UTR region of the NM_181839.2 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
regulation of G2/M transition of mitotic cell cycle
negative regulation of protein import into nucleus
negative regulation of cAMP-dependent protein kinase activity
cAMP-dependent protein kinase inhibitor activity
protein binding
protein kinase A catalytic subunit binding
negative regulation of transcription from RNA polymerase II promoter
regulation of G2/M transition of mitotic cell cycle
negative regulation of protein import into nucleus
negative regulation of cAMP-dependent protein kinase activity
cAMP-dependent protein kinase inhibitor activity
protein binding
protein kinase A catalytic subunit binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0800008028.hg.1 | ENST00000352966.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008028.hg.1 | ENST00000352966.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008028.hg.1 | ENST00000352966.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008028.hg.1 | ENST00000352966.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008028.hg.1 | ENST00000352966.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008028.hg.1 | ENST00000352966.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008028.hg.1 | ENST00000352966.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008028.hg.1 | ENST00000352966.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008028.hg.1 | ENST00000396418.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008028.hg.1 | ENST00000396418.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008028.hg.1 | ENST00000396418.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008028.hg.1 | ENST00000396418.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008028.hg.1 | ENST00000396418.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008028.hg.1 | ENST00000396418.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008028.hg.1 | ENST00000396418.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008028.hg.1 | ENST00000396418.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008028.hg.1 | ENST00000396418.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008028.hg.1 | ENST00000396418.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008028.hg.1 | ENST00000396418.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008028.hg.1 | ENST00000396418.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008028.hg.1 | ENST00000518467.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008028.hg.1 | ENST00000518467.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008028.hg.1 | ENST00000518467.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008028.hg.1 | ENST00000518467.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008028.hg.1 | ENST00000518467.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008028.hg.1 | NM_006823.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008028.hg.1 | NM_006823.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008028.hg.1 | NM_006823.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008028.hg.1 | NM_006823.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008028.hg.1 | NM_006823.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008028.hg.1 | NM_006823.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008028.hg.1 | NM_006823.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008028.hg.1 | NM_006823.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008028.hg.1 | NM_006823.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008028.hg.1 | NM_006823.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008028.hg.1 | NM_006823.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008028.hg.1 | NM_006823.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008028.hg.1 | NM_006823.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008028.hg.1 | NM_181839.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008028.hg.1 | NM_181839.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008028.hg.1 | NM_181839.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008028.hg.1 | NM_181839.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008028.hg.1 | NM_181839.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008028.hg.1 | NM_181839.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008028.hg.1 | NM_181839.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008028.hg.1 | NM_181839.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008028.hg.1 | NM_181839.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008028.hg.1 | NM_181839.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008028.hg.1 | NM_181839.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008028.hg.1 | NM_181839.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08006906.hg.1 | TC08000505.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR08006906.hg.1 | NM_006823.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08006906.hg.1 | NM_181839.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08006908.hg.1 | TC08000505.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR08006908.hg.1 | ENST00000352966.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08006908.hg.1 | ENST00000396418.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08006908.hg.1 | NM_006823.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08006908.hg.1 | NM_181839.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08006912.hg.1 | TC08000505.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR08006912.hg.1 | ENST00000352966.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08006912.hg.1 | ENST00000396418.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08006912.hg.1 | ENST00000518467.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08006912.hg.1 | NM_006823.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08006912.hg.1 | NM_181839.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08006907.hg.1 | TC08000505.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR08006907.hg.1 | ENST00000396418.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08006907.hg.1 | NM_006823.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08006907.hg.1 | NM_181839.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800155281.hg.1 | NM_006823.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800155281.hg.1 | NM_181839.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800155281.hg.1 | NM_006823.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800155281.hg.1 | NM_181839.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800155281.hg.1 | TC0800008028.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0800155282.hg.1 | ENST00000396418.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800155282.hg.1 | NM_006823.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800155282.hg.1 | NM_181839.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800155282.hg.1 | ENST00000396418.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800155282.hg.1 | NM_006823.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800155282.hg.1 | NM_181839.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800155282.hg.1 | TC0800008028.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0800155283.hg.1 | ENST00000352966.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800155283.hg.1 | ENST00000396418.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800155283.hg.1 | NM_006823.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800155283.hg.1 | NM_181839.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800155283.hg.1 | ENST00000352966.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800155283.hg.1 | ENST00000396418.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800155283.hg.1 | NM_006823.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800155283.hg.1 | NM_181839.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800155283.hg.1 | TC0800008028.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0800155288.hg.1 | ENST00000352966.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800155288.hg.1 | ENST00000396418.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800155288.hg.1 | ENST00000518467.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800155288.hg.1 | NM_006823.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800155288.hg.1 | NM_181839.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800155288.hg.1 | ENST00000352966.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800155288.hg.1 | ENST00000396418.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800155288.hg.1 | ENST00000518467.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800155288.hg.1 | NM_006823.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800155288.hg.1 | NM_181839.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800155288.hg.1 | TC0800008028.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0800155287.hg.1 | ENST00000352966.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800155287.hg.1 | ENST00000396418.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800155287.hg.1 | ENST00000518467.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800155287.hg.1 | NM_006823.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800155287.hg.1 | NM_181839.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800155287.hg.1 | ENST00000352966.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800155287.hg.1 | ENST00000396418.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800155287.hg.1 | ENST00000518467.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800155287.hg.1 | NM_006823.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800155287.hg.1 | NM_181839.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800155287.hg.1 | TC0800008028.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0800008028.hg.1 | ENST00000352966.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008028.hg.1 | ENST00000352966.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008028.hg.1 | ENST00000352966.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008028.hg.1 | ENST00000352966.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008028.hg.1 | ENST00000352966.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008028.hg.1 | ENST00000352966.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008028.hg.1 | ENST00000352966.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008028.hg.1 | ENST00000352966.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008028.hg.1 | ENST00000396418.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008028.hg.1 | ENST00000396418.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008028.hg.1 | ENST00000396418.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008028.hg.1 | ENST00000396418.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008028.hg.1 | ENST00000396418.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008028.hg.1 | ENST00000396418.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008028.hg.1 | ENST00000396418.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008028.hg.1 | ENST00000396418.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008028.hg.1 | ENST00000396418.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008028.hg.1 | ENST00000396418.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008028.hg.1 | ENST00000396418.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008028.hg.1 | ENST00000396418.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008028.hg.1 | ENST00000518467.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008028.hg.1 | ENST00000518467.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008028.hg.1 | ENST00000518467.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008028.hg.1 | ENST00000518467.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008028.hg.1 | ENST00000518467.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008028.hg.1 | NM_006823.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008028.hg.1 | NM_006823.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008028.hg.1 | NM_006823.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008028.hg.1 | NM_006823.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008028.hg.1 | NM_006823.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008028.hg.1 | NM_006823.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008028.hg.1 | NM_006823.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008028.hg.1 | NM_006823.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008028.hg.1 | NM_006823.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008028.hg.1 | NM_006823.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008028.hg.1 | NM_006823.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008028.hg.1 | NM_006823.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008028.hg.1 | NM_006823.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008028.hg.1 | NM_181839.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008028.hg.1 | NM_181839.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008028.hg.1 | NM_181839.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008028.hg.1 | NM_181839.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008028.hg.1 | NM_181839.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008028.hg.1 | NM_181839.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008028.hg.1 | NM_181839.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008028.hg.1 | NM_181839.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008028.hg.1 | NM_181839.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008028.hg.1 | NM_181839.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008028.hg.1 | NM_181839.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008028.hg.1 | NM_181839.2 | Refseq |

PKIA
Genomic Map