Assay Details
| Gene Symbol: CRHBP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001882.3 | NP_001873.2 | 1 - 2 | 357 | 84 | |
| GenBank mRNA | AK223457.1 | - | 1 - 2 | 234 | 84 | |
| AK307729.1 | - | 1 - 2 | 167 | 84 | ||
| AK311052.1 | - | 1 - 2 | 163 | 84 | ||
| BC018038.1 | - | 1 - 2 | 167 | 4812893 | 84 | |
| CB959505.1 | - | 1 - 2 | 384 | 84 | ||
| CR541666.1 | - | 1 - 2 | 77 | 84 | ||
| X58022.1 | - | 1 - 2 | 123 | 84 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC05000375.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
synaptic transmission, dopaminergic
maternal aggressive behavior
inflammatory response
signal transduction
female pregnancy
learning or memory
hormone-mediated signaling pathway
cellular response to stress
cellular response to drug
cellular response to potassium ion
hormone metabolic process
regulated exocytosis
behavioral response to ethanol
regulation of corticotropin secretion
negative regulation of corticotropin secretion
cellular response to calcium ion
cellular response to cocaine
cellular response to cAMP
cellular response to tumor necrosis factor
cellular response to estrogen stimulus
cellular response to estradiol stimulus
cellular response to gonadotropin-releasing hormone
negative regulation of corticotropin-releasing hormone receptor activity
regulation of N-methyl-D-aspartate selective glutamate receptor activity
protein binding
peptide binding
corticotropin-releasing hormone binding
synaptic transmission, dopaminergic
maternal aggressive behavior
inflammatory response
signal transduction
female pregnancy
learning or memory
hormone-mediated signaling pathway
cellular response to stress
cellular response to drug
cellular response to potassium ion
hormone metabolic process
regulated exocytosis
behavioral response to ethanol
regulation of corticotropin secretion
negative regulation of corticotropin secretion
cellular response to calcium ion
cellular response to cocaine
cellular response to cAMP
cellular response to tumor necrosis factor
cellular response to estrogen stimulus
cellular response to estradiol stimulus
cellular response to gonadotropin-releasing hormone
negative regulation of corticotropin-releasing hormone receptor activity
regulation of N-methyl-D-aspartate selective glutamate receptor activity
protein binding
peptide binding
corticotropin-releasing hormone binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR05005243.hg.1 | TC05000375.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR05005243.hg.1 | ENST00000274368.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05005243.hg.1 | ENST00000506501.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05005243.hg.1 | NM_001882.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013193.hg.1 | ENST00000274368.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013193.hg.1 | ENST00000274368.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013193.hg.1 | ENST00000274368.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013193.hg.1 | ENST00000274368.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013193.hg.1 | ENST00000274368.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013193.hg.1 | ENST00000274368.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013193.hg.1 | ENST00000274368.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013193.hg.1 | ENST00000274368.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013193.hg.1 | ENST00000274368.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013193.hg.1 | ENST00000274368.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013193.hg.1 | ENST00000274368.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013193.hg.1 | ENST00000274368.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013193.hg.1 | ENST00000274368.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013193.hg.1 | ENST00000503763.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013193.hg.1 | ENST00000503763.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013193.hg.1 | ENST00000503763.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013193.hg.1 | ENST00000503763.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013193.hg.1 | ENST00000506501.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013193.hg.1 | ENST00000506501.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013193.hg.1 | ENST00000506501.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013193.hg.1 | ENST00000506501.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013193.hg.1 | ENST00000506501.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013193.hg.1 | ENST00000506501.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013193.hg.1 | ENST00000506501.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013193.hg.1 | ENST00000506501.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013193.hg.1 | ENST00000512446.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013193.hg.1 | ENST00000512446.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013193.hg.1 | ENST00000512446.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013193.hg.1 | ENST00000512446.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013193.hg.1 | ENST00000512446.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013193.hg.1 | NM_001882.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013193.hg.1 | NM_001882.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013193.hg.1 | NM_001882.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013193.hg.1 | NM_001882.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013193.hg.1 | NM_001882.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013193.hg.1 | NM_001882.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013193.hg.1 | NM_001882.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013193.hg.1 | NM_001882.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013193.hg.1 | NM_001882.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013193.hg.1 | NM_001882.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013193.hg.1 | NM_001882.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013193.hg.1 | NM_001882.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05005241.hg.1 | TC05000375.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR05005241.hg.1 | ENST00000274368.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05005241.hg.1 | NM_001882.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05005245.hg.1 | TC05000375.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR05005245.hg.1 | ENST00000274368.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05005245.hg.1 | ENST00000506501.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05005245.hg.1 | NM_001882.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500193384.hg.1 | ENST00000274368.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500193384.hg.1 | ENST00000506501.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500193384.hg.1 | NM_001882.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500193384.hg.1 | ENST00000274368.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500193384.hg.1 | ENST00000506501.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500193384.hg.1 | NM_001882.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500193384.hg.1 | TC0500013193.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0500193385.hg.1 | ENST00000274368.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500193385.hg.1 | ENST00000506501.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500193385.hg.1 | NM_001882.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500193385.hg.1 | ENST00000274368.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500193385.hg.1 | ENST00000506501.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500193385.hg.1 | NM_001882.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500193385.hg.1 | TC0500013193.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0500193382.hg.1 | ENST00000274368.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500193382.hg.1 | NM_001882.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500193382.hg.1 | ENST00000274368.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500193382.hg.1 | NM_001882.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500193382.hg.1 | TC0500013193.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0500193387.hg.1 | ENST00000274368.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500193387.hg.1 | ENST00000506501.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500193387.hg.1 | ENST00000512446.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500193387.hg.1 | NM_001882.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500193387.hg.1 | ENST00000274368.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500193387.hg.1 | ENST00000506501.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500193387.hg.1 | ENST00000512446.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500193387.hg.1 | NM_001882.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500193387.hg.1 | TC0500013193.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0500013193.hg.1 | ENST00000274368.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013193.hg.1 | ENST00000274368.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013193.hg.1 | ENST00000274368.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013193.hg.1 | ENST00000274368.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013193.hg.1 | ENST00000274368.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013193.hg.1 | ENST00000274368.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013193.hg.1 | ENST00000274368.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013193.hg.1 | ENST00000274368.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013193.hg.1 | ENST00000274368.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013193.hg.1 | ENST00000274368.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013193.hg.1 | ENST00000274368.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013193.hg.1 | ENST00000274368.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013193.hg.1 | ENST00000274368.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013193.hg.1 | ENST00000503763.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013193.hg.1 | ENST00000503763.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013193.hg.1 | ENST00000503763.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013193.hg.1 | ENST00000503763.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013193.hg.1 | ENST00000506501.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013193.hg.1 | ENST00000506501.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013193.hg.1 | ENST00000506501.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013193.hg.1 | ENST00000506501.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013193.hg.1 | ENST00000506501.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013193.hg.1 | ENST00000506501.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013193.hg.1 | ENST00000506501.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013193.hg.1 | ENST00000506501.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013193.hg.1 | ENST00000512446.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013193.hg.1 | ENST00000512446.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013193.hg.1 | ENST00000512446.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013193.hg.1 | ENST00000512446.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013193.hg.1 | ENST00000512446.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013193.hg.1 | NM_001882.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013193.hg.1 | NM_001882.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013193.hg.1 | NM_001882.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013193.hg.1 | NM_001882.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013193.hg.1 | NM_001882.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013193.hg.1 | NM_001882.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013193.hg.1 | NM_001882.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013193.hg.1 | NM_001882.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013193.hg.1 | NM_001882.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013193.hg.1 | NM_001882.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013193.hg.1 | NM_001882.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013193.hg.1 | NM_001882.3 | Refseq |

CRHBP
Genomic Map