Assay Details
| Gene Symbol: PRDM11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001256695.1 | NP_001243624.1 | 3 - 4 | 468 | 81 | |
| NM_001256696.1 | NP_001243625.1 | 3 - 4 | 468 | 81 | ||
| NR_046338.1 | 4 - 5 | 574 | 81 | |||
| XM_011520222.2 | XP_011518524.1 | 3 - 4 | 616 | 81 | ||
| XM_011520223.2 | XP_011518525.1 | 3 - 4 | 298 | 81 | ||
| XM_011520224.2 | XP_011518526.1 | 3 - 4 | 611 | 81 | ||
| XM_011520225.2 | XP_011518527.1 | 3 - 4 | 248 | 81 | ||
| XM_011520228.2 | XP_011518530.1 | 3 - 4 | 609 | 81 | ||
| XM_011520230.2 | XP_011518532.1 | 3 - 4 | 601 | 81 | ||
| XM_011520231.2 | XP_011518533.1 | 3 - 4 | 603 | 81 | ||
| GenBank mRNA | AK094792.1 | - | 3 - 4 | 468 | 81 | |
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | ENST00000424263.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | ENST00000424263.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | ENST00000424263.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | ENST00000424263.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | ENST00000424263.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | ENST00000424263.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | ENST00000424263.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | ENST00000424263.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | ENST00000424263.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | ENST00000424263.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | ENST00000526442.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | ENST00000526442.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | ENST00000526442.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | ENST00000526442.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | ENST00000526442.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | ENST00000526442.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | ENST00000528980.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | ENST00000528980.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | ENST00000528980.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | ENST00000528980.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | ENST00000530656.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | ENST00000530656.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | ENST00000530656.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | ENST00000530656.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | ENST00000530656.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | ENST00000530656.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | ENST00000530656.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | ENST00000530656.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | ENST00000530656.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | ENST00000534751.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | ENST00000534751.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | ENST00000534751.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | ENST00000534751.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | ENST00000534751.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | ENST00000534751.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | ENST00000622142.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | ENST00000622142.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | ENST00000622142.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | ENST00000622142.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | ENST00000622142.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | ENST00000622142.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | ENST00000622142.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | ENST00000622142.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | ENST00000622142.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | ENST00000622142.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | ENST00000622142.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | NM_001256695.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | NM_001256695.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | NM_001256695.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | NM_001256695.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | NM_001256695.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | NM_001256695.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | NM_001256695.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | NM_001256695.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | NM_001256695.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | NM_001256695.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | NM_001256695.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | NM_001256696.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | NM_001256696.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | NM_001256696.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | NM_001256696.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | NM_001256696.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | NM_001256696.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | NM_001256696.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | NM_001256696.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | NM_001256696.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | NM_001256696.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | NR_046338.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | NR_046338.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | NR_046338.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | NR_046338.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | NR_046338.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | NR_046338.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | NR_046338.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | NR_046338.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | NR_046338.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | NR_046338.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | NR_046338.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007408.hg.1 | NR_046338.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100152390.hg.1 | ENST00000424263.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100152390.hg.1 | ENST00000526442.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100152390.hg.1 | ENST00000528980.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100152390.hg.1 | ENST00000530656.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100152390.hg.1 | ENST00000534751.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100152390.hg.1 | ENST00000622142.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100152390.hg.1 | NM_001256695.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100152390.hg.1 | NM_001256696.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100152390.hg.1 | NR_046338.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100152390.hg.1 | ENST00000424263.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100152390.hg.1 | ENST00000526442.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100152390.hg.1 | ENST00000528980.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100152390.hg.1 | ENST00000530656.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100152390.hg.1 | ENST00000534751.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100152390.hg.1 | ENST00000622142.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100152390.hg.1 | NM_001256695.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100152390.hg.1 | NM_001256696.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100152390.hg.1 | NR_046338.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100152390.hg.1 | TC1100007408.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1100152389.hg.1 | ENST00000424263.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100152389.hg.1 | ENST00000526442.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100152389.hg.1 | ENST00000530656.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100152389.hg.1 | ENST00000534751.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100152389.hg.1 | ENST00000622142.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100152389.hg.1 | NM_001256695.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100152389.hg.1 | NM_001256696.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100152389.hg.1 | NR_046338.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100152389.hg.1 | ENST00000424263.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100152389.hg.1 | ENST00000526442.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100152389.hg.1 | ENST00000530656.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100152389.hg.1 | ENST00000534751.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100152389.hg.1 | ENST00000622142.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100152389.hg.1 | NM_001256695.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100152389.hg.1 | NM_001256696.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100152389.hg.1 | NR_046338.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100152389.hg.1 | TC1100007408.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | ENST00000424263.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | ENST00000424263.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | ENST00000424263.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | ENST00000424263.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | ENST00000424263.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | ENST00000424263.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | ENST00000424263.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | ENST00000424263.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | ENST00000424263.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | ENST00000424263.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | ENST00000526442.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | ENST00000526442.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | ENST00000526442.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | ENST00000526442.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | ENST00000526442.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | ENST00000526442.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | ENST00000528980.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | ENST00000528980.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | ENST00000528980.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | ENST00000528980.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | ENST00000530656.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | ENST00000530656.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | ENST00000530656.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | ENST00000530656.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | ENST00000530656.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | ENST00000530656.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | ENST00000530656.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | ENST00000530656.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | ENST00000530656.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | ENST00000534751.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | ENST00000534751.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | ENST00000534751.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | ENST00000534751.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | ENST00000534751.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | ENST00000534751.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | ENST00000622142.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | ENST00000622142.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | ENST00000622142.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | ENST00000622142.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | ENST00000622142.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | ENST00000622142.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | ENST00000622142.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | ENST00000622142.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | ENST00000622142.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | ENST00000622142.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | ENST00000622142.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | NM_001256695.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | NM_001256695.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | NM_001256695.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | NM_001256695.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | NM_001256695.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | NM_001256695.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | NM_001256695.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | NM_001256695.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | NM_001256695.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | NM_001256695.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | NM_001256695.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | NM_001256696.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | NM_001256696.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | NM_001256696.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | NM_001256696.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | NM_001256696.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | NM_001256696.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | NM_001256696.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | NM_001256696.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | NM_001256696.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | NM_001256696.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | NR_046338.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | NR_046338.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | NR_046338.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | NR_046338.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | NR_046338.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | NR_046338.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | NR_046338.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | NR_046338.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | NR_046338.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | NR_046338.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | NR_046338.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007408.hg.1 | NR_046338.1 | Refseq |

PRDM11
Genomic Map