Assay Details
| Gene Symbol: LTA4H | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000895.2 | NP_000886.1 | 8 - 9 | 993 | 120 | |
| NM_001256643.1 | NP_001243572.1 | 8 - 9 | 924 | 120 | ||
| NM_001256644.1 | NP_001243573.1 | 8 - 9 | 924 | 120 | ||
| NR_132659.1 | 8 - 9 | 932 | 120 | |||
| XM_005268871.1 | XP_005268928.1 | 8 - 9 | 983 | 120 | ||
| XM_011538348.1 | XP_011536650.1 | 8 - 9 | 983 | 120 | ||
| XM_011538349.2 | XP_011536651.1 | 8 - 9 | 983 | 120 | ||
| GenBank mRNA | AB209740.1 | - | 9 - 10 | 929 | 120 | |
| AK293630.1 | - | 8 - 9 | 912 | 120 | ||
| AK298017.1 | - | 8 - 9 | 924 | 120 | ||
| AK301308.1 | - | 8 - 9 | 802 | 120 | ||
| AK308184.1 | - | 8 - 9 | 922 | 120 | ||
| BC032528.1 | - | 8 - 9 | 910 | 5502553 | 120 | |
| BX647158.1 | - | 8 - 9 | 881 | 120 | ||
| CR457068.1 | - | 8 - 9 | 851 | 120 | ||
| J02959.1 | - | 8 - 9 | 881 | 120 | ||
| J03459.1 | - | 8 - 9 | 919 | 120 | ||
More Information
Updated Mapping Information
Set Membership: |
|

LTA4H
Genomic Map