Assay Details
| Gene Symbol: MIB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020774.3 | NP_065825.1 | 18 - 19 | 2922 | 64 | |
| XM_011526098.1 | XP_011524400.1 | 10 - 11 | 1923 | 64 | ||
| XM_017025873.1 | XP_016881362.1 | 15 - 16 | 2614 | 64 | ||
| GenBank mRNA | AB037744.1 | - | 6 - 7 | 838 | 64 | |
| AK075157.1 | - | 6 - 7 | 789 | 64 | ||
| AK091266.1 | - | 17 - 18 | 2352 | 64 | ||
| AL713705.1 | - | 3 - 4 | 351 | 64 | ||
| AY147849.1 | - | 18 - 19 | 2940 | 64 | ||
| AY149908.1 | - | 18 - 19 | 2667 | 64 | ||
| BC022403.1 | - | 8 - 9 | 1043 | 64 | ||
| BC110581.1 | - | 18 - 19 | 2667 | 40008394 | 64 | |
| BC110582.1 | - | 18 - 19 | 2667 | 40008395 | 64 | |
| CR749635.1 | - | 15 - 16 | 2087 | 64 | ||
More Information
Updated Mapping Information
Set Membership: |
|

MIB1
Genomic Map