Assay Details
| Gene Symbol: MYBPC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_004533.3 | NP_004524.3 | 13 - 14 | 1522 | 66 | |
| GenBank mRNA | BC130536.1 | - | 13 - 14 | 1489 | 40146567 | 66 |
| BC136389.1 | - | 13 - 14 | 1489 | 9020376 | 66 | |
| CF552888.1 | - | 4 - 5 | 475 | 66 | ||
| X73113.1 | - | 13 - 14 | 1490 | 66 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC19000759.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR19010825.hg.1 | ENST00000357701.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19010825.hg.1 | NM_004533.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19010825.hg.1 | TC19000759.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008584.hg.1 | ENST00000357701.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008584.hg.1 | ENST00000357701.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008584.hg.1 | ENST00000357701.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008584.hg.1 | ENST00000357701.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008584.hg.1 | ENST00000357701.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008584.hg.1 | ENST00000357701.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008584.hg.1 | ENST00000357701.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008584.hg.1 | ENST00000357701.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008584.hg.1 | ENST00000357701.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008584.hg.1 | ENST00000357701.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008584.hg.1 | ENST00000357701.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008584.hg.1 | ENST00000357701.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008584.hg.1 | ENST00000357701.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008584.hg.1 | ENST00000357701.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008584.hg.1 | ENST00000357701.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008584.hg.1 | ENST00000357701.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008584.hg.1 | ENST00000357701.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008584.hg.1 | ENST00000357701.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008584.hg.1 | ENST00000357701.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008584.hg.1 | ENST00000357701.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008584.hg.1 | ENST00000357701.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008584.hg.1 | ENST00000357701.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008584.hg.1 | ENST00000357701.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008584.hg.1 | ENST00000357701.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008584.hg.1 | ENST00000357701.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008584.hg.1 | ENST00000357701.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008584.hg.1 | ENST00000357701.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008584.hg.1 | ENST00000357701.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008584.hg.1 | ENST00000597498.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008584.hg.1 | ENST00000597498.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008584.hg.1 | ENST00000597498.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008584.hg.1 | NM_004533.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008584.hg.1 | NM_004533.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008584.hg.1 | NM_004533.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008584.hg.1 | NM_004533.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008584.hg.1 | NM_004533.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008584.hg.1 | NM_004533.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008584.hg.1 | NM_004533.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008584.hg.1 | NM_004533.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008584.hg.1 | NM_004533.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008584.hg.1 | NM_004533.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008584.hg.1 | NM_004533.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008584.hg.1 | NM_004533.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008584.hg.1 | NM_004533.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008584.hg.1 | NM_004533.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008584.hg.1 | NM_004533.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008584.hg.1 | NM_004533.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008584.hg.1 | NM_004533.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008584.hg.1 | NM_004533.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008584.hg.1 | NM_004533.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008584.hg.1 | NM_004533.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008584.hg.1 | NM_004533.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008584.hg.1 | NM_004533.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008584.hg.1 | NM_004533.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008584.hg.1 | NM_004533.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008584.hg.1 | NM_004533.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008584.hg.1 | NM_004533.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008584.hg.1 | NM_004533.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008584.hg.1 | NM_004533.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900167039.hg.1 | ENST00000357701.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900167039.hg.1 | NM_004533.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900167039.hg.1 | TC1900008584.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900167039.hg.1 | ENST00000357701.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900167039.hg.1 | NM_004533.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008584.hg.1 | ENST00000357701.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008584.hg.1 | ENST00000357701.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008584.hg.1 | ENST00000357701.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008584.hg.1 | ENST00000357701.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008584.hg.1 | ENST00000357701.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008584.hg.1 | ENST00000357701.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008584.hg.1 | ENST00000357701.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008584.hg.1 | ENST00000357701.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008584.hg.1 | ENST00000357701.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008584.hg.1 | ENST00000357701.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008584.hg.1 | ENST00000357701.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008584.hg.1 | ENST00000357701.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008584.hg.1 | ENST00000357701.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008584.hg.1 | ENST00000357701.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008584.hg.1 | ENST00000357701.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008584.hg.1 | ENST00000357701.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008584.hg.1 | ENST00000357701.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008584.hg.1 | ENST00000357701.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008584.hg.1 | ENST00000357701.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008584.hg.1 | ENST00000357701.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008584.hg.1 | ENST00000357701.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008584.hg.1 | ENST00000357701.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008584.hg.1 | ENST00000357701.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008584.hg.1 | ENST00000357701.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008584.hg.1 | ENST00000357701.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008584.hg.1 | ENST00000357701.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008584.hg.1 | ENST00000357701.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008584.hg.1 | ENST00000357701.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008584.hg.1 | ENST00000597498.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008584.hg.1 | ENST00000597498.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008584.hg.1 | ENST00000597498.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008584.hg.1 | NM_004533.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008584.hg.1 | NM_004533.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008584.hg.1 | NM_004533.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008584.hg.1 | NM_004533.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008584.hg.1 | NM_004533.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008584.hg.1 | NM_004533.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008584.hg.1 | NM_004533.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008584.hg.1 | NM_004533.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008584.hg.1 | NM_004533.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008584.hg.1 | NM_004533.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008584.hg.1 | NM_004533.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008584.hg.1 | NM_004533.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008584.hg.1 | NM_004533.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008584.hg.1 | NM_004533.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008584.hg.1 | NM_004533.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008584.hg.1 | NM_004533.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008584.hg.1 | NM_004533.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008584.hg.1 | NM_004533.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008584.hg.1 | NM_004533.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008584.hg.1 | NM_004533.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008584.hg.1 | NM_004533.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008584.hg.1 | NM_004533.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008584.hg.1 | NM_004533.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008584.hg.1 | NM_004533.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008584.hg.1 | NM_004533.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008584.hg.1 | NM_004533.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008584.hg.1 | NM_004533.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008584.hg.1 | NM_004533.3 | Refseq |

MYBPC2
Genomic Map