Assay Details
| Gene Symbol: DNM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005336.2 | NP_001005336.1 | 10 - 11 | 1454 | 101 | |
| NM_004408.3 | NP_004399.2 | 10 - 11 | 1454 | 101 | ||
| XM_005251763.2 | XP_005251820.1 | 10 - 11 | 1454 | 101 | ||
| XM_005251769.2 | XP_005251826.1 | 10 - 11 | 1454 | 101 | ||
| XM_011518336.2 | XP_011516638.1 | 10 - 11 | 1454 | 101 | ||
| XM_017014369.1 | XP_016869858.1 | 10 - 11 | 1454 | 101 | ||
| XM_017014370.1 | XP_016869859.1 | 10 - 11 | 1454 | 101 | ||
| XM_017014371.1 | XP_016869860.1 | 10 - 11 | 1454 | 101 | ||
| XM_017014372.1 | XP_016869861.1 | 10 - 11 | 1454 | 101 | ||
| XM_017014373.1 | XP_016869862.1 | 10 - 11 | 1454 | 101 | ||
| GenBank mRNA | AB209124.1 | - | 9 - 10 | 1139 | 101 | |
| AK295101.1 | - | 10 - 11 | 1247 | 101 | ||
| BC050279.1 | - | 10 - 11 | 1430 | 5284480 | 101 | |
| BC063850.1 | - | 10 - 11 | 1387 | 6184100 | 101 | |
| L07807.1 | - | 10 - 11 | 1404 | 101 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC09000679.hg.1
Set Membership: |
|

DNM1
Genomic Map