Assay Details
| Gene Symbol: CLASP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_015282.2 | NP_056097.1 | 32 - 33 | 3767 | 69 | |
| GenBank mRNA | AB014522.1 | - | 24 - 25 | 2630 | 69 | |
| AF347693.1 | - | 31 - 32 | 3542 | 69 | ||
| BX648933.1 | - | 14 - 15 | 1416 | 69 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC02002264.hg.1
Set Membership: |
|

CLASP1
Genomic Map