Assay Details
| Gene Symbol: EPC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_015630.3 | NP_056445.3 | 5 - 6 | 846 | 149 | |
| XM_011510941.2 | XP_011509243.1 | 5 - 6 | 1057 | 149 | ||
| XM_011510943.2 | XP_011509245.1 | 4 - 5 | 730 | 149 | ||
| GenBank mRNA | AF286904.1 | - | 5 - 6 | 838 | 149 | |
| AK001433.1 | - | 3 - 4 | 437 | 149 | ||
| AK125789.1 | - | 6 - 7 | 915 | 149 | ||
| BC093818.1 | - | 5 - 6 | 758 | 7939663 | 149 | |
| BC093820.1 | - | 5 - 6 | 758 | 7939665 | 149 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC02000926.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR02013519.hg.1 | TC02000926.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02013519.hg.1 | ENST00000258484.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02013519.hg.1 | NM_015630.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02013520.hg.1 | TC02000926.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02013520.hg.1 | ENST00000258484.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02013520.hg.1 | NM_015630.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | ENST00000258484.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | ENST00000258484.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | ENST00000258484.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | ENST00000258484.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | ENST00000258484.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | ENST00000258484.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | ENST00000258484.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | ENST00000258484.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | ENST00000258484.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | ENST00000258484.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | ENST00000258484.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | ENST00000258484.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | ENST00000258484.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | ENST00000258484.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | ENST00000258484.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | ENST00000258484.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | ENST00000258484.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | ENST00000258484.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | ENST00000258484.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | ENST00000397424.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | ENST00000397424.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | ENST00000397424.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | ENST00000397424.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | ENST00000397424.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | ENST00000397424.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | ENST00000409654.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | ENST00000409654.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | ENST00000409654.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | ENST00000409654.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | ENST00000409654.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | ENST00000409654.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | ENST00000449013.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | ENST00000449013.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | ENST00000457184.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | ENST00000457184.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | ENST00000457184.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | ENST00000457184.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | ENST00000457184.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | ENST00000457184.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | ENST00000491099.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | ENST00000491099.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | ENST00000491099.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | ENST00000491099.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | ENST00000491099.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | ENST00000491099.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | NM_015630.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | NM_015630.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | NM_015630.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | NM_015630.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | NM_015630.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | NM_015630.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | NM_015630.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | NM_015630.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | NM_015630.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | NM_015630.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | NM_015630.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | NM_015630.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | NM_015630.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | NM_015630.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | NM_015630.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | NM_015630.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | NM_015630.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | NM_015630.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | NM_015630.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | ENST00000258484.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | ENST00000258484.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | ENST00000258484.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | ENST00000258484.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | ENST00000258484.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | ENST00000258484.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | ENST00000258484.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | ENST00000258484.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | ENST00000258484.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | ENST00000258484.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | ENST00000258484.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | ENST00000258484.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | ENST00000258484.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | ENST00000258484.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | ENST00000258484.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | ENST00000258484.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | ENST00000258484.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | ENST00000258484.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | ENST00000258484.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | ENST00000397424.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | ENST00000397424.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | ENST00000397424.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | ENST00000397424.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | ENST00000397424.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | ENST00000397424.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | ENST00000409654.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | ENST00000409654.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | ENST00000409654.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | ENST00000409654.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | ENST00000409654.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | ENST00000409654.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | ENST00000449013.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | ENST00000449013.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | ENST00000457184.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | ENST00000457184.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | ENST00000457184.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | ENST00000457184.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | ENST00000457184.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | ENST00000457184.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | ENST00000491099.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | ENST00000491099.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | ENST00000491099.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | ENST00000491099.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | ENST00000491099.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | ENST00000491099.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | NM_015630.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | NM_015630.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | NM_015630.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | NM_015630.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | NM_015630.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | NM_015630.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | NM_015630.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | NM_015630.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | NM_015630.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | NM_015630.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | NM_015630.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | NM_015630.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | NM_015630.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | NM_015630.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | NM_015630.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | NM_015630.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | NM_015630.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | NM_015630.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | NM_015630.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200167203.hg.1 | ENST00000258484.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200167203.hg.1 | NM_015630.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200167203.hg.1 | ENST00000258484.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200167203.hg.1 | NM_015630.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200167203.hg.1 | TC0200009612.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200167202.hg.1 | ENST00000258484.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200167202.hg.1 | NM_015630.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200167202.hg.1 | ENST00000258484.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200167202.hg.1 | NM_015630.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200167202.hg.1 | TC0200009612.hg.1 | Clariom D TC |

EPC2
Genomic Map