Assay Details
| Gene Symbol: RNF20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019592.6 | NP_062538.5 | 18 - 19 | 2744 | 99 | |
| XM_011518862.1 | XP_011517164.1 | 18 - 19 | 3021 | 99 | ||
| GenBank mRNA | AF265230.1 | - | 18 - 19 | 2742 | 99 | |
| AK002051.1 | - | 6 - 7 | 844 | 99 | ||
| AK022300.1 | - | 13 - 14 | 2016 | 99 | ||
| AK022532.1 | - | 18 - 19 | 2740 | 99 | ||
| AK314401.1 | - | 18 - 19 | 2699 | 99 | ||
| AL832910.1 | - | 4 - 5 | 583 | 99 | ||
| AL834272.1 | - | 8 - 9 | 1296 | 99 | ||
| BC110584.1 | - | 18 - 19 | 2737 | 40009044 | 99 | |
| BC110585.1 | - | 18 - 19 | 2737 | 40009047 | 99 | |
| BC152309.1 | - | 18 - 19 | 2737 | 40009034 | 99 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC09000519.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
protein polyubiquitination
regulation of transcription, DNA-templated
ubiquitin-dependent protein catabolic process
regulation of mitotic cell cycle
histone monoubiquitination
negative regulation of cell migration
positive regulation of histone methylation
histone H2B ubiquitination
positive regulation of transcription, DNA-templated
negative regulation of mRNA polyadenylation
positive regulation of histone H2B ubiquitination
p53 binding
chromatin binding
transcription coactivator activity
mRNA 3'-UTR binding
ubiquitin-protein transferase activity
protein binding
zinc ion binding
ligase activity
ubiquitin protein ligase binding
histone binding
protein polyubiquitination
regulation of transcription, DNA-templated
ubiquitin-dependent protein catabolic process
regulation of mitotic cell cycle
histone monoubiquitination
negative regulation of cell migration
positive regulation of histone methylation
histone H2B ubiquitination
positive regulation of transcription, DNA-templated
negative regulation of mRNA polyadenylation
positive regulation of histone H2B ubiquitination
p53 binding
chromatin binding
transcription coactivator activity
mRNA 3'-UTR binding
ubiquitin-protein transferase activity
protein binding
zinc ion binding
ligase activity
ubiquitin protein ligase binding
histone binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0900008257.hg.1 | ENST00000374819.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008257.hg.1 | ENST00000374819.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008257.hg.1 | ENST00000374819.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008257.hg.1 | ENST00000374819.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008257.hg.1 | ENST00000374819.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008257.hg.1 | ENST00000374819.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008257.hg.1 | ENST00000389120.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008257.hg.1 | ENST00000389120.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008257.hg.1 | ENST00000389120.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008257.hg.1 | ENST00000389120.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008257.hg.1 | ENST00000389120.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008257.hg.1 | ENST00000389120.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008257.hg.1 | ENST00000389120.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008257.hg.1 | ENST00000389120.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008257.hg.1 | ENST00000389120.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008257.hg.1 | ENST00000389120.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008257.hg.1 | ENST00000389120.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008257.hg.1 | ENST00000389120.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008257.hg.1 | ENST00000389120.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008257.hg.1 | ENST00000389120.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008257.hg.1 | ENST00000389120.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008257.hg.1 | ENST00000389120.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008257.hg.1 | ENST00000389120.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008257.hg.1 | ENST00000389120.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008257.hg.1 | ENST00000389120.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008257.hg.1 | ENST00000389120.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008257.hg.1 | ENST00000389120.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008257.hg.1 | ENST00000389120.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008257.hg.1 | ENST00000389120.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008257.hg.1 | ENST00000389120.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008257.hg.1 | ENST00000389120.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008257.hg.1 | ENST00000466817.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008257.hg.1 | ENST00000466817.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008257.hg.1 | ENST00000466817.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008257.hg.1 | ENST00000466817.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008257.hg.1 | ENST00000466817.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008257.hg.1 | ENST00000466817.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008257.hg.1 | ENST00000478072.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008257.hg.1 | ENST00000478072.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008257.hg.1 | ENST00000478072.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008257.hg.1 | ENST00000478072.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008257.hg.1 | ENST00000479306.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008257.hg.1 | ENST00000479306.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008257.hg.1 | ENST00000479306.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008257.hg.1 | ENST00000479306.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008257.hg.1 | ENST00000479306.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008257.hg.1 | ENST00000481046.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008257.hg.1 | ENST00000481046.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008257.hg.1 | ENST00000481046.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008257.hg.1 | ENST00000481046.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008257.hg.1 | ENST00000481046.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008257.hg.1 | NM_019592.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008257.hg.1 | NM_019592.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008257.hg.1 | NM_019592.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008257.hg.1 | NM_019592.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008257.hg.1 | NM_019592.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008257.hg.1 | NM_019592.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008257.hg.1 | NM_019592.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008257.hg.1 | NM_019592.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008257.hg.1 | NM_019592.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008257.hg.1 | NM_019592.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008257.hg.1 | NM_019592.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008257.hg.1 | NM_019592.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008257.hg.1 | NM_019592.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008257.hg.1 | NM_019592.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008257.hg.1 | NM_019592.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008257.hg.1 | NM_019592.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008257.hg.1 | NM_019592.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008257.hg.1 | NM_019592.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008257.hg.1 | NM_019592.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008257.hg.1 | NM_019592.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008257.hg.1 | NM_019592.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008257.hg.1 | NM_019592.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008257.hg.1 | NM_019592.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008257.hg.1 | NM_019592.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008257.hg.1 | NM_019592.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09005732.hg.1 | TC09000519.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09005732.hg.1 | ENST00000389120.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09005732.hg.1 | NM_019592.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008257.hg.1 | ENST00000374819.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008257.hg.1 | ENST00000374819.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008257.hg.1 | ENST00000374819.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008257.hg.1 | ENST00000374819.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008257.hg.1 | ENST00000374819.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008257.hg.1 | ENST00000374819.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008257.hg.1 | ENST00000389120.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008257.hg.1 | ENST00000389120.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008257.hg.1 | ENST00000389120.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008257.hg.1 | ENST00000389120.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008257.hg.1 | ENST00000389120.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008257.hg.1 | ENST00000389120.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008257.hg.1 | ENST00000389120.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008257.hg.1 | ENST00000389120.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008257.hg.1 | ENST00000389120.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008257.hg.1 | ENST00000389120.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008257.hg.1 | ENST00000389120.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008257.hg.1 | ENST00000389120.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008257.hg.1 | ENST00000389120.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008257.hg.1 | ENST00000389120.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008257.hg.1 | ENST00000389120.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008257.hg.1 | ENST00000389120.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008257.hg.1 | ENST00000389120.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008257.hg.1 | ENST00000389120.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008257.hg.1 | ENST00000389120.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008257.hg.1 | ENST00000389120.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008257.hg.1 | ENST00000389120.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008257.hg.1 | ENST00000389120.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008257.hg.1 | ENST00000389120.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008257.hg.1 | ENST00000389120.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008257.hg.1 | ENST00000389120.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008257.hg.1 | ENST00000466817.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008257.hg.1 | ENST00000466817.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008257.hg.1 | ENST00000466817.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008257.hg.1 | ENST00000466817.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008257.hg.1 | ENST00000466817.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008257.hg.1 | ENST00000466817.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008257.hg.1 | ENST00000478072.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008257.hg.1 | ENST00000478072.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008257.hg.1 | ENST00000478072.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008257.hg.1 | ENST00000478072.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008257.hg.1 | ENST00000479306.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008257.hg.1 | ENST00000479306.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008257.hg.1 | ENST00000479306.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008257.hg.1 | ENST00000479306.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008257.hg.1 | ENST00000479306.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008257.hg.1 | ENST00000481046.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008257.hg.1 | ENST00000481046.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008257.hg.1 | ENST00000481046.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008257.hg.1 | ENST00000481046.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008257.hg.1 | ENST00000481046.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008257.hg.1 | NM_019592.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008257.hg.1 | NM_019592.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008257.hg.1 | NM_019592.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008257.hg.1 | NM_019592.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008257.hg.1 | NM_019592.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008257.hg.1 | NM_019592.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008257.hg.1 | NM_019592.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008257.hg.1 | NM_019592.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008257.hg.1 | NM_019592.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008257.hg.1 | NM_019592.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008257.hg.1 | NM_019592.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008257.hg.1 | NM_019592.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008257.hg.1 | NM_019592.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008257.hg.1 | NM_019592.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008257.hg.1 | NM_019592.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008257.hg.1 | NM_019592.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008257.hg.1 | NM_019592.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008257.hg.1 | NM_019592.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008257.hg.1 | NM_019592.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008257.hg.1 | NM_019592.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008257.hg.1 | NM_019592.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008257.hg.1 | NM_019592.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008257.hg.1 | NM_019592.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008257.hg.1 | NM_019592.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008257.hg.1 | NM_019592.6 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900155358.hg.1 | ENST00000389120.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900155358.hg.1 | NM_019592.6 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900155358.hg.1 | ENST00000389120.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900155358.hg.1 | NM_019592.6 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900155358.hg.1 | TC0900008257.hg.1 | Clariom D TC |

RNF20
Genomic Map