Assay Details
| Gene Symbol: MTCH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_014341.2 | NP_055156.1 | 8 - 9 | 1230 | 95 | |
| NR_130739.1 | 8 - 9 | 1323 | 95 | |||
| XM_005248979.1 | XP_005249036.1 | 8 - 9 | 1230 | 95 | ||
| XM_011514450.1 | XP_011512752.1 | 7 - 8 | 738 | 95 | ||
| GenBank mRNA | AF151822.1 | - | 8 - 9 | 702 | 95 | |
| AF176006.3 | - | 8 - 9 | 852 | 95 | ||
| AK022783.1 | - | 8 - 9 | 850 | 95 | ||
| AK130842.1 | - | 8 - 9 | 934 | 95 | ||
| AK303318.1 | - | 7 - 8 | 651 | 95 | ||
| AK315067.1 | - | 8 - 9 | 1231 | 95 | ||
| AY358519.1 | - | 5 - 6 | 311 | 95 | ||
| AY947344.1 | - | 8 - 9 | 821 | 95 | ||
| BC000702.2 | - | 8 - 9 | 782 | 95 | ||
| BC006507.2 | - | 8 - 9 | 582 | 95 | ||
| BC009962.2 | - | 8 - 9 | 831 | 95 | ||
| BC110914.1 | - | 8 - 9 | 873 | 5179722 | 95 | |
| BC125056.1 | - | 8 - 9 | 847 | 95 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

MTCH1
Genomic Map