Assay Details
| Gene Symbol: MTA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001282755.1 | NP_001269684.1 | 2 - 3 | 531 | 88 | |
| XM_005264459.2 | XP_005264516.1 | 2 - 3 | 531 | 88 | ||
| GenBank mRNA | AK127245.1 | - | 2 - 3 | 531 | 88 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001282755.1, XM_005264459.2 transcripts.
Set Membership: |
|

MTA3
Genomic Map