Assay Details
| Gene Symbol: ATP6V0B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039457.2 | NP_001034546.1 | 1 - 2 | 463 | 70 | |
| GenBank mRNA | BG475609.1 | - | 1 - 2 | 179 | 70 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the NM_001039457.2 transcripts.
Set Membership: |
|

ATP6V0B
Genomic Map