Assay Details
| Gene Symbol: AGPAT5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018361.3 | NP_060831.2 | 1 - 2 | 521 | 125 | |
| XM_011534751.2 | XP_011533053.1 | 1 - 2 | 291 | 125 | ||
| GenBank mRNA | AF375789.1 | - | 1 - 2 | 483 | 125 | |
| AK002072.1 | - | 1 - 2 | 185 | 125 | ||
| AL136587.1 | - | 1 - 2 | 211 | 125 | ||
| BC023550.2 | - | 1 - 2 | 465 | 4129973 | 125 | |
| BC068519.1 | - | 1 - 2 | 185 | 125 | ||
| BC080537.1 | - | 1 - 2 | 490 | 5015128 | 125 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC08000022.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR08000345.hg.1 | TC08000022.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR08000345.hg.1 | ENST00000285518.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08000345.hg.1 | NM_018361.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08000340.hg.1 | TC08000022.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR08000340.hg.1 | ENST00000285518.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08000340.hg.1 | NM_018361.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08000348.hg.1 | TC08000022.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR08000348.hg.1 | ENST00000285518.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08000348.hg.1 | NM_018361.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08000342.hg.1 | TC08000022.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR08000342.hg.1 | ENST00000285518.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08000342.hg.1 | NM_018361.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08000343.hg.1 | TC08000022.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR08000343.hg.1 | ENST00000285518.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08000343.hg.1 | NM_018361.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08000344.hg.1 | TC08000022.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR08000344.hg.1 | ENST00000285518.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08000344.hg.1 | NM_018361.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | ENST00000285518.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | ENST00000285518.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | ENST00000285518.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | ENST00000285518.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | ENST00000285518.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | ENST00000285518.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | ENST00000285518.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | ENST00000285518.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | ENST00000285518.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | ENST00000285518.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | ENST00000285518.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | ENST00000285518.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | ENST00000285518.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | ENST00000285518.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | ENST00000285518.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | ENST00000285518.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | ENST00000285518.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | ENST00000285518.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | ENST00000518327.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | ENST00000518327.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | ENST00000518327.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | ENST00000518327.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | ENST00000518327.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | ENST00000518327.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | ENST00000523234.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | ENST00000523234.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | ENST00000523234.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | ENST00000523234.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | ENST00000523234.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | ENST00000523234.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | ENST00000523234.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | ENST00000523234.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | ENST00000523234.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | ENST00000523234.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | ENST00000523234.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | ENST00000523234.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | ENST00000523234.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | ENST00000523586.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | ENST00000523586.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | ENST00000523586.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | ENST00000523586.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | ENST00000523586.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | ENST00000530716.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | ENST00000530716.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | ENST00000530716.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | ENST00000530716.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | ENST00000530716.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | ENST00000533159.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | ENST00000533159.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | ENST00000533159.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | ENST00000533159.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | ENST00000533648.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | ENST00000533648.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | NM_018361.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | NM_018361.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | NM_018361.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | NM_018361.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | NM_018361.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | NM_018361.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | NM_018361.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | NM_018361.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | NM_018361.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | NM_018361.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | NM_018361.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | NM_018361.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | NM_018361.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | NM_018361.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | NM_018361.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | NM_018361.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | NM_018361.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | NM_018361.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006566.hg.1 | NM_018361.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800144768.hg.1 | ENST00000285518.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800144768.hg.1 | ENST00000523586.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800144768.hg.1 | NM_018361.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800144768.hg.1 | ENST00000285518.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800144768.hg.1 | ENST00000523586.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800144768.hg.1 | NM_018361.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800144768.hg.1 | TC0800006566.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0800144765.hg.1 | ENST00000285518.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800144765.hg.1 | ENST00000518327.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800144765.hg.1 | ENST00000523234.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800144765.hg.1 | ENST00000523586.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800144765.hg.1 | NM_018361.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800144765.hg.1 | ENST00000285518.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800144765.hg.1 | ENST00000518327.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800144765.hg.1 | ENST00000523234.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800144765.hg.1 | ENST00000523586.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800144765.hg.1 | NM_018361.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800144765.hg.1 | TC0800006566.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | ENST00000285518.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | ENST00000285518.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | ENST00000285518.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | ENST00000285518.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | ENST00000285518.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | ENST00000285518.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | ENST00000285518.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | ENST00000285518.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | ENST00000285518.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | ENST00000285518.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | ENST00000285518.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | ENST00000285518.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | ENST00000285518.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | ENST00000285518.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | ENST00000285518.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | ENST00000285518.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | ENST00000285518.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | ENST00000285518.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | ENST00000518327.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | ENST00000518327.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | ENST00000518327.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | ENST00000518327.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | ENST00000518327.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | ENST00000518327.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | ENST00000523234.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | ENST00000523234.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | ENST00000523234.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | ENST00000523234.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | ENST00000523234.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | ENST00000523234.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | ENST00000523234.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | ENST00000523234.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | ENST00000523234.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | ENST00000523234.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | ENST00000523234.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | ENST00000523234.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | ENST00000523234.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | ENST00000523586.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | ENST00000523586.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | ENST00000523586.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | ENST00000523586.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | ENST00000523586.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | ENST00000530716.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | ENST00000530716.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | ENST00000530716.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | ENST00000530716.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | ENST00000530716.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | ENST00000533159.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | ENST00000533159.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | ENST00000533159.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | ENST00000533159.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | ENST00000533648.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | ENST00000533648.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | NM_018361.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | NM_018361.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | NM_018361.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | NM_018361.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | NM_018361.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | NM_018361.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | NM_018361.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | NM_018361.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | NM_018361.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | NM_018361.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | NM_018361.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | NM_018361.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | NM_018361.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | NM_018361.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | NM_018361.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | NM_018361.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | NM_018361.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | NM_018361.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006566.hg.1 | NM_018361.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800144764.hg.1 | ENST00000285518.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800144764.hg.1 | ENST00000518327.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800144764.hg.1 | ENST00000523234.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800144764.hg.1 | NM_018361.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800144764.hg.1 | ENST00000285518.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800144764.hg.1 | ENST00000518327.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800144764.hg.1 | ENST00000523234.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800144764.hg.1 | NM_018361.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800144764.hg.1 | TC0800006566.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0800144761.hg.1 | ENST00000285518.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800144761.hg.1 | NM_018361.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800144761.hg.1 | ENST00000285518.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800144761.hg.1 | NM_018361.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800144761.hg.1 | TC0800006566.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0800144759.hg.1 | ENST00000285518.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800144759.hg.1 | NM_018361.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800144759.hg.1 | ENST00000285518.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800144759.hg.1 | NM_018361.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800144759.hg.1 | TC0800006566.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0800144760.hg.1 | ENST00000285518.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800144760.hg.1 | NM_018361.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800144760.hg.1 | ENST00000285518.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800144760.hg.1 | NM_018361.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800144760.hg.1 | TC0800006566.hg.1 | Clariom D TC |

AGPAT5
Genomic Map