Assay Details
| Gene Symbol: EHHADH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001166415.1 | NP_001159887.1 | 5 - 6 | 767 | 74 | |
| NM_001966.3 | NP_001957.2 | 5 - 6 | 644 | 74 | ||
| XM_006713525.1 | XP_006713588.1 | 6 - 7 | 626 | 74 | ||
| XM_011512517.1 | XP_011510819.1 | 5 - 6 | 1471 | 74 | ||
| XM_017005839.1 | XP_016861328.1 | 6 - 7 | 1502 | 74 | ||
| GenBank mRNA | AK223460.1 | - | 5 - 6 | 608 | 74 | |
| AK291798.1 | - | 5 - 6 | 595 | 74 | ||
| AK301521.1 | - | 5 - 6 | 705 | 74 | ||
| BC030982.1 | - | 5 - 6 | 595 | 74 | ||
| BC038948.1 | - | 5 - 6 | 585 | 5533337 | 74 | |
| BC110460.2 | - | 5 - 6 | 575 | 40026238 | 74 | |
| L07077.1 | - | 5 - 6 | 575 | 74 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC03002076.hg.1
Additional Information:
This assay is in the 5'UTR region of the XM_006713525.1,XM_017005839.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
internal protein amino acid acetylation
fatty acid beta-oxidation
cellular lipid metabolic process
3-hydroxyacyl-CoA dehydrogenase activity
dodecenoyl-CoA delta-isomerase activity
enoyl-CoA hydratase activity
receptor binding
protein binding
long-chain-enoyl-CoA hydratase activity
enzyme binding
internal protein amino acid acetylation
fatty acid beta-oxidation
cellular lipid metabolic process
3-hydroxyacyl-CoA dehydrogenase activity
dodecenoyl-CoA delta-isomerase activity
enoyl-CoA hydratase activity
receptor binding
protein binding
long-chain-enoyl-CoA hydratase activity
enzyme binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR03036742.hg.1 | TC03002076.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03036742.hg.1 | ENST00000231887.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03036742.hg.1 | ENST00000456310.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03036742.hg.1 | ENST00000537544.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03036742.hg.1 | NM_001166415.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03036742.hg.1 | NM_001966.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013408.hg.1 | ENST00000231887.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013408.hg.1 | ENST00000231887.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013408.hg.1 | ENST00000231887.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013408.hg.1 | ENST00000231887.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013408.hg.1 | ENST00000231887.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013408.hg.1 | ENST00000231887.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013408.hg.1 | ENST00000231887.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013408.hg.1 | ENST00000231887.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013408.hg.1 | ENST00000231887.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013408.hg.1 | ENST00000231887.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013408.hg.1 | ENST00000231887.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013408.hg.1 | ENST00000231887.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013408.hg.1 | ENST00000231887.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013408.hg.1 | ENST00000231887.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013408.hg.1 | ENST00000440662.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013408.hg.1 | ENST00000440662.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013408.hg.1 | ENST00000440662.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013408.hg.1 | ENST00000456310.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013408.hg.1 | ENST00000456310.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013408.hg.1 | ENST00000456310.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013408.hg.1 | ENST00000456310.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013408.hg.1 | ENST00000456310.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013408.hg.1 | ENST00000456310.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013408.hg.1 | ENST00000456310.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013408.hg.1 | ENST00000456310.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013408.hg.1 | ENST00000456310.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013408.hg.1 | ENST00000456310.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013408.hg.1 | ENST00000456310.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013408.hg.1 | ENST00000456310.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013408.hg.1 | ENST00000456310.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013408.hg.1 | ENST00000465178.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013408.hg.1 | ENST00000465178.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013408.hg.1 | ENST00000465178.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013408.hg.1 | ENST00000475987.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013408.hg.1 | ENST00000475987.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013408.hg.1 | ENST00000475987.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013408.hg.1 | ENST00000475987.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013408.hg.1 | ENST00000475987.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013408.hg.1 | ENST00000475987.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013408.hg.1 | ENST00000475987.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013408.hg.1 | ENST00000483104.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013408.hg.1 | ENST00000483104.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013408.hg.1 | ENST00000483104.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013408.hg.1 | NM_001166415.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013408.hg.1 | NM_001166415.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013408.hg.1 | NM_001166415.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013408.hg.1 | NM_001166415.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013408.hg.1 | NM_001166415.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013408.hg.1 | NM_001166415.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013408.hg.1 | NM_001166415.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013408.hg.1 | NM_001166415.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013408.hg.1 | NM_001166415.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013408.hg.1 | NM_001166415.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013408.hg.1 | NM_001166415.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013408.hg.1 | NM_001166415.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013408.hg.1 | NM_001166415.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013408.hg.1 | NM_001166415.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013408.hg.1 | NM_001166415.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013408.hg.1 | NM_001966.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013408.hg.1 | NM_001966.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013408.hg.1 | NM_001966.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013408.hg.1 | NM_001966.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013408.hg.1 | NM_001966.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013408.hg.1 | NM_001966.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013408.hg.1 | NM_001966.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013408.hg.1 | NM_001966.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013408.hg.1 | NM_001966.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013408.hg.1 | NM_001966.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013408.hg.1 | NM_001966.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013408.hg.1 | NM_001966.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013408.hg.1 | NM_001966.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013408.hg.1 | NM_001966.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03036743.hg.1 | TC03002076.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03036743.hg.1 | ENST00000231887.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03036743.hg.1 | ENST00000456310.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03036743.hg.1 | ENST00000537544.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03036743.hg.1 | NM_001166415.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03036743.hg.1 | NM_001966.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300199933.hg.1 | ENST00000231887.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300199933.hg.1 | ENST00000456310.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300199933.hg.1 | ENST00000483104.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300199933.hg.1 | NM_001166415.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300199933.hg.1 | NM_001966.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300199933.hg.1 | ENST00000231887.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300199933.hg.1 | ENST00000456310.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300199933.hg.1 | ENST00000483104.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300199933.hg.1 | NM_001166415.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300199933.hg.1 | NM_001966.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300199933.hg.1 | TC0300013408.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0300013408.hg.1 | ENST00000231887.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013408.hg.1 | ENST00000231887.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013408.hg.1 | ENST00000231887.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013408.hg.1 | ENST00000231887.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013408.hg.1 | ENST00000231887.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013408.hg.1 | ENST00000231887.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013408.hg.1 | ENST00000231887.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013408.hg.1 | ENST00000231887.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013408.hg.1 | ENST00000231887.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013408.hg.1 | ENST00000231887.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013408.hg.1 | ENST00000231887.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013408.hg.1 | ENST00000231887.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013408.hg.1 | ENST00000231887.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013408.hg.1 | ENST00000231887.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013408.hg.1 | ENST00000440662.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013408.hg.1 | ENST00000440662.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013408.hg.1 | ENST00000440662.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013408.hg.1 | ENST00000456310.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013408.hg.1 | ENST00000456310.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013408.hg.1 | ENST00000456310.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013408.hg.1 | ENST00000456310.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013408.hg.1 | ENST00000456310.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013408.hg.1 | ENST00000456310.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013408.hg.1 | ENST00000456310.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013408.hg.1 | ENST00000456310.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013408.hg.1 | ENST00000456310.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013408.hg.1 | ENST00000456310.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013408.hg.1 | ENST00000456310.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013408.hg.1 | ENST00000456310.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013408.hg.1 | ENST00000456310.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013408.hg.1 | ENST00000465178.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013408.hg.1 | ENST00000465178.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013408.hg.1 | ENST00000465178.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013408.hg.1 | ENST00000475987.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013408.hg.1 | ENST00000475987.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013408.hg.1 | ENST00000475987.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013408.hg.1 | ENST00000475987.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013408.hg.1 | ENST00000475987.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013408.hg.1 | ENST00000475987.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013408.hg.1 | ENST00000475987.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013408.hg.1 | ENST00000483104.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013408.hg.1 | ENST00000483104.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013408.hg.1 | ENST00000483104.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013408.hg.1 | NM_001166415.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013408.hg.1 | NM_001166415.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013408.hg.1 | NM_001166415.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013408.hg.1 | NM_001166415.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013408.hg.1 | NM_001166415.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013408.hg.1 | NM_001166415.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013408.hg.1 | NM_001166415.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013408.hg.1 | NM_001166415.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013408.hg.1 | NM_001166415.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013408.hg.1 | NM_001166415.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013408.hg.1 | NM_001166415.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013408.hg.1 | NM_001166415.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013408.hg.1 | NM_001166415.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013408.hg.1 | NM_001166415.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013408.hg.1 | NM_001166415.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013408.hg.1 | NM_001966.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013408.hg.1 | NM_001966.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013408.hg.1 | NM_001966.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013408.hg.1 | NM_001966.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013408.hg.1 | NM_001966.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013408.hg.1 | NM_001966.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013408.hg.1 | NM_001966.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013408.hg.1 | NM_001966.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013408.hg.1 | NM_001966.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013408.hg.1 | NM_001966.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013408.hg.1 | NM_001966.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013408.hg.1 | NM_001966.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013408.hg.1 | NM_001966.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013408.hg.1 | NM_001966.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300199932.hg.1 | ENST00000231887.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300199932.hg.1 | ENST00000456310.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300199932.hg.1 | NM_001166415.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300199932.hg.1 | NM_001966.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300199932.hg.1 | ENST00000231887.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300199932.hg.1 | ENST00000456310.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300199932.hg.1 | NM_001166415.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300199932.hg.1 | NM_001966.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300199932.hg.1 | TC0300013408.hg.1 | Clariom D TC |

EHHADH
Genomic Map