Assay Details
| Gene Symbol: MRPL47 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177988.1 | NP_817125.1 | 1 - 2 | 119 | 164 | |
| GenBank mRNA | BC021575.1 | - | 1 - 2 | 119 | 3161966 | 164 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC03002039.hg
Additional Information:
This assay is in the 5'UTR region of the BC021575.1,NM_177988.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR03036105.hg.1 | TC03002039.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03036105.hg.1 | ENST00000259038.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03036105.hg.1 | ENST00000392659.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03036105.hg.1 | ENST00000476781.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03036105.hg.1 | NM_020409.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03036105.hg.1 | NM_177988.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03036107.hg.1 | TC03002039.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03036107.hg.1 | ENST00000259038.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03036107.hg.1 | ENST00000392659.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03036107.hg.1 | ENST00000476781.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03036107.hg.1 | NM_020409.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03036107.hg.1 | NM_177988.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013270.hg.1 | ENST00000259038.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013270.hg.1 | ENST00000259038.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013270.hg.1 | ENST00000259038.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013270.hg.1 | ENST00000259038.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013270.hg.1 | ENST00000259038.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013270.hg.1 | ENST00000259038.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013270.hg.1 | ENST00000259038.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013270.hg.1 | ENST00000259038.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013270.hg.1 | ENST00000259038.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013270.hg.1 | ENST00000259038.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013270.hg.1 | ENST00000392659.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013270.hg.1 | ENST00000392659.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013270.hg.1 | ENST00000392659.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013270.hg.1 | ENST00000392659.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013270.hg.1 | ENST00000392659.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013270.hg.1 | ENST00000392659.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013270.hg.1 | ENST00000392659.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013270.hg.1 | ENST00000392659.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013270.hg.1 | ENST00000476781.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013270.hg.1 | ENST00000476781.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013270.hg.1 | ENST00000476781.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013270.hg.1 | ENST00000476781.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013270.hg.1 | ENST00000476781.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013270.hg.1 | ENST00000476781.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013270.hg.1 | ENST00000476781.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013270.hg.1 | ENST00000476781.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013270.hg.1 | ENST00000476781.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013270.hg.1 | ENST00000476781.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013270.hg.1 | ENST00000476781.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013270.hg.1 | ENST00000476781.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013270.hg.1 | NM_020409.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013270.hg.1 | NM_020409.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013270.hg.1 | NM_020409.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013270.hg.1 | NM_020409.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013270.hg.1 | NM_020409.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013270.hg.1 | NM_020409.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013270.hg.1 | NM_020409.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013270.hg.1 | NM_020409.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013270.hg.1 | NM_020409.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013270.hg.1 | NM_020409.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013270.hg.1 | NM_020409.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013270.hg.1 | NM_177988.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013270.hg.1 | NM_177988.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013270.hg.1 | NM_177988.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013270.hg.1 | NM_177988.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013270.hg.1 | NM_177988.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013270.hg.1 | NM_177988.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013270.hg.1 | NM_177988.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013270.hg.1 | NM_177988.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013270.hg.1 | NM_177988.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03036106.hg.1 | TC03002039.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03036106.hg.1 | ENST00000259038.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03036106.hg.1 | ENST00000392659.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03036106.hg.1 | ENST00000476781.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03036106.hg.1 | NM_020409.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03036106.hg.1 | NM_177988.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300198872.hg.1 | ENST00000259038.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300198872.hg.1 | ENST00000392659.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300198872.hg.1 | ENST00000476781.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300198872.hg.1 | NM_020409.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300198872.hg.1 | NM_177988.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300198872.hg.1 | ENST00000259038.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300198872.hg.1 | ENST00000392659.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300198872.hg.1 | ENST00000476781.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300198872.hg.1 | NM_020409.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300198872.hg.1 | NM_177988.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300198872.hg.1 | TC0300013270.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0300198869.hg.1 | ENST00000259038.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300198869.hg.1 | ENST00000392659.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300198869.hg.1 | ENST00000476781.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300198869.hg.1 | NM_020409.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300198869.hg.1 | NM_177988.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300198869.hg.1 | ENST00000259038.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300198869.hg.1 | ENST00000392659.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300198869.hg.1 | ENST00000476781.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300198869.hg.1 | NM_020409.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300198869.hg.1 | NM_177988.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300198869.hg.1 | TC0300013270.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0300013270.hg.1 | ENST00000259038.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013270.hg.1 | ENST00000259038.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013270.hg.1 | ENST00000259038.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013270.hg.1 | ENST00000259038.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013270.hg.1 | ENST00000259038.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013270.hg.1 | ENST00000259038.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013270.hg.1 | ENST00000259038.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013270.hg.1 | ENST00000259038.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013270.hg.1 | ENST00000259038.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013270.hg.1 | ENST00000259038.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013270.hg.1 | ENST00000392659.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013270.hg.1 | ENST00000392659.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013270.hg.1 | ENST00000392659.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013270.hg.1 | ENST00000392659.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013270.hg.1 | ENST00000392659.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013270.hg.1 | ENST00000392659.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013270.hg.1 | ENST00000392659.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013270.hg.1 | ENST00000392659.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013270.hg.1 | ENST00000476781.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013270.hg.1 | ENST00000476781.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013270.hg.1 | ENST00000476781.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013270.hg.1 | ENST00000476781.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013270.hg.1 | ENST00000476781.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013270.hg.1 | ENST00000476781.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013270.hg.1 | ENST00000476781.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013270.hg.1 | ENST00000476781.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013270.hg.1 | ENST00000476781.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013270.hg.1 | ENST00000476781.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013270.hg.1 | ENST00000476781.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013270.hg.1 | ENST00000476781.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013270.hg.1 | NM_020409.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013270.hg.1 | NM_020409.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013270.hg.1 | NM_020409.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013270.hg.1 | NM_020409.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013270.hg.1 | NM_020409.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013270.hg.1 | NM_020409.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013270.hg.1 | NM_020409.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013270.hg.1 | NM_020409.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013270.hg.1 | NM_020409.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013270.hg.1 | NM_020409.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013270.hg.1 | NM_020409.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013270.hg.1 | NM_177988.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013270.hg.1 | NM_177988.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013270.hg.1 | NM_177988.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013270.hg.1 | NM_177988.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013270.hg.1 | NM_177988.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013270.hg.1 | NM_177988.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013270.hg.1 | NM_177988.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013270.hg.1 | NM_177988.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013270.hg.1 | NM_177988.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300198867.hg.1 | ENST00000259038.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300198867.hg.1 | ENST00000392659.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300198867.hg.1 | ENST00000476781.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300198867.hg.1 | NM_020409.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300198867.hg.1 | NM_177988.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300198867.hg.1 | ENST00000259038.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300198867.hg.1 | ENST00000392659.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300198867.hg.1 | ENST00000476781.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300198867.hg.1 | NM_020409.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300198867.hg.1 | NM_177988.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300198867.hg.1 | TC0300013270.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0300198870.hg.1 | ENST00000259038.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300198870.hg.1 | ENST00000392659.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300198870.hg.1 | ENST00000476781.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300198870.hg.1 | NM_020409.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300198870.hg.1 | NM_177988.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300198870.hg.1 | ENST00000259038.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300198870.hg.1 | ENST00000392659.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300198870.hg.1 | ENST00000476781.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300198870.hg.1 | NM_020409.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300198870.hg.1 | NM_177988.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300198870.hg.1 | TC0300013270.hg.1 | Clariom D TC |

MRPL47
Genomic Map