Assay Details
| Gene Symbol: SARS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_006513.3 | NP_006504.2 | 3 - 4 | 384 | 87 | |
| XM_006710813.2 | XP_006710876.1 | 3 - 4 | 384 | 87 | ||
| GenBank mRNA | AK222677.1 | - | 3 - 4 | 345 | 87 | |
| AK312771.1 | - | 3 - 4 | 345 | 87 | ||
| BC000716.1 | - | 3 - 4 | 293 | 3505218 | 87 | |
| BC009390.2 | - | 3 - 4 | 331 | 4129592 | 87 | |
| BC111398.1 | - | 3 - 4 | 363 | 4133889 | 87 | |
| BP363347.1 | - | 3 - 4 | 384 | 87 | ||
| D49914.1 | - | 3 - 4 | 287 | 87 | ||
| X91257.1 | - | 3 - 4 | 359 | 87 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01000940.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01014812.hg.1 | TC01000940.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01014812.hg.1 | ENST00000234677.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01014812.hg.1 | ENST00000369923.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01014812.hg.1 | NM_006513.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01014813.hg.1 | TC01000940.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01014813.hg.1 | ENST00000234677.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01014813.hg.1 | ENST00000369923.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01014813.hg.1 | NM_006513.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000234677.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000234677.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000234677.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000234677.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000234677.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000234677.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000234677.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000234677.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000234677.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000234677.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000234677.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000234677.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000369923.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000369923.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000369923.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000369923.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000369923.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000369923.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000369923.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000369923.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000369923.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000369923.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000369923.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000369923.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000369923.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000369923.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000468588.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000468588.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000468588.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000468588.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000468588.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000468588.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000468588.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000468588.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000468588.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000471705.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000471705.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000471705.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000477544.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000477544.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000477544.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000477544.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000477544.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000482384.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000482384.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | NM_006513.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | NM_006513.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | NM_006513.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | NM_006513.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | NM_006513.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | NM_006513.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | NM_006513.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | NM_006513.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | NM_006513.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | NM_006513.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | NM_006513.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | NM_006513.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | NM_006513.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | NM_006513.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | NR_034072.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | NR_034072.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | NR_034072.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | NR_034072.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100166646.hg.1 | ENST00000234677.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100166646.hg.1 | ENST00000369923.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100166646.hg.1 | ENST00000477544.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100166646.hg.1 | NM_006513.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100166646.hg.1 | ENST00000234677.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100166646.hg.1 | ENST00000369923.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100166646.hg.1 | ENST00000477544.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100166646.hg.1 | NM_006513.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100166646.hg.1 | TC0100009344.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000234677.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000234677.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000234677.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000234677.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000234677.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000234677.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000234677.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000234677.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000234677.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000234677.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000234677.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000234677.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000369923.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000369923.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000369923.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000369923.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000369923.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000369923.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000369923.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000369923.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000369923.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000369923.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000369923.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000369923.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000369923.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000369923.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000468588.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000468588.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000468588.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000468588.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000468588.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000468588.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000468588.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000468588.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000468588.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000471705.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000471705.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000471705.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000477544.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000477544.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000477544.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000477544.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000477544.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000482384.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000482384.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | NM_006513.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | NM_006513.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | NM_006513.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | NM_006513.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | NM_006513.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | NM_006513.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | NM_006513.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | NM_006513.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | NM_006513.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | NM_006513.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | NM_006513.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | NM_006513.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | NM_006513.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | NM_006513.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | NR_034072.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | NR_034072.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | NR_034072.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | NR_034072.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100166647.hg.1 | ENST00000234677.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100166647.hg.1 | ENST00000369923.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100166647.hg.1 | NM_006513.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100166647.hg.1 | ENST00000234677.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100166647.hg.1 | ENST00000369923.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100166647.hg.1 | NM_006513.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100166647.hg.1 | TC0100009344.hg.1 | Clariom D TC |

SARS
Genomic Map