Assay Details
| Gene Symbol: ETNK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039481.1 | NP_001034570.1 | 2 - 3 | 699 | 95 | |
| GenBank mRNA | BC006111.1 | - | 2 - 3 | 651 | 95 | |
| BT007084.1 | - | 2 - 3 | 410 | 95 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC12000239.hg.1
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR12002939.hg.1 | ENST00000335148.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12002939.hg.1 | NM_001039481.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12002939.hg.1 | TC12000239.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | ENST00000266517.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | ENST00000266517.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | ENST00000266517.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | ENST00000266517.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | ENST00000266517.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | ENST00000266517.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | ENST00000266517.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | ENST00000266517.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | ENST00000266517.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | ENST00000266517.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | ENST00000266517.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | ENST00000266517.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | ENST00000266517.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | ENST00000266517.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | ENST00000266517.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | ENST00000266517.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | ENST00000335148.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | ENST00000335148.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | ENST00000335148.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | ENST00000335148.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | ENST00000335148.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | ENST00000335148.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | ENST00000335148.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | ENST00000335148.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | ENST00000538218.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | ENST00000538218.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | ENST00000538218.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | ENST00000538218.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | ENST00000538218.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | ENST00000538218.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | ENST00000538218.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | ENST00000538218.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | ENST00000538218.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | ENST00000538218.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | ENST00000538218.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | ENST00000538218.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | ENST00000538218.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | ENST00000539974.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | ENST00000539974.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | ENST00000539974.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | ENST00000544191.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | ENST00000544191.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | ENST00000544191.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | ENST00000544191.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | ENST00000545979.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | ENST00000545979.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | NM_001039481.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | NM_001039481.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | NM_001039481.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | NM_001039481.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | NM_001039481.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | NM_001039481.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | NM_001039481.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | NM_018638.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | NM_018638.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | NM_018638.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | NM_018638.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | NM_018638.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | NM_018638.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | NM_018638.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | NM_018638.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | NM_018638.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | NM_018638.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | NM_018638.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | NM_018638.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | NM_018638.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | NM_018638.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007070.hg.1 | NM_018638.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | ENST00000266517.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | ENST00000266517.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | ENST00000266517.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | ENST00000266517.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | ENST00000266517.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | ENST00000266517.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | ENST00000266517.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | ENST00000266517.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | ENST00000266517.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | ENST00000266517.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | ENST00000266517.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | ENST00000266517.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | ENST00000266517.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | ENST00000266517.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | ENST00000266517.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | ENST00000266517.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | ENST00000335148.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | ENST00000335148.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | ENST00000335148.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | ENST00000335148.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | ENST00000335148.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | ENST00000335148.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | ENST00000335148.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | ENST00000335148.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | ENST00000538218.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | ENST00000538218.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | ENST00000538218.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | ENST00000538218.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | ENST00000538218.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | ENST00000538218.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | ENST00000538218.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | ENST00000538218.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | ENST00000538218.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | ENST00000538218.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | ENST00000538218.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | ENST00000538218.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | ENST00000538218.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | ENST00000539974.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | ENST00000539974.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | ENST00000539974.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | ENST00000544191.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | ENST00000544191.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | ENST00000544191.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | ENST00000544191.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | ENST00000545979.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | ENST00000545979.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | NM_001039481.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | NM_001039481.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | NM_001039481.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | NM_001039481.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | NM_001039481.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | NM_001039481.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | NM_001039481.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | NM_018638.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | NM_018638.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | NM_018638.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | NM_018638.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | NM_018638.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | NM_018638.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | NM_018638.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | NM_018638.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | NM_018638.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | NM_018638.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | NM_018638.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | NM_018638.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | NM_018638.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | NM_018638.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007070.hg.1 | NM_018638.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200149569.hg.1 | ENST00000266517.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200149569.hg.1 | ENST00000335148.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200149569.hg.1 | ENST00000538218.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200149569.hg.1 | ENST00000539974.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200149569.hg.1 | NM_001039481.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200149569.hg.1 | NM_018638.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200149569.hg.1 | ENST00000266517.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200149569.hg.1 | ENST00000335148.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200149569.hg.1 | ENST00000538218.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200149569.hg.1 | ENST00000539974.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200149569.hg.1 | NM_001039481.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200149569.hg.1 | NM_018638.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200149569.hg.1 | TC1200007070.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1200149570.hg.1 | ENST00000335148.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200149570.hg.1 | NM_001039481.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200149570.hg.1 | ENST00000335148.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200149570.hg.1 | NM_001039481.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200149570.hg.1 | TC1200007070.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1200149571.hg.1 | ENST00000335148.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200149571.hg.1 | NM_001039481.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200149571.hg.1 | ENST00000335148.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200149571.hg.1 | NM_001039481.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200149571.hg.1 | TC1200007070.hg.1 | Clariom D TC |

ETNK1
Genomic Map