Assay Details
| Gene Symbol: HSPG2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291860.1 | NP_001278789.1 | 40 - 41 | 5093 | 136 | |
| NM_005529.6 | NP_005520.4 | 40 - 41 | 5090 | 136 | ||
| XM_011541318.2 | XP_011539620.1 | 45 - 46 | 5574 | 136 | ||
| XM_017001120.1 | XP_016856609.1 | 41 - 42 | 5220 | 136 | ||
| XM_017001121.1 | XP_016856610.1 | 40 - 41 | 5169 | 136 | ||
| XM_017001122.1 | XP_016856611.1 | 40 - 41 | 5166 | 136 | ||
| GenBank mRNA | M85289.1 | - | 40 - 41 | 5090 | 136 | |
More Information
Updated Mapping Information
Set Membership: |
|

HSPG2
Genomic Map