Assay Details
| Gene Symbol: ZNF713 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_182633.2 | NP_872439.2 | 2 - 3 | 533 | 82 | |
| GenBank mRNA | AL539604.3 | - | 2 - 3 | 328 | 82 | |
| BC094796.1 | - | 2 - 3 | 354 | 82 | ||
| DA224934.1 | - | 2 - 3 | 523 | 82 | ||
| DA690512.1 | - | 2 - 3 | 353 | 82 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC07003324.hg.1
Additional Information:
This assay is in the 5'UTR region of the NM_182633.2 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR07004959.hg.1 | TC07003324.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR07004959.hg.1 | ENST00000429591.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07004959.hg.1 | NM_182633.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07004948.hg.1 | TC07003324.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR07004948.hg.1 | ENST00000429591.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07004948.hg.1 | ENST00000482436.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07004948.hg.1 | NM_182633.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07004952.hg.1 | TC07003324.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR07004952.hg.1 | ENST00000429591.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07004952.hg.1 | ENST00000482436.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07004952.hg.1 | NM_182633.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013361.hg.1 | ENST00000411863.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013361.hg.1 | ENST00000411863.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013361.hg.1 | ENST00000411863.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013361.hg.1 | ENST00000411863.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013361.hg.1 | ENST00000411863.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013361.hg.1 | ENST00000411863.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013361.hg.1 | ENST00000411863.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013361.hg.1 | ENST00000411863.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013361.hg.1 | ENST00000411863.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013361.hg.1 | ENST00000411863.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013361.hg.1 | ENST00000429591.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013361.hg.1 | ENST00000429591.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013361.hg.1 | ENST00000429591.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013361.hg.1 | ENST00000429591.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013361.hg.1 | ENST00000429591.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013361.hg.1 | ENST00000429591.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013361.hg.1 | ENST00000429591.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013361.hg.1 | ENST00000429591.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013361.hg.1 | ENST00000429591.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013361.hg.1 | ENST00000429591.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013361.hg.1 | ENST00000429591.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013361.hg.1 | ENST00000429591.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013361.hg.1 | ENST00000429591.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013361.hg.1 | ENST00000466630.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013361.hg.1 | ENST00000466630.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013361.hg.1 | ENST00000466630.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013361.hg.1 | ENST00000466630.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013361.hg.1 | ENST00000466630.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013361.hg.1 | ENST00000466630.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013361.hg.1 | ENST00000466630.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013361.hg.1 | ENST00000466630.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013361.hg.1 | ENST00000466630.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013361.hg.1 | ENST00000466630.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013361.hg.1 | ENST00000466630.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013361.hg.1 | ENST00000466630.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013361.hg.1 | ENST00000466630.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013361.hg.1 | ENST00000466630.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013361.hg.1 | ENST00000466630.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013361.hg.1 | ENST00000482436.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013361.hg.1 | ENST00000482436.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013361.hg.1 | ENST00000482436.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013361.hg.1 | ENST00000482436.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013361.hg.1 | ENST00000482436.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013361.hg.1 | ENST00000482436.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013361.hg.1 | ENST00000484120.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013361.hg.1 | ENST00000484120.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013361.hg.1 | ENST00000484120.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013361.hg.1 | ENST00000633730.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013361.hg.1 | ENST00000633730.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013361.hg.1 | ENST00000633730.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013361.hg.1 | ENST00000633730.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013361.hg.1 | ENST00000633730.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013361.hg.1 | ENST00000633730.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013361.hg.1 | NM_182633.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013361.hg.1 | NM_182633.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013361.hg.1 | NM_182633.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013361.hg.1 | NM_182633.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013361.hg.1 | NM_182633.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013361.hg.1 | NM_182633.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013361.hg.1 | NM_182633.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013361.hg.1 | NM_182633.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013361.hg.1 | NM_182633.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013361.hg.1 | NM_182633.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013361.hg.1 | NM_182633.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013361.hg.1 | NM_182633.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013361.hg.1 | NM_182633.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013361.hg.1 | NM_182633.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07004955.hg.1 | TC07003324.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR07004955.hg.1 | ENST00000429591.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07004955.hg.1 | ENST00000482436.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07004955.hg.1 | NM_182633.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07004950.hg.1 | TC07003324.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR07004950.hg.1 | ENST00000429591.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07004950.hg.1 | ENST00000482436.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07004950.hg.1 | NM_182633.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700196779.hg.1 | ENST00000411863.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700196779.hg.1 | ENST00000429591.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700196779.hg.1 | ENST00000466630.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700196779.hg.1 | ENST00000484120.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700196779.hg.1 | NM_182633.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700196779.hg.1 | ENST00000411863.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700196779.hg.1 | ENST00000429591.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700196779.hg.1 | ENST00000466630.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700196779.hg.1 | ENST00000484120.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700196779.hg.1 | NM_182633.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700196779.hg.1 | TC0700013361.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0700196773.hg.1 | NM_182633.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700196773.hg.1 | NM_182633.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700196773.hg.1 | TC0700013361.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0700013361.hg.1 | ENST00000411863.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013361.hg.1 | ENST00000411863.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013361.hg.1 | ENST00000411863.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013361.hg.1 | ENST00000411863.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013361.hg.1 | ENST00000411863.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013361.hg.1 | ENST00000411863.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013361.hg.1 | ENST00000411863.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013361.hg.1 | ENST00000411863.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013361.hg.1 | ENST00000411863.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013361.hg.1 | ENST00000411863.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013361.hg.1 | ENST00000429591.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013361.hg.1 | ENST00000429591.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013361.hg.1 | ENST00000429591.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013361.hg.1 | ENST00000429591.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013361.hg.1 | ENST00000429591.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013361.hg.1 | ENST00000429591.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013361.hg.1 | ENST00000429591.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013361.hg.1 | ENST00000429591.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013361.hg.1 | ENST00000429591.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013361.hg.1 | ENST00000429591.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013361.hg.1 | ENST00000429591.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013361.hg.1 | ENST00000429591.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013361.hg.1 | ENST00000429591.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013361.hg.1 | ENST00000466630.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013361.hg.1 | ENST00000466630.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013361.hg.1 | ENST00000466630.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013361.hg.1 | ENST00000466630.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013361.hg.1 | ENST00000466630.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013361.hg.1 | ENST00000466630.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013361.hg.1 | ENST00000466630.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013361.hg.1 | ENST00000466630.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013361.hg.1 | ENST00000466630.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013361.hg.1 | ENST00000466630.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013361.hg.1 | ENST00000466630.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013361.hg.1 | ENST00000466630.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013361.hg.1 | ENST00000466630.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013361.hg.1 | ENST00000466630.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013361.hg.1 | ENST00000466630.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013361.hg.1 | ENST00000482436.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013361.hg.1 | ENST00000482436.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013361.hg.1 | ENST00000482436.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013361.hg.1 | ENST00000482436.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013361.hg.1 | ENST00000482436.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013361.hg.1 | ENST00000482436.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013361.hg.1 | ENST00000484120.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013361.hg.1 | ENST00000484120.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013361.hg.1 | ENST00000484120.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013361.hg.1 | ENST00000633730.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013361.hg.1 | ENST00000633730.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013361.hg.1 | ENST00000633730.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013361.hg.1 | ENST00000633730.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013361.hg.1 | ENST00000633730.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013361.hg.1 | ENST00000633730.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013361.hg.1 | NM_182633.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013361.hg.1 | NM_182633.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013361.hg.1 | NM_182633.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013361.hg.1 | NM_182633.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013361.hg.1 | NM_182633.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013361.hg.1 | NM_182633.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013361.hg.1 | NM_182633.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013361.hg.1 | NM_182633.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013361.hg.1 | NM_182633.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013361.hg.1 | NM_182633.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013361.hg.1 | NM_182633.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013361.hg.1 | NM_182633.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013361.hg.1 | NM_182633.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013361.hg.1 | NM_182633.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700196775.hg.1 | ENST00000429591.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700196775.hg.1 | ENST00000466630.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700196775.hg.1 | NM_182633.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700196775.hg.1 | ENST00000429591.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700196775.hg.1 | ENST00000466630.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700196775.hg.1 | NM_182633.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700196775.hg.1 | TC0700013361.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0700196778.hg.1 | ENST00000411863.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700196778.hg.1 | ENST00000429591.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700196778.hg.1 | ENST00000466630.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700196778.hg.1 | ENST00000484120.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700196778.hg.1 | NM_182633.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700196778.hg.1 | ENST00000411863.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700196778.hg.1 | ENST00000429591.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700196778.hg.1 | ENST00000466630.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700196778.hg.1 | ENST00000484120.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700196778.hg.1 | NM_182633.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700196778.hg.1 | TC0700013361.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0700196776.hg.1 | ENST00000411863.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700196776.hg.1 | ENST00000429591.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700196776.hg.1 | ENST00000466630.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700196776.hg.1 | NM_182633.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700196776.hg.1 | ENST00000411863.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700196776.hg.1 | ENST00000429591.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700196776.hg.1 | ENST00000466630.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700196776.hg.1 | NM_182633.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700196776.hg.1 | TC0700013361.hg.1 | Clariom D TC |

ZNF713
Genomic Map