| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007919.hg.1 |
ENST00000465453.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008245.hg.1 |
ENST00000511924.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013362.hg.1 |
ENST00000285298.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013362.hg.1 |
ENST00000285298.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013362.hg.1 |
ENST00000285298.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013362.hg.1 |
ENST00000285298.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013362.hg.1 |
ENST00000285298.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013362.hg.1 |
ENST00000285298.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013362.hg.1 |
ENST00000443449.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013362.hg.1 |
ENST00000443449.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013362.hg.1 |
ENST00000443449.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013362.hg.1 |
NM_015969.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013362.hg.1 |
NM_015969.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013362.hg.1 |
NM_015969.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700013362.hg.1 |
ENST00000285298.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013362.hg.1 |
ENST00000285298.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013362.hg.1 |
ENST00000285298.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013362.hg.1 |
ENST00000285298.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013362.hg.1 |
ENST00000285298.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013362.hg.1 |
ENST00000285298.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013362.hg.1 |
ENST00000443449.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013362.hg.1 |
ENST00000443449.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013362.hg.1 |
ENST00000443449.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013362.hg.1 |
NM_015969.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700013362.hg.1 |
NM_015969.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700013362.hg.1 |
NM_015969.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00008245.hg.1 |
ENST00000511924.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700196804.hg.1 |
ENST00000285298.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700196804.hg.1 |
ENST00000443449.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700196804.hg.1 |
NM_015969.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700196804.hg.1 |
ENST00000285298.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700196804.hg.1 |
ENST00000443449.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700196804.hg.1 |
NM_015969.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700196804.hg.1 |
TC0700013362.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0700196805.hg.1 |
ENST00000285298.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700196805.hg.1 |
ENST00000443449.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700196805.hg.1 |
NM_015969.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700196805.hg.1 |
ENST00000285298.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700196805.hg.1 |
ENST00000443449.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700196805.hg.1 |
NM_015969.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700196805.hg.1 |
TC0700013362.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0700196799.hg.1 |
ENST00000285298.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700196799.hg.1 |
ENST00000285298.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700196799.hg.1 |
TC0700013362.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0300007919.hg.1 |
ENST00000465453.1 |
Ensembl |