Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | SLC8A1 antisense RNA 1 | SLC8A1-AS1 | -29.6kb | Anti-sense | Chr. 2 - 39917634 - 40255209 on NCBI Build GRCh38 | |
| Downstream | uncharacterized LOC388942 | LOC388942 | 1.6mb | Anti-sense | Chr. 2 - 41877555 - 41894046 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map