Assay Details
| Gene Symbol: FEZ2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042548.1 | NP_001036013.1 | 6 - 7 | 1021 | 151 | |
| XM_006712153.2 | XP_006712216.1 | 6 - 7 | 843 | 151 | ||
| GenBank mRNA | BC089390.1 | - | 6 - 7 | 967 | 151 | |
More Information
Set Membership: |
|

FEZ2
Genomic Map