Assay Details
| Gene Symbol: TTLL12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_015140.3 | NP_055955.1 | 6 - 7 | 982 | 122 | |
| GenBank mRNA | BC001070.2 | - | 6 - 7 | 946 | 3504490 | 122 |
| CT841516.1 | - | 6 - 7 | 923 | 122 | ||
| D63487.1 | - | 6 - 7 | 904 | 122 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC22000859.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | ENST00000216129.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | ENST00000216129.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | ENST00000216129.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | ENST00000216129.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | ENST00000216129.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | ENST00000216129.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | ENST00000216129.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | ENST00000216129.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | ENST00000216129.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | ENST00000216129.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | ENST00000216129.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | ENST00000216129.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | ENST00000216129.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | ENST00000216129.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | ENST00000216129.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | ENST00000216129.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | ENST00000216129.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | ENST00000216129.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | ENST00000216129.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | ENST00000216129.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | ENST00000216129.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | ENST00000484118.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | ENST00000484118.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | ENST00000484118.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | ENST00000484711.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | ENST00000484711.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | ENST00000484711.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | ENST00000484711.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | ENST00000484711.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | ENST00000484711.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | ENST00000484711.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | ENST00000484711.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | ENST00000494035.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | ENST00000494035.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | ENST00000494035.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | ENST00000494035.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | ENST00000494035.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | ENST00000494035.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | ENST00000494035.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | NM_015140.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | NM_015140.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | NM_015140.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | NM_015140.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | NM_015140.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | NM_015140.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | NM_015140.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | NM_015140.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | NM_015140.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | NM_015140.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | NM_015140.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | NM_015140.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | NM_015140.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | NM_015140.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | NM_015140.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | NM_015140.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | NM_015140.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | NM_015140.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | NM_015140.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | NM_015140.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | NM_015140.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22014526.hg.1 | ENST00000216129.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22014526.hg.1 | ENST00000423379.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22014526.hg.1 | NM_015140.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22014526.hg.1 | TC22000859.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR2200164616.hg.1 | TC2200008906.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200164616.hg.1 | ENST00000216129.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200164616.hg.1 | ENST00000484118.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200164616.hg.1 | NM_015140.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200164616.hg.1 | ENST00000216129.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200164616.hg.1 | ENST00000484118.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200164616.hg.1 | NM_015140.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | ENST00000216129.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | ENST00000216129.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | ENST00000216129.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | ENST00000216129.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | ENST00000216129.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | ENST00000216129.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | ENST00000216129.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | ENST00000216129.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | ENST00000216129.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | ENST00000216129.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | ENST00000216129.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | ENST00000216129.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | ENST00000216129.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | ENST00000216129.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | ENST00000216129.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | ENST00000216129.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | ENST00000216129.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | ENST00000216129.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | ENST00000216129.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | ENST00000216129.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | ENST00000216129.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | ENST00000484118.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | ENST00000484118.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | ENST00000484118.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | ENST00000484711.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | ENST00000484711.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | ENST00000484711.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | ENST00000484711.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | ENST00000484711.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | ENST00000484711.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | ENST00000484711.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | ENST00000484711.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | ENST00000494035.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | ENST00000494035.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | ENST00000494035.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | ENST00000494035.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | ENST00000494035.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | ENST00000494035.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | ENST00000494035.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | NM_015140.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | NM_015140.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | NM_015140.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | NM_015140.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | NM_015140.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | NM_015140.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | NM_015140.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | NM_015140.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | NM_015140.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | NM_015140.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | NM_015140.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | NM_015140.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | NM_015140.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | NM_015140.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | NM_015140.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | NM_015140.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | NM_015140.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | NM_015140.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | NM_015140.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | NM_015140.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | NM_015140.3 | Refseq |

TTLL12
Genomic Map