Assay Details
| Gene Symbol: RHBDF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005498.3 | NP_001005498.2 | 6 - 7 | 1021 | 234 | |
| NM_024599.5 | NP_078875.4 | 7 - 8 | 1183 | 234 | ||
| XM_005257669.3 | XP_005257726.1 | 7 - 8 | 1544 | 234 | ||
| XM_005257670.1 | XP_005257727.1 | 7 - 8 | 1053 | 234 | ||
| XM_011525249.2 | XP_011523551.1 | 7 - 8 | 2500 | 234 | ||
| XM_011525250.2 | XP_011523552.1 | 8 - 9 | 1210 | 234 | ||
| XM_011525251.2 | XP_011523553.1 | 7 - 8 | 1027 | 234 | ||
| XM_017025079.1 | XP_016880568.1 | 7 - 8 | 1457 | 234 | ||
| GenBank mRNA | AK025994.1 | - | 2 - 3 | 207 | 234 | |
| AK057882.1 | - | 4 - 5 | 877 | 234 | ||
| AK074034.1 | - | 5 - 6 | 1488 | 234 | ||
| AK292135.1 | - | 7 - 8 | 1176 | 234 | ||
| AK292166.1 | - | 6 - 7 | 976 | 234 | ||
| AK303434.1 | - | 5 - 6 | 837 | 234 | ||
| AL833680.1 | - | 1 - 2 | 76 | 234 | ||
| AY500247.1 | - | 6 - 7 | 872 | 234 | ||
| BC016034.1 | - | 7 - 8 | 1163 | 3910543 | 234 | |
| BC035829.1 | - | 2 - 3 | 269 | 234 | ||
More Information
Set Membership: |
|

RHBDF2
Genomic Map