Assay Details
| Gene Symbol: LOC100132593 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK098270.1 | - | 1 | 227 | 87 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | myosin light chain 10 | MYL10 |
MYLC2PL, PLRLC |
-545.9kb | Anti-sense | Chr. 7 - 101613325 - 101629296 on NCBI Build GRCh38 |
| Downstream | SH2B adaptor protein 2 | SH2B2 | APS | 107.6kb | Sense | Chr. 7 - 102284526 - 102321711 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

LOC100132593
Genomic Map