Assay Details
| Gene Symbol: UBE2Q1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017582.6 | NP_060052.3 | 2 - 3 | 522 | 69 | |
| GenBank mRNA | AK293986.1 | - | 2 - 3 | 145 | 69 | |
| AY112698.1 | - | 2 - 3 | 497 | 69 | ||
| AY948200.1 | - | 2 - 3 | 505 | 69 | ||
| BC070158.1 | - | 1 - 2 | 47 | 69 | ||
| BC107057.2 | - | 3 - 4 | 190 | 69 | ||
| BC148185.2 | - | 2 - 3 | 199 | 69 | ||
| BQ278712.1 | - | 522 | 69 | |||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01003293.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK293986.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01050804.hg.1 | TC01003293.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01050804.hg.1 | ENST00000292211.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01050804.hg.1 | ENST00000497453.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01050804.hg.1 | NM_017582.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015918.hg.1 | ENST00000292211.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015918.hg.1 | ENST00000292211.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015918.hg.1 | ENST00000292211.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015918.hg.1 | ENST00000292211.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015918.hg.1 | ENST00000292211.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015918.hg.1 | ENST00000292211.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015918.hg.1 | ENST00000292211.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015918.hg.1 | ENST00000292211.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015918.hg.1 | ENST00000292211.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015918.hg.1 | ENST00000292211.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015918.hg.1 | ENST00000292211.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015918.hg.1 | ENST00000292211.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015918.hg.1 | ENST00000292211.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015918.hg.1 | ENST00000292211.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015918.hg.1 | ENST00000292211.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015918.hg.1 | ENST00000292211.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015918.hg.1 | ENST00000292211.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015918.hg.1 | ENST00000292211.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015918.hg.1 | ENST00000292211.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015918.hg.1 | ENST00000467683.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015918.hg.1 | ENST00000467683.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015918.hg.1 | ENST00000467683.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015918.hg.1 | ENST00000467683.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015918.hg.1 | ENST00000467683.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015918.hg.1 | ENST00000467683.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015918.hg.1 | ENST00000467683.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015918.hg.1 | ENST00000474181.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015918.hg.1 | ENST00000474181.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015918.hg.1 | ENST00000474181.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015918.hg.1 | ENST00000474181.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015918.hg.1 | ENST00000474181.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015918.hg.1 | ENST00000476612.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015918.hg.1 | ENST00000476612.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015918.hg.1 | ENST00000476612.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015918.hg.1 | ENST00000476612.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015918.hg.1 | ENST00000483639.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015918.hg.1 | ENST00000483639.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015918.hg.1 | ENST00000483639.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015918.hg.1 | ENST00000483639.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015918.hg.1 | ENST00000483639.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015918.hg.1 | ENST00000483639.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015918.hg.1 | ENST00000483639.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015918.hg.1 | ENST00000483639.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015918.hg.1 | ENST00000491572.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015918.hg.1 | ENST00000491572.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015918.hg.1 | ENST00000491572.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015918.hg.1 | ENST00000491572.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015918.hg.1 | ENST00000491572.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015918.hg.1 | ENST00000497453.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015918.hg.1 | ENST00000497453.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015918.hg.1 | ENST00000497453.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015918.hg.1 | ENST00000497453.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015918.hg.1 | ENST00000497453.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015918.hg.1 | ENST00000497453.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015918.hg.1 | ENST00000497453.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015918.hg.1 | ENST00000497453.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015918.hg.1 | ENST00000497453.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015918.hg.1 | NM_017582.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015918.hg.1 | NM_017582.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015918.hg.1 | NM_017582.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015918.hg.1 | NM_017582.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015918.hg.1 | NM_017582.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015918.hg.1 | NM_017582.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015918.hg.1 | NM_017582.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015918.hg.1 | NM_017582.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015918.hg.1 | NM_017582.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015918.hg.1 | NM_017582.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015918.hg.1 | NM_017582.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015918.hg.1 | NM_017582.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015918.hg.1 | NM_017582.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015918.hg.1 | NM_017582.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015918.hg.1 | NM_017582.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015918.hg.1 | NM_017582.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015918.hg.1 | NM_017582.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015918.hg.1 | NM_017582.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015918.hg.1 | NM_017582.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01050806.hg.1 | TC01003293.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01050806.hg.1 | ENST00000292211.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01050806.hg.1 | NM_017582.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01050807.hg.1 | TC01003293.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01050807.hg.1 | ENST00000292211.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01050807.hg.1 | NM_017582.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01050803.hg.1 | TC01003293.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01050803.hg.1 | ENST00000292211.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01050803.hg.1 | ENST00000497453.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01050803.hg.1 | NM_017582.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01050805.hg.1 | TC01003293.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01050805.hg.1 | ENST00000292211.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01050805.hg.1 | ENST00000497453.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01050805.hg.1 | NM_017582.6 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100219837.hg.1 | ENST00000292211.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100219837.hg.1 | NM_017582.6 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100219837.hg.1 | ENST00000292211.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100219837.hg.1 | NM_017582.6 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100219837.hg.1 | TC0100015918.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100219836.hg.1 | ENST00000292211.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100219836.hg.1 | ENST00000497453.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100219836.hg.1 | NM_017582.6 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100219836.hg.1 | ENST00000292211.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100219836.hg.1 | ENST00000497453.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100219836.hg.1 | NM_017582.6 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100219836.hg.1 | TC0100015918.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100219835.hg.1 | ENST00000292211.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100219835.hg.1 | ENST00000497453.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100219835.hg.1 | NM_017582.6 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100219835.hg.1 | ENST00000292211.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100219835.hg.1 | ENST00000497453.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100219835.hg.1 | NM_017582.6 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100219835.hg.1 | TC0100015918.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100015918.hg.1 | ENST00000292211.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015918.hg.1 | ENST00000292211.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015918.hg.1 | ENST00000292211.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015918.hg.1 | ENST00000292211.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015918.hg.1 | ENST00000292211.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015918.hg.1 | ENST00000292211.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015918.hg.1 | ENST00000292211.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015918.hg.1 | ENST00000292211.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015918.hg.1 | ENST00000292211.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015918.hg.1 | ENST00000292211.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015918.hg.1 | ENST00000292211.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015918.hg.1 | ENST00000292211.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015918.hg.1 | ENST00000292211.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015918.hg.1 | ENST00000292211.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015918.hg.1 | ENST00000292211.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015918.hg.1 | ENST00000292211.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015918.hg.1 | ENST00000292211.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015918.hg.1 | ENST00000292211.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015918.hg.1 | ENST00000292211.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015918.hg.1 | ENST00000467683.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015918.hg.1 | ENST00000467683.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015918.hg.1 | ENST00000467683.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015918.hg.1 | ENST00000467683.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015918.hg.1 | ENST00000467683.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015918.hg.1 | ENST00000467683.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015918.hg.1 | ENST00000467683.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015918.hg.1 | ENST00000474181.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015918.hg.1 | ENST00000474181.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015918.hg.1 | ENST00000474181.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015918.hg.1 | ENST00000474181.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015918.hg.1 | ENST00000474181.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015918.hg.1 | ENST00000476612.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015918.hg.1 | ENST00000476612.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015918.hg.1 | ENST00000476612.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015918.hg.1 | ENST00000476612.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015918.hg.1 | ENST00000483639.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015918.hg.1 | ENST00000483639.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015918.hg.1 | ENST00000483639.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015918.hg.1 | ENST00000483639.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015918.hg.1 | ENST00000483639.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015918.hg.1 | ENST00000483639.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015918.hg.1 | ENST00000483639.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015918.hg.1 | ENST00000483639.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015918.hg.1 | ENST00000491572.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015918.hg.1 | ENST00000491572.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015918.hg.1 | ENST00000491572.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015918.hg.1 | ENST00000491572.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015918.hg.1 | ENST00000491572.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015918.hg.1 | ENST00000497453.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015918.hg.1 | ENST00000497453.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015918.hg.1 | ENST00000497453.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015918.hg.1 | ENST00000497453.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015918.hg.1 | ENST00000497453.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015918.hg.1 | ENST00000497453.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015918.hg.1 | ENST00000497453.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015918.hg.1 | ENST00000497453.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015918.hg.1 | ENST00000497453.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015918.hg.1 | NM_017582.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015918.hg.1 | NM_017582.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015918.hg.1 | NM_017582.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015918.hg.1 | NM_017582.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015918.hg.1 | NM_017582.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015918.hg.1 | NM_017582.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015918.hg.1 | NM_017582.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015918.hg.1 | NM_017582.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015918.hg.1 | NM_017582.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015918.hg.1 | NM_017582.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015918.hg.1 | NM_017582.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015918.hg.1 | NM_017582.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015918.hg.1 | NM_017582.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015918.hg.1 | NM_017582.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015918.hg.1 | NM_017582.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015918.hg.1 | NM_017582.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015918.hg.1 | NM_017582.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015918.hg.1 | NM_017582.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015918.hg.1 | NM_017582.6 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100219834.hg.1 | ENST00000292211.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100219834.hg.1 | ENST00000497453.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100219834.hg.1 | NM_017582.6 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100219834.hg.1 | ENST00000292211.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100219834.hg.1 | ENST00000497453.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100219834.hg.1 | NM_017582.6 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100219834.hg.1 | TC0100015918.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100219838.hg.1 | ENST00000292211.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100219838.hg.1 | NM_017582.6 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100219838.hg.1 | ENST00000292211.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100219838.hg.1 | NM_017582.6 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100219838.hg.1 | TC0100015918.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 175938 | - | - |

UBE2Q1
Genomic Map