Assay Details
| Gene Symbol: UBE2Q1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017582.6 | NP_060052.3 | 7 - 8 | 970 | 85 | |
| GenBank mRNA | AK056388.1 | - | 3 - 4 | 270 | 85 | |
| AK293986.1 | - | 7 - 8 | 593 | 85 | ||
| AY112698.1 | - | 7 - 8 | 945 | 85 | ||
| AY948200.1 | - | 7 - 8 | 953 | 85 | ||
| BC000848.1 | - | 3 - 4 | 304 | 85 | ||
| BC009286.2 | - | 2 - 3 | 108 | 85 | ||
| BC061583.1 | - | 3 - 4 | 282 | 85 | ||
| BC070158.1 | - | 6 - 7 | 495 | 85 | ||
| BC087836.1 | - | 5 - 6 | 381 | 85 | ||
| BC107057.2 | - | 8 - 9 | 638 | 85 | ||
| BC148185.2 | - | 7 - 8 | 647 | 85 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01003293.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100015918.hg.1 | ENST00000292211.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015918.hg.1 | ENST00000292211.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015918.hg.1 | ENST00000292211.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015918.hg.1 | ENST00000292211.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015918.hg.1 | ENST00000292211.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015918.hg.1 | ENST00000292211.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015918.hg.1 | ENST00000292211.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015918.hg.1 | ENST00000292211.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015918.hg.1 | ENST00000292211.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015918.hg.1 | ENST00000292211.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015918.hg.1 | ENST00000292211.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015918.hg.1 | ENST00000292211.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015918.hg.1 | ENST00000292211.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015918.hg.1 | ENST00000292211.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015918.hg.1 | ENST00000292211.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015918.hg.1 | ENST00000292211.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015918.hg.1 | ENST00000292211.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015918.hg.1 | ENST00000292211.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015918.hg.1 | ENST00000292211.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015918.hg.1 | ENST00000467683.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015918.hg.1 | ENST00000467683.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015918.hg.1 | ENST00000467683.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015918.hg.1 | ENST00000467683.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015918.hg.1 | ENST00000467683.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015918.hg.1 | ENST00000467683.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015918.hg.1 | ENST00000467683.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015918.hg.1 | ENST00000474181.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015918.hg.1 | ENST00000474181.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015918.hg.1 | ENST00000474181.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015918.hg.1 | ENST00000474181.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015918.hg.1 | ENST00000474181.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015918.hg.1 | ENST00000476612.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015918.hg.1 | ENST00000476612.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015918.hg.1 | ENST00000476612.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015918.hg.1 | ENST00000476612.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015918.hg.1 | ENST00000483639.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015918.hg.1 | ENST00000483639.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015918.hg.1 | ENST00000483639.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015918.hg.1 | ENST00000483639.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015918.hg.1 | ENST00000483639.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015918.hg.1 | ENST00000483639.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015918.hg.1 | ENST00000483639.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015918.hg.1 | ENST00000483639.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015918.hg.1 | ENST00000491572.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015918.hg.1 | ENST00000491572.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015918.hg.1 | ENST00000491572.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015918.hg.1 | ENST00000491572.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015918.hg.1 | ENST00000491572.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015918.hg.1 | ENST00000497453.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015918.hg.1 | ENST00000497453.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015918.hg.1 | ENST00000497453.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015918.hg.1 | ENST00000497453.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015918.hg.1 | ENST00000497453.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015918.hg.1 | ENST00000497453.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015918.hg.1 | ENST00000497453.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015918.hg.1 | ENST00000497453.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015918.hg.1 | ENST00000497453.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015918.hg.1 | NM_017582.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015918.hg.1 | NM_017582.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015918.hg.1 | NM_017582.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015918.hg.1 | NM_017582.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015918.hg.1 | NM_017582.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015918.hg.1 | NM_017582.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015918.hg.1 | NM_017582.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015918.hg.1 | NM_017582.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015918.hg.1 | NM_017582.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015918.hg.1 | NM_017582.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015918.hg.1 | NM_017582.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015918.hg.1 | NM_017582.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015918.hg.1 | NM_017582.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015918.hg.1 | NM_017582.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015918.hg.1 | NM_017582.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015918.hg.1 | NM_017582.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015918.hg.1 | NM_017582.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015918.hg.1 | NM_017582.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015918.hg.1 | NM_017582.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01050794.hg.1 | TC01003293.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01050794.hg.1 | ENST00000292211.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01050794.hg.1 | NM_017582.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015918.hg.1 | ENST00000292211.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015918.hg.1 | ENST00000292211.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015918.hg.1 | ENST00000292211.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015918.hg.1 | ENST00000292211.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015918.hg.1 | ENST00000292211.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015918.hg.1 | ENST00000292211.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015918.hg.1 | ENST00000292211.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015918.hg.1 | ENST00000292211.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015918.hg.1 | ENST00000292211.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015918.hg.1 | ENST00000292211.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015918.hg.1 | ENST00000292211.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015918.hg.1 | ENST00000292211.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015918.hg.1 | ENST00000292211.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015918.hg.1 | ENST00000292211.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015918.hg.1 | ENST00000292211.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015918.hg.1 | ENST00000292211.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015918.hg.1 | ENST00000292211.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015918.hg.1 | ENST00000292211.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015918.hg.1 | ENST00000292211.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015918.hg.1 | ENST00000467683.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015918.hg.1 | ENST00000467683.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015918.hg.1 | ENST00000467683.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015918.hg.1 | ENST00000467683.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015918.hg.1 | ENST00000467683.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015918.hg.1 | ENST00000467683.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015918.hg.1 | ENST00000467683.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015918.hg.1 | ENST00000474181.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015918.hg.1 | ENST00000474181.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015918.hg.1 | ENST00000474181.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015918.hg.1 | ENST00000474181.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015918.hg.1 | ENST00000474181.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015918.hg.1 | ENST00000476612.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015918.hg.1 | ENST00000476612.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015918.hg.1 | ENST00000476612.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015918.hg.1 | ENST00000476612.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015918.hg.1 | ENST00000483639.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015918.hg.1 | ENST00000483639.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015918.hg.1 | ENST00000483639.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015918.hg.1 | ENST00000483639.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015918.hg.1 | ENST00000483639.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015918.hg.1 | ENST00000483639.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015918.hg.1 | ENST00000483639.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015918.hg.1 | ENST00000483639.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015918.hg.1 | ENST00000491572.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015918.hg.1 | ENST00000491572.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015918.hg.1 | ENST00000491572.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015918.hg.1 | ENST00000491572.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015918.hg.1 | ENST00000491572.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015918.hg.1 | ENST00000497453.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015918.hg.1 | ENST00000497453.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015918.hg.1 | ENST00000497453.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015918.hg.1 | ENST00000497453.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015918.hg.1 | ENST00000497453.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015918.hg.1 | ENST00000497453.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015918.hg.1 | ENST00000497453.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015918.hg.1 | ENST00000497453.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015918.hg.1 | ENST00000497453.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015918.hg.1 | NM_017582.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015918.hg.1 | NM_017582.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015918.hg.1 | NM_017582.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015918.hg.1 | NM_017582.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015918.hg.1 | NM_017582.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015918.hg.1 | NM_017582.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015918.hg.1 | NM_017582.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015918.hg.1 | NM_017582.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015918.hg.1 | NM_017582.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015918.hg.1 | NM_017582.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015918.hg.1 | NM_017582.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015918.hg.1 | NM_017582.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015918.hg.1 | NM_017582.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015918.hg.1 | NM_017582.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015918.hg.1 | NM_017582.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015918.hg.1 | NM_017582.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015918.hg.1 | NM_017582.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015918.hg.1 | NM_017582.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015918.hg.1 | NM_017582.6 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100219825.hg.1 | ENST00000292211.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100219825.hg.1 | ENST00000474181.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100219825.hg.1 | ENST00000483639.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100219825.hg.1 | NM_017582.6 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100219825.hg.1 | ENST00000292211.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100219825.hg.1 | ENST00000474181.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100219825.hg.1 | ENST00000483639.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100219825.hg.1 | NM_017582.6 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100219825.hg.1 | TC0100015918.hg.1 | Clariom D TC |

UBE2Q1
Genomic Map