Assay Details
| Gene Symbol: LAMA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_005559.3 | NP_005550.2 | 31 - 32 | 4560 | 72 | |
| XM_011525655.2 | XP_011523957.1 | 31 - 32 | 4554 | 72 | ||
| XM_011525656.2 | XP_011523958.1 | 21 - 22 | 2975 | 72 | ||
| XM_011525657.1 | XP_011523959.1 | 31 - 32 | 4538 | 72 | ||
| GenBank mRNA | BC144590.1 | - | 30 - 31 | 5481 | 72 | |
| X58531.1 | - | 31 - 32 | 4560 | 72 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC18000309.hg.1
Set Membership: |
|

LAMA1
Genomic Map