Assay Details
| Gene Symbol: ILF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001267809.1 | NP_001254738.1 | 9 - 10 | 782 | 77 | |
| NM_004515.3 | NP_004506.2 | 9 - 10 | 763 | 77 | ||
| GenBank mRNA | AK223326.1 | - | 9 - 10 | 683 | 77 | |
| AK302214.1 | - | 9 - 10 | 726 | 77 | ||
| AK313364.1 | - | 9 - 10 | 722 | 77 | ||
| BC000382.2 | - | 9 - 10 | 688 | 2820505 | 77 | |
| BM800544.1 | - | 9 - 10 | 739 | 77 | ||
| HQ322261.1 | - | 8 - 9 | 657 | 77 | ||
| HQ322262.1 | - | 8 - 9 | 657 | 77 | ||
| U10323.1 | - | 9 - 10 | 696 | 77 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100015875.hg.1 | ENST00000361891.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015875.hg.1 | ENST00000361891.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015875.hg.1 | ENST00000361891.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015875.hg.1 | ENST00000361891.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015875.hg.1 | ENST00000361891.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015875.hg.1 | ENST00000361891.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015875.hg.1 | ENST00000361891.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015875.hg.1 | ENST00000361891.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015875.hg.1 | ENST00000361891.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015875.hg.1 | ENST00000361891.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015875.hg.1 | ENST00000361891.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015875.hg.1 | ENST00000361891.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015875.hg.1 | ENST00000361891.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015875.hg.1 | ENST00000361891.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015875.hg.1 | ENST00000361891.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015875.hg.1 | ENST00000361891.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015875.hg.1 | ENST00000361891.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015875.hg.1 | ENST00000361891.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015875.hg.1 | ENST00000368681.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015875.hg.1 | ENST00000368681.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015875.hg.1 | ENST00000368681.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015875.hg.1 | ENST00000368681.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015875.hg.1 | ENST00000368681.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015875.hg.1 | ENST00000368681.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015875.hg.1 | ENST00000368681.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015875.hg.1 | ENST00000368681.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015875.hg.1 | ENST00000368684.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015875.hg.1 | ENST00000368684.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015875.hg.1 | ENST00000368684.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015875.hg.1 | ENST00000368684.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015875.hg.1 | ENST00000368684.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015875.hg.1 | ENST00000368684.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015875.hg.1 | ENST00000480213.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015875.hg.1 | ENST00000480213.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015875.hg.1 | ENST00000480213.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015875.hg.1 | ENST00000480213.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015875.hg.1 | ENST00000480213.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015875.hg.1 | ENST00000480213.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015875.hg.1 | ENST00000480213.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015875.hg.1 | ENST00000480213.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015875.hg.1 | ENST00000615950.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015875.hg.1 | ENST00000615950.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015875.hg.1 | ENST00000615950.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015875.hg.1 | ENST00000615950.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015875.hg.1 | ENST00000615950.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015875.hg.1 | ENST00000615950.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015875.hg.1 | ENST00000615950.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015875.hg.1 | ENST00000615950.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015875.hg.1 | ENST00000615950.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015875.hg.1 | ENST00000615950.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015875.hg.1 | ENST00000615950.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015875.hg.1 | ENST00000615950.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015875.hg.1 | ENST00000615950.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015875.hg.1 | ENST00000615950.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015875.hg.1 | ENST00000615950.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015875.hg.1 | ENST00000615950.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015875.hg.1 | ENST00000615950.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015875.hg.1 | ENST00000615950.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015875.hg.1 | ENST00000615950.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015875.hg.1 | NM_001267809.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015875.hg.1 | NM_001267809.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015875.hg.1 | NM_001267809.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015875.hg.1 | NM_001267809.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015875.hg.1 | NM_001267809.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015875.hg.1 | NM_001267809.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015875.hg.1 | NM_001267809.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015875.hg.1 | NM_001267809.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015875.hg.1 | NM_001267809.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015875.hg.1 | NM_001267809.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015875.hg.1 | NM_001267809.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015875.hg.1 | NM_001267809.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015875.hg.1 | NM_001267809.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015875.hg.1 | NM_001267809.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015875.hg.1 | NM_001267809.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015875.hg.1 | NM_001267809.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015875.hg.1 | NM_001267809.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015875.hg.1 | NM_001267809.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015875.hg.1 | NM_001267809.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015875.hg.1 | NM_004515.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015875.hg.1 | NM_004515.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015875.hg.1 | NM_004515.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015875.hg.1 | NM_004515.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015875.hg.1 | NM_004515.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015875.hg.1 | NM_004515.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015875.hg.1 | NM_004515.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015875.hg.1 | NM_004515.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015875.hg.1 | NM_004515.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015875.hg.1 | NM_004515.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015875.hg.1 | NM_004515.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015875.hg.1 | NM_004515.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015875.hg.1 | NM_004515.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015875.hg.1 | NM_004515.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015875.hg.1 | NM_004515.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015875.hg.1 | NM_004515.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015875.hg.1 | NM_004515.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015875.hg.1 | NM_004515.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015875.hg.1 | NM_004515.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100219312.hg.1 | ENST00000361891.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100219312.hg.1 | ENST00000480213.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100219312.hg.1 | ENST00000615950.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100219312.hg.1 | NM_001267809.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100219312.hg.1 | NM_004515.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100219312.hg.1 | ENST00000361891.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100219312.hg.1 | ENST00000480213.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100219312.hg.1 | ENST00000615950.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100219312.hg.1 | NM_001267809.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100219312.hg.1 | NM_004515.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100219312.hg.1 | TC0100015875.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100015875.hg.1 | ENST00000361891.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015875.hg.1 | ENST00000361891.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015875.hg.1 | ENST00000361891.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015875.hg.1 | ENST00000361891.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015875.hg.1 | ENST00000361891.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015875.hg.1 | ENST00000361891.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015875.hg.1 | ENST00000361891.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015875.hg.1 | ENST00000361891.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015875.hg.1 | ENST00000361891.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015875.hg.1 | ENST00000361891.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015875.hg.1 | ENST00000361891.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015875.hg.1 | ENST00000361891.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015875.hg.1 | ENST00000361891.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015875.hg.1 | ENST00000361891.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015875.hg.1 | ENST00000361891.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015875.hg.1 | ENST00000361891.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015875.hg.1 | ENST00000361891.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015875.hg.1 | ENST00000361891.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015875.hg.1 | ENST00000368681.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015875.hg.1 | ENST00000368681.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015875.hg.1 | ENST00000368681.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015875.hg.1 | ENST00000368681.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015875.hg.1 | ENST00000368681.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015875.hg.1 | ENST00000368681.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015875.hg.1 | ENST00000368681.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015875.hg.1 | ENST00000368681.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015875.hg.1 | ENST00000368684.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015875.hg.1 | ENST00000368684.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015875.hg.1 | ENST00000368684.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015875.hg.1 | ENST00000368684.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015875.hg.1 | ENST00000368684.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015875.hg.1 | ENST00000368684.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015875.hg.1 | ENST00000480213.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015875.hg.1 | ENST00000480213.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015875.hg.1 | ENST00000480213.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015875.hg.1 | ENST00000480213.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015875.hg.1 | ENST00000480213.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015875.hg.1 | ENST00000480213.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015875.hg.1 | ENST00000480213.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015875.hg.1 | ENST00000480213.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015875.hg.1 | ENST00000615950.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015875.hg.1 | ENST00000615950.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015875.hg.1 | ENST00000615950.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015875.hg.1 | ENST00000615950.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015875.hg.1 | ENST00000615950.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015875.hg.1 | ENST00000615950.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015875.hg.1 | ENST00000615950.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015875.hg.1 | ENST00000615950.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015875.hg.1 | ENST00000615950.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015875.hg.1 | ENST00000615950.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015875.hg.1 | ENST00000615950.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015875.hg.1 | ENST00000615950.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015875.hg.1 | ENST00000615950.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015875.hg.1 | ENST00000615950.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015875.hg.1 | ENST00000615950.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015875.hg.1 | ENST00000615950.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015875.hg.1 | ENST00000615950.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015875.hg.1 | ENST00000615950.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015875.hg.1 | ENST00000615950.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015875.hg.1 | NM_001267809.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015875.hg.1 | NM_001267809.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015875.hg.1 | NM_001267809.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015875.hg.1 | NM_001267809.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015875.hg.1 | NM_001267809.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015875.hg.1 | NM_001267809.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015875.hg.1 | NM_001267809.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015875.hg.1 | NM_001267809.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015875.hg.1 | NM_001267809.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015875.hg.1 | NM_001267809.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015875.hg.1 | NM_001267809.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015875.hg.1 | NM_001267809.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015875.hg.1 | NM_001267809.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015875.hg.1 | NM_001267809.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015875.hg.1 | NM_001267809.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015875.hg.1 | NM_001267809.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015875.hg.1 | NM_001267809.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015875.hg.1 | NM_001267809.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015875.hg.1 | NM_001267809.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015875.hg.1 | NM_004515.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015875.hg.1 | NM_004515.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015875.hg.1 | NM_004515.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015875.hg.1 | NM_004515.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015875.hg.1 | NM_004515.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015875.hg.1 | NM_004515.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015875.hg.1 | NM_004515.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015875.hg.1 | NM_004515.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015875.hg.1 | NM_004515.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015875.hg.1 | NM_004515.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015875.hg.1 | NM_004515.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015875.hg.1 | NM_004515.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015875.hg.1 | NM_004515.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015875.hg.1 | NM_004515.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015875.hg.1 | NM_004515.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015875.hg.1 | NM_004515.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015875.hg.1 | NM_004515.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015875.hg.1 | NM_004515.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015875.hg.1 | NM_004515.3 | Refseq |

ILF2
Genomic Map