Assay Details
| Gene Symbol: DHRS13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144683.3 | NP_653284.2 | 1 - 2 | 252 | 124 | |
| GenBank mRNA | AY358566.1 | - | 1 - 2 | 252 | 124 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC17001312.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR17017497.hg.1 | ENST00000378895.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17017497.hg.1 | ENST00000394901.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17017497.hg.1 | NM_144683.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17017497.hg.1 | TC17001312.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR17017492.hg.1 | ENST00000378895.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17017492.hg.1 | ENST00000394901.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17017492.hg.1 | NM_144683.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17017492.hg.1 | TC17001312.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010221.hg.1 | ENST00000378895.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010221.hg.1 | ENST00000378895.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010221.hg.1 | ENST00000378895.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010221.hg.1 | ENST00000378895.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010221.hg.1 | ENST00000378895.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010221.hg.1 | ENST00000378895.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010221.hg.1 | ENST00000378895.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010221.hg.1 | ENST00000378895.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010221.hg.1 | ENST00000378895.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010221.hg.1 | ENST00000394901.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010221.hg.1 | ENST00000394901.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010221.hg.1 | ENST00000394901.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010221.hg.1 | ENST00000394901.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010221.hg.1 | ENST00000394901.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010221.hg.1 | ENST00000394901.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010221.hg.1 | ENST00000394901.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010221.hg.1 | ENST00000394901.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010221.hg.1 | ENST00000394901.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010221.hg.1 | ENST00000394901.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010221.hg.1 | ENST00000426464.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010221.hg.1 | ENST00000426464.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010221.hg.1 | ENST00000426464.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010221.hg.1 | ENST00000426464.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010221.hg.1 | ENST00000426464.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010221.hg.1 | ENST00000426464.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010221.hg.1 | ENST00000581759.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010221.hg.1 | ENST00000581759.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010221.hg.1 | ENST00000581759.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010221.hg.1 | ENST00000581974.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010221.hg.1 | ENST00000581974.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010221.hg.1 | NM_144683.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010221.hg.1 | NM_144683.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010221.hg.1 | NM_144683.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010221.hg.1 | NM_144683.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010221.hg.1 | NM_144683.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010221.hg.1 | NM_144683.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010221.hg.1 | NM_144683.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010221.hg.1 | NM_144683.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010221.hg.1 | NM_144683.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17017494.hg.1 | ENST00000378895.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17017494.hg.1 | ENST00000394901.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17017494.hg.1 | ENST00000426464.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17017494.hg.1 | NM_144683.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17017494.hg.1 | TC17001312.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1700180324.hg.1 | ENST00000378895.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700180324.hg.1 | ENST00000394901.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700180324.hg.1 | NM_144683.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700180324.hg.1 | ENST00000378895.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700180324.hg.1 | ENST00000394901.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700180324.hg.1 | NM_144683.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700180324.hg.1 | TC1700010221.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700180321.hg.1 | ENST00000378895.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700180321.hg.1 | ENST00000394901.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700180321.hg.1 | ENST00000426464.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700180321.hg.1 | NM_144683.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700180321.hg.1 | ENST00000378895.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700180321.hg.1 | ENST00000394901.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700180321.hg.1 | ENST00000426464.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700180321.hg.1 | NM_144683.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700180321.hg.1 | TC1700010221.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700180319.hg.1 | ENST00000378895.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700180319.hg.1 | ENST00000394901.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700180319.hg.1 | NM_144683.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700180319.hg.1 | ENST00000378895.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700180319.hg.1 | ENST00000394901.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700180319.hg.1 | NM_144683.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700180319.hg.1 | TC1700010221.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1700010221.hg.1 | ENST00000378895.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010221.hg.1 | ENST00000378895.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010221.hg.1 | ENST00000378895.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010221.hg.1 | ENST00000378895.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010221.hg.1 | ENST00000378895.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010221.hg.1 | ENST00000378895.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010221.hg.1 | ENST00000378895.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010221.hg.1 | ENST00000378895.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010221.hg.1 | ENST00000378895.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010221.hg.1 | ENST00000394901.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010221.hg.1 | ENST00000394901.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010221.hg.1 | ENST00000394901.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010221.hg.1 | ENST00000394901.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010221.hg.1 | ENST00000394901.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010221.hg.1 | ENST00000394901.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010221.hg.1 | ENST00000394901.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010221.hg.1 | ENST00000394901.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010221.hg.1 | ENST00000394901.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010221.hg.1 | ENST00000394901.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010221.hg.1 | ENST00000426464.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010221.hg.1 | ENST00000426464.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010221.hg.1 | ENST00000426464.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010221.hg.1 | ENST00000426464.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010221.hg.1 | ENST00000426464.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010221.hg.1 | ENST00000426464.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010221.hg.1 | ENST00000581759.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010221.hg.1 | ENST00000581759.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010221.hg.1 | ENST00000581759.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010221.hg.1 | ENST00000581974.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010221.hg.1 | ENST00000581974.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010221.hg.1 | NM_144683.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010221.hg.1 | NM_144683.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010221.hg.1 | NM_144683.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010221.hg.1 | NM_144683.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010221.hg.1 | NM_144683.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010221.hg.1 | NM_144683.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010221.hg.1 | NM_144683.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010221.hg.1 | NM_144683.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010221.hg.1 | NM_144683.3 | Refseq |

DHRS13
Genomic Map