Assay Details
| Gene Symbol: PRPF3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_004698.2 | NP_004689.1 | 14 - 15 | 2009 | 75 | |
| XM_011510129.1 | XP_011508431.1 | 15 - 16 | 2051 | 75 | ||
| XM_011510130.2 | XP_011508432.1 | 11 - 12 | 2571 | 75 | ||
| XM_017002790.1 | XP_016858279.1 | 11 - 12 | 1445 | 75 | ||
| GenBank mRNA | AF001947.1 | - | 13 - 14 | 1902 | 75 | |
| AF016370.1 | - | 14 - 15 | 1919 | 75 | ||
| AK222825.1 | - | 14 - 15 | 1935 | 75 | ||
| AK297139.1 | - | 14 - 15 | 1893 | 75 | ||
| AK297265.1 | - | 13 - 14 | 1788 | 75 | ||
| AK312891.1 | - | 14 - 15 | 1930 | 75 | ||
| BC000184.2 | - | 13 - 14 | 1896 | 3351519 | 75 | |
| BC001954.1 | - | 13 - 14 | 1904 | 2905308 | 75 | |
| BQ002848.1 | - | 2 - 3 | 75 | |||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01001185.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Molecular Function -
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01018213.hg.1 | TC01001185.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01018213.hg.1 | ENST00000324862.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01018213.hg.1 | ENST00000414970.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01018213.hg.1 | ENST00000467329.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01018213.hg.1 | NM_004698.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000324862.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000324862.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000324862.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000324862.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000324862.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000324862.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000324862.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000324862.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000324862.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000324862.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000324862.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000324862.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000324862.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000324862.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000324862.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000324862.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000324862.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000324862.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000324862.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000324862.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000467329.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000467329.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000467329.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000467329.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000467329.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000467329.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000467329.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000467329.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000467329.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000467329.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000467329.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000467329.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000467329.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000467329.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000467329.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000467329.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000467329.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000467329.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000467514.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000467514.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000467514.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000467514.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000470824.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000470824.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000470824.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000470824.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000476970.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000476970.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000476970.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000476970.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000476970.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000493553.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000493553.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000493553.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000493553.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000496202.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000496202.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000496202.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000496202.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000496202.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000496202.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000496202.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000496202.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000496202.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000496202.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | NM_004698.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | NM_004698.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | NM_004698.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | NM_004698.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | NM_004698.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | NM_004698.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | NM_004698.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | NM_004698.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | NM_004698.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | NM_004698.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | NM_004698.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | NM_004698.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | NM_004698.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | NM_004698.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | NM_004698.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | NM_004698.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | NM_004698.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | NM_004698.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | NM_004698.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | NM_004698.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | NM_004698.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01018211.hg.1 | TC01001185.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01018211.hg.1 | ENST00000324862.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01018211.hg.1 | ENST00000414970.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01018211.hg.1 | ENST00000467329.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01018211.hg.1 | NM_004698.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100171511.hg.1 | ENST00000324862.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100171511.hg.1 | ENST00000467329.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100171511.hg.1 | ENST00000476970.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100171511.hg.1 | NM_004698.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100171511.hg.1 | ENST00000324862.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100171511.hg.1 | ENST00000467329.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100171511.hg.1 | ENST00000476970.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100171511.hg.1 | NM_004698.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100171511.hg.1 | TC0100009902.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000324862.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000324862.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000324862.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000324862.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000324862.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000324862.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000324862.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000324862.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000324862.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000324862.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000324862.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000324862.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000324862.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000324862.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000324862.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000324862.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000324862.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000324862.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000324862.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000324862.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000467329.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000467329.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000467329.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000467329.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000467329.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000467329.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000467329.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000467329.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000467329.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000467329.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000467329.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000467329.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000467329.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000467329.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000467329.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000467329.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000467329.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000467329.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000467514.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000467514.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000467514.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000467514.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000470824.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000470824.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000470824.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000470824.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000476970.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000476970.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000476970.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000476970.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000476970.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000493553.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000493553.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000493553.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000493553.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000496202.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000496202.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000496202.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000496202.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000496202.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000496202.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000496202.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000496202.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000496202.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000496202.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | NM_004698.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | NM_004698.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | NM_004698.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | NM_004698.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | NM_004698.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | NM_004698.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | NM_004698.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | NM_004698.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | NM_004698.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | NM_004698.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | NM_004698.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | NM_004698.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | NM_004698.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | NM_004698.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | NM_004698.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | NM_004698.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | NM_004698.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | NM_004698.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | NM_004698.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | NM_004698.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | NM_004698.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100171513.hg.1 | ENST00000324862.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100171513.hg.1 | ENST00000467329.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100171513.hg.1 | ENST00000470824.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100171513.hg.1 | ENST00000476970.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100171513.hg.1 | NM_004698.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100171513.hg.1 | ENST00000324862.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100171513.hg.1 | ENST00000467329.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100171513.hg.1 | ENST00000470824.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100171513.hg.1 | ENST00000476970.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100171513.hg.1 | NM_004698.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100171513.hg.1 | TC0100009902.hg.1 | Clariom D TC |

PRPF3
Genomic Map