Assay Details
| Gene Symbol: CBX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001127228.1 | NP_001120700.1 | 2 - 3 | 430 | 74 | |
| NM_006807.4 | NP_006798.1 | 2 - 3 | 620 | 74 | ||
| GenBank mRNA | AK312615.1 | - | 2 - 3 | 369 | 74 | |
| BC002609.1 | - | 2 - 3 | 354 | 3140815 | 74 | |
| BC021302.2 | - | 2 - 3 | 421 | 5013102 | 74 | |
| CR456766.1 | - | 1 - 2 | 139 | 74 | ||
| DA020204.1 | - | 2 - 3 | 430 | 74 | ||
| DA781730.1 | - | 2 - 3 | 435 | 74 | ||
| DB026977.1 | - | 2 - 3 | 351 | 74 | ||
| U35451.1 | - | 2 - 3 | 355 | 74 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC17001635.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009166.hg.1 | ENST00000453841.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17021890.hg.1 | ENST00000225603.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17021890.hg.1 | NM_001127228.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17021890.hg.1 | TC17001635.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR17021889.hg.1 | ENST00000225603.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17021889.hg.1 | NM_001127228.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17021889.hg.1 | TC17001635.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR17021881.hg.1 | ENST00000225603.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17021881.hg.1 | ENST00000393408.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17021881.hg.1 | NM_001127228.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17021881.hg.1 | NM_006807.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17021881.hg.1 | TC17001635.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009399.hg.1 | ENST00000475330.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010979.hg.1 | ENST00000225603.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010979.hg.1 | ENST00000225603.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010979.hg.1 | ENST00000225603.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010979.hg.1 | ENST00000225603.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010979.hg.1 | ENST00000225603.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010979.hg.1 | ENST00000225603.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010979.hg.1 | ENST00000225603.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010979.hg.1 | ENST00000225603.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010979.hg.1 | ENST00000225603.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010979.hg.1 | ENST00000225603.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010979.hg.1 | ENST00000225603.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010979.hg.1 | ENST00000393408.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010979.hg.1 | ENST00000393408.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010979.hg.1 | ENST00000393408.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010979.hg.1 | ENST00000393408.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010979.hg.1 | ENST00000393408.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010979.hg.1 | ENST00000393408.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010979.hg.1 | ENST00000393408.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010979.hg.1 | ENST00000393408.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010979.hg.1 | ENST00000393408.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010979.hg.1 | ENST00000393408.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010979.hg.1 | ENST00000402583.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010979.hg.1 | ENST00000402583.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010979.hg.1 | ENST00000402583.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010979.hg.1 | ENST00000402583.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010979.hg.1 | ENST00000402583.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010979.hg.1 | ENST00000444685.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010979.hg.1 | ENST00000444685.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010979.hg.1 | ENST00000444685.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010979.hg.1 | ENST00000444685.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010979.hg.1 | ENST00000444685.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010979.hg.1 | ENST00000495350.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010979.hg.1 | ENST00000495350.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010979.hg.1 | ENST00000495350.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010979.hg.1 | ENST00000495350.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010979.hg.1 | ENST00000495350.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010979.hg.1 | ENST00000581003.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010979.hg.1 | ENST00000581003.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010979.hg.1 | ENST00000581003.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010979.hg.1 | ENST00000581003.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010979.hg.1 | ENST00000581003.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010979.hg.1 | ENST00000581003.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010979.hg.1 | ENST00000581003.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010979.hg.1 | ENST00000581003.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010979.hg.1 | NM_001127228.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010979.hg.1 | NM_001127228.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010979.hg.1 | NM_001127228.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010979.hg.1 | NM_001127228.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010979.hg.1 | NM_001127228.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010979.hg.1 | NM_001127228.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010979.hg.1 | NM_001127228.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010979.hg.1 | NM_001127228.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010979.hg.1 | NM_001127228.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010979.hg.1 | NM_001127228.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010979.hg.1 | NM_001127228.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010979.hg.1 | NM_006807.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010979.hg.1 | NM_006807.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010979.hg.1 | NM_006807.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010979.hg.1 | NM_006807.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010979.hg.1 | NM_006807.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010979.hg.1 | NM_006807.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010979.hg.1 | NM_006807.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010979.hg.1 | NM_006807.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010979.hg.1 | NM_006807.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010979.hg.1 | NM_006807.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006707.hg.1 | ENST00000439415.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17021891.hg.1 | ENST00000225603.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17021891.hg.1 | NM_001127228.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17021891.hg.1 | TC17001635.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1700188311.hg.1 | ENST00000225603.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700188311.hg.1 | ENST00000393408.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700188311.hg.1 | ENST00000402583.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700188311.hg.1 | ENST00000444685.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700188311.hg.1 | ENST00000495350.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700188311.hg.1 | ENST00000581003.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700188311.hg.1 | NM_001127228.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700188311.hg.1 | NM_006807.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700188311.hg.1 | ENST00000225603.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700188311.hg.1 | ENST00000393408.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700188311.hg.1 | ENST00000402583.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700188311.hg.1 | ENST00000444685.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700188311.hg.1 | ENST00000495350.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700188311.hg.1 | ENST00000581003.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700188311.hg.1 | NM_001127228.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700188311.hg.1 | NM_006807.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700188311.hg.1 | TC1700010979.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700188321.hg.1 | ENST00000225603.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700188321.hg.1 | ENST00000581003.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700188321.hg.1 | NM_001127228.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700188321.hg.1 | ENST00000225603.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700188321.hg.1 | ENST00000581003.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700188321.hg.1 | NM_001127228.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700188321.hg.1 | TC1700010979.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700188322.hg.1 | ENST00000225603.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700188322.hg.1 | NM_001127228.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700188322.hg.1 | ENST00000225603.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700188322.hg.1 | NM_001127228.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700188322.hg.1 | TC1700010979.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0X00009166.hg.1 | ENST00000453841.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009399.hg.1 | ENST00000475330.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010979.hg.1 | ENST00000225603.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010979.hg.1 | ENST00000225603.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010979.hg.1 | ENST00000225603.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010979.hg.1 | ENST00000225603.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010979.hg.1 | ENST00000225603.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010979.hg.1 | ENST00000225603.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010979.hg.1 | ENST00000225603.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010979.hg.1 | ENST00000225603.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010979.hg.1 | ENST00000225603.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010979.hg.1 | ENST00000225603.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010979.hg.1 | ENST00000225603.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010979.hg.1 | ENST00000393408.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010979.hg.1 | ENST00000393408.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010979.hg.1 | ENST00000393408.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010979.hg.1 | ENST00000393408.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010979.hg.1 | ENST00000393408.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010979.hg.1 | ENST00000393408.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010979.hg.1 | ENST00000393408.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010979.hg.1 | ENST00000393408.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010979.hg.1 | ENST00000393408.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010979.hg.1 | ENST00000393408.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010979.hg.1 | ENST00000402583.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010979.hg.1 | ENST00000402583.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010979.hg.1 | ENST00000402583.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010979.hg.1 | ENST00000402583.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010979.hg.1 | ENST00000402583.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010979.hg.1 | ENST00000444685.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010979.hg.1 | ENST00000444685.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010979.hg.1 | ENST00000444685.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010979.hg.1 | ENST00000444685.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010979.hg.1 | ENST00000444685.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010979.hg.1 | ENST00000495350.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010979.hg.1 | ENST00000495350.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010979.hg.1 | ENST00000495350.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010979.hg.1 | ENST00000495350.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010979.hg.1 | ENST00000495350.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010979.hg.1 | ENST00000581003.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010979.hg.1 | ENST00000581003.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010979.hg.1 | ENST00000581003.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010979.hg.1 | ENST00000581003.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010979.hg.1 | ENST00000581003.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010979.hg.1 | ENST00000581003.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010979.hg.1 | ENST00000581003.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010979.hg.1 | ENST00000581003.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010979.hg.1 | NM_001127228.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010979.hg.1 | NM_001127228.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010979.hg.1 | NM_001127228.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010979.hg.1 | NM_001127228.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010979.hg.1 | NM_001127228.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010979.hg.1 | NM_001127228.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010979.hg.1 | NM_001127228.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010979.hg.1 | NM_001127228.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010979.hg.1 | NM_001127228.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010979.hg.1 | NM_001127228.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010979.hg.1 | NM_001127228.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010979.hg.1 | NM_006807.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010979.hg.1 | NM_006807.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010979.hg.1 | NM_006807.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010979.hg.1 | NM_006807.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010979.hg.1 | NM_006807.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010979.hg.1 | NM_006807.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010979.hg.1 | NM_006807.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010979.hg.1 | NM_006807.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010979.hg.1 | NM_006807.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010979.hg.1 | NM_006807.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700188323.hg.1 | ENST00000225603.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700188323.hg.1 | NM_001127228.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700188323.hg.1 | ENST00000225603.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700188323.hg.1 | NM_001127228.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700188323.hg.1 | TC1700010979.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0X00006707.hg.1 | ENST00000439415.1 | Ensembl |

CBX1
Genomic Map