Assay Details
| Gene Symbol: MAL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022440.2 | NP_071885.1 | 1 - 2 | 236 | 86 | |
| GenBank mRNA | X76681.1 | - | 1 - 2 | 98 | 86 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: MAL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022440.2 | NP_071885.1 | 1 - 2 | 236 | 86 | |
| GenBank mRNA | X76681.1 | - | 1 - 2 | 98 | 86 | |
Set Membership: |
|