Assay Details
| Gene Symbol: CA12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001218.4 | NP_001209.1 | 8 - 9 | 1263 | 66 | |
| GenBank mRNA | AF037335.1 | - | 8 - 9 | 988 | 66 | |
| AF051882.1 | - | 8 - 9 | 880 | 66 | ||
| AK315769.1 | - | 8 - 9 | 1029 | 66 | ||
| BC023981.1 | - | 8 - 9 | 1003 | 4042589 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | ENST00000178638.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | ENST00000178638.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | ENST00000178638.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | ENST00000178638.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | ENST00000178638.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | ENST00000178638.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | ENST00000178638.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | ENST00000178638.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | ENST00000178638.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | ENST00000178638.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | ENST00000178638.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | ENST00000178638.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | ENST00000178638.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | ENST00000178638.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | ENST00000178638.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | ENST00000178638.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | ENST00000178638.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | ENST00000178638.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | ENST00000344366.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | ENST00000344366.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | ENST00000344366.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | ENST00000344366.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | ENST00000344366.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | ENST00000344366.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | ENST00000344366.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | ENST00000344366.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | ENST00000344366.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | ENST00000344366.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | ENST00000344366.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | ENST00000344366.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | ENST00000344366.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | ENST00000422263.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | ENST00000422263.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | ENST00000422263.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | ENST00000422263.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | ENST00000422263.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | ENST00000422263.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | ENST00000422263.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | ENST00000422263.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | ENST00000422263.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | ENST00000422263.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | ENST00000422263.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | ENST00000422263.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | ENST00000558287.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | ENST00000558287.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | ENST00000558287.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | ENST00000558287.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | ENST00000560293.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | ENST00000560293.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | ENST00000560666.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | ENST00000560666.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | ENST00000560666.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | ENST00000560666.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | NM_001218.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | NM_001218.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | NM_001218.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | NM_001218.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | NM_001218.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | NM_001218.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | NM_001218.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | NM_001218.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | NM_001218.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | NM_001218.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | NM_001218.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | NM_001218.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | NM_001218.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | NM_001218.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | NM_001218.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | NM_001218.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | NM_001293642.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | NM_001293642.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | NM_001293642.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | NM_001293642.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | NM_001293642.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | NM_001293642.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | NM_001293642.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | NM_001293642.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | NM_001293642.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | NM_001293642.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | NM_001293642.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | NM_001293642.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | NM_001293642.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | NM_001293642.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | NM_206925.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | NM_206925.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | NM_206925.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | NM_206925.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | NM_206925.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | NM_206925.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | NM_206925.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | NM_206925.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | NM_206925.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | NM_206925.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | NM_206925.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | NM_206925.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | NM_206925.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | NM_206925.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | NM_206925.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | ENST00000178638.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | ENST00000178638.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | ENST00000178638.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | ENST00000178638.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | ENST00000178638.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | ENST00000178638.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | ENST00000178638.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | ENST00000178638.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | ENST00000178638.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | ENST00000178638.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | ENST00000178638.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | ENST00000178638.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | ENST00000178638.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | ENST00000178638.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | ENST00000178638.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | ENST00000178638.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | ENST00000178638.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | ENST00000178638.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | ENST00000344366.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | ENST00000344366.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | ENST00000344366.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | ENST00000344366.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | ENST00000344366.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | ENST00000344366.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | ENST00000344366.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | ENST00000344366.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | ENST00000344366.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | ENST00000344366.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | ENST00000344366.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | ENST00000344366.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | ENST00000344366.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | ENST00000422263.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | ENST00000422263.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | ENST00000422263.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | ENST00000422263.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | ENST00000422263.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | ENST00000422263.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | ENST00000422263.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | ENST00000422263.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | ENST00000422263.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | ENST00000422263.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | ENST00000422263.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | ENST00000422263.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | ENST00000558287.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | ENST00000558287.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | ENST00000558287.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | ENST00000558287.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | ENST00000560293.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | ENST00000560293.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | ENST00000560666.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | ENST00000560666.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | ENST00000560666.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | ENST00000560666.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | NM_001218.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | NM_001218.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | NM_001218.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | NM_001218.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | NM_001218.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | NM_001218.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | NM_001218.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | NM_001218.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | NM_001218.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | NM_001218.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | NM_001218.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | NM_001218.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | NM_001218.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | NM_001218.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | NM_001218.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | NM_001218.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | NM_001293642.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | NM_001293642.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | NM_001293642.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | NM_001293642.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | NM_001293642.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | NM_001293642.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | NM_001293642.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | NM_001293642.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | NM_001293642.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | NM_001293642.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | NM_001293642.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | NM_001293642.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | NM_001293642.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | NM_001293642.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | NM_206925.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | NM_206925.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | NM_206925.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | NM_206925.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | NM_206925.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | NM_206925.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | NM_206925.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | NM_206925.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | NM_206925.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | NM_206925.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | NM_206925.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | NM_206925.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | NM_206925.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | NM_206925.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | NM_206925.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500171615.hg.1 | ENST00000178638.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500171615.hg.1 | NM_001218.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500171615.hg.1 | ENST00000178638.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500171615.hg.1 | NM_001218.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500171615.hg.1 | TC1500009709.hg.1 | Clariom D TC |

CA12
Genomic Map