Assay Details
| Gene Symbol: CA12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001293642.1 | NP_001280571.1 | 7 - 8 | 1086 | 64 | |
| NM_206925.2 | NP_996808.1 | 8 - 9 | 1266 | 64 | ||
| NR_135511.1 | 8 - 9 | 1369 | 64 | |||
| GenBank mRNA | AK096845.1 | - | 7 - 8 | 822 | 64 | |
| BC000278.1 | - | 8 - 9 | 988 | 3357439 | 64 | |
| BC011691.2 | - | 8 - 9 | 991 | 3344734 | 64 | |
| BT006656.1 | - | 8 - 9 | 876 | 64 | ||
More Information
Set Membership: |
|

CA12
Genomic Map