Assay Details
Gene Symbol: HDAC9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF124924.1 | - | 8 - 9 | 1165 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
