Assay Details
| Gene Symbol: ARHGEF39 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_032818.2 | NP_116207.2 | 7 - 8 | 1019 | 75 | |
| XM_017015226.1 | XP_016870715.1 | 6 - 7 | 814 | 75 | ||
| GenBank mRNA | AK001187.1 | - | 2 - 3 | 279 | 75 | |
| AK225529.1 | - | 7 - 8 | 75 | |||
| BC033666.1 | - | 7 - 8 | 956 | 5000577 | 75 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC09001057.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR09014025.hg.1 | TC09001057.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09014025.hg.1 | ENST00000343259.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09014025.hg.1 | ENST00000378387.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09014025.hg.1 | ENST00000378395.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09014025.hg.1 | ENST00000490970.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09014025.hg.1 | NM_032818.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09014026.hg.1 | TC09001057.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09014026.hg.1 | ENST00000343259.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09014026.hg.1 | ENST00000378387.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09014026.hg.1 | ENST00000378395.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09014026.hg.1 | ENST00000490970.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09014026.hg.1 | NM_032818.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009948.hg.1 | ENST00000378387.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009948.hg.1 | ENST00000378387.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009948.hg.1 | ENST00000378387.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009948.hg.1 | ENST00000378387.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009948.hg.1 | ENST00000378387.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009948.hg.1 | ENST00000378387.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009948.hg.1 | ENST00000378387.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009948.hg.1 | ENST00000378387.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009948.hg.1 | ENST00000378387.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009948.hg.1 | ENST00000378387.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009948.hg.1 | ENST00000378387.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009948.hg.1 | ENST00000378387.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009948.hg.1 | ENST00000378387.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009948.hg.1 | ENST00000378387.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009948.hg.1 | ENST00000378387.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009948.hg.1 | ENST00000378387.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009948.hg.1 | ENST00000378387.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009948.hg.1 | ENST00000378387.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009948.hg.1 | ENST00000468876.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009948.hg.1 | ENST00000468876.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009948.hg.1 | ENST00000468876.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009948.hg.1 | ENST00000468876.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009948.hg.1 | ENST00000468876.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009948.hg.1 | ENST00000468876.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009948.hg.1 | ENST00000475323.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009948.hg.1 | ENST00000475323.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009948.hg.1 | ENST00000475323.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009948.hg.1 | ENST00000475323.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009948.hg.1 | ENST00000475323.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009948.hg.1 | ENST00000475323.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009948.hg.1 | ENST00000475323.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009948.hg.1 | ENST00000475323.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009948.hg.1 | ENST00000475323.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009948.hg.1 | ENST00000475323.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009948.hg.1 | ENST00000475323.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009948.hg.1 | ENST00000475323.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009948.hg.1 | ENST00000475323.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009948.hg.1 | ENST00000475323.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009948.hg.1 | ENST00000488918.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009948.hg.1 | ENST00000488918.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009948.hg.1 | ENST00000488918.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009948.hg.1 | ENST00000490638.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009948.hg.1 | ENST00000490638.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009948.hg.1 | ENST00000490638.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009948.hg.1 | ENST00000490638.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009948.hg.1 | ENST00000490638.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009948.hg.1 | ENST00000490638.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009948.hg.1 | ENST00000490638.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009948.hg.1 | ENST00000490638.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009948.hg.1 | ENST00000490638.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009948.hg.1 | ENST00000490638.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009948.hg.1 | ENST00000490638.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009948.hg.1 | ENST00000490638.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009948.hg.1 | ENST00000490638.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009948.hg.1 | ENST00000490638.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009948.hg.1 | ENST00000490638.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009948.hg.1 | ENST00000490970.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009948.hg.1 | ENST00000490970.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009948.hg.1 | ENST00000490970.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009948.hg.1 | ENST00000490970.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009948.hg.1 | ENST00000490970.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009948.hg.1 | ENST00000490970.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009948.hg.1 | ENST00000490970.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009948.hg.1 | ENST00000490970.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009948.hg.1 | ENST00000490970.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009948.hg.1 | ENST00000490970.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009948.hg.1 | ENST00000490970.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009948.hg.1 | NM_032818.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009948.hg.1 | NM_032818.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009948.hg.1 | NM_032818.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009948.hg.1 | NM_032818.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009948.hg.1 | NM_032818.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009948.hg.1 | NM_032818.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009948.hg.1 | NM_032818.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009948.hg.1 | NM_032818.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009948.hg.1 | NM_032818.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009948.hg.1 | NM_032818.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009948.hg.1 | NM_032818.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009948.hg.1 | NM_032818.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009948.hg.1 | NM_032818.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009948.hg.1 | NM_032818.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009948.hg.1 | NM_032818.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009948.hg.1 | NM_032818.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009948.hg.1 | NM_032818.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009948.hg.1 | NM_032818.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900168390.hg.1 | ENST00000378387.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900168390.hg.1 | ENST00000475323.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900168390.hg.1 | ENST00000488918.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900168390.hg.1 | ENST00000490638.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900168390.hg.1 | ENST00000490970.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900168390.hg.1 | NM_032818.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900168390.hg.1 | ENST00000378387.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900168390.hg.1 | ENST00000475323.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900168390.hg.1 | ENST00000488918.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900168390.hg.1 | ENST00000490638.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900168390.hg.1 | ENST00000490970.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900168390.hg.1 | NM_032818.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900168390.hg.1 | TC0900009948.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0900168391.hg.1 | ENST00000378387.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900168391.hg.1 | ENST00000475323.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900168391.hg.1 | ENST00000490638.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900168391.hg.1 | ENST00000490970.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900168391.hg.1 | NM_032818.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900168391.hg.1 | ENST00000378387.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900168391.hg.1 | ENST00000475323.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900168391.hg.1 | ENST00000490638.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900168391.hg.1 | ENST00000490970.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900168391.hg.1 | NM_032818.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900168391.hg.1 | TC0900009948.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0900009948.hg.1 | ENST00000378387.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009948.hg.1 | ENST00000378387.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009948.hg.1 | ENST00000378387.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009948.hg.1 | ENST00000378387.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009948.hg.1 | ENST00000378387.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009948.hg.1 | ENST00000378387.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009948.hg.1 | ENST00000378387.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009948.hg.1 | ENST00000378387.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009948.hg.1 | ENST00000378387.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009948.hg.1 | ENST00000378387.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009948.hg.1 | ENST00000378387.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009948.hg.1 | ENST00000378387.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009948.hg.1 | ENST00000378387.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009948.hg.1 | ENST00000378387.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009948.hg.1 | ENST00000378387.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009948.hg.1 | ENST00000378387.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009948.hg.1 | ENST00000378387.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009948.hg.1 | ENST00000378387.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009948.hg.1 | ENST00000468876.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009948.hg.1 | ENST00000468876.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009948.hg.1 | ENST00000468876.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009948.hg.1 | ENST00000468876.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009948.hg.1 | ENST00000468876.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009948.hg.1 | ENST00000468876.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009948.hg.1 | ENST00000475323.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009948.hg.1 | ENST00000475323.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009948.hg.1 | ENST00000475323.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009948.hg.1 | ENST00000475323.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009948.hg.1 | ENST00000475323.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009948.hg.1 | ENST00000475323.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009948.hg.1 | ENST00000475323.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009948.hg.1 | ENST00000475323.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009948.hg.1 | ENST00000475323.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009948.hg.1 | ENST00000475323.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009948.hg.1 | ENST00000475323.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009948.hg.1 | ENST00000475323.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009948.hg.1 | ENST00000475323.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009948.hg.1 | ENST00000475323.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009948.hg.1 | ENST00000488918.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009948.hg.1 | ENST00000488918.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009948.hg.1 | ENST00000488918.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009948.hg.1 | ENST00000490638.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009948.hg.1 | ENST00000490638.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009948.hg.1 | ENST00000490638.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009948.hg.1 | ENST00000490638.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009948.hg.1 | ENST00000490638.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009948.hg.1 | ENST00000490638.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009948.hg.1 | ENST00000490638.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009948.hg.1 | ENST00000490638.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009948.hg.1 | ENST00000490638.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009948.hg.1 | ENST00000490638.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009948.hg.1 | ENST00000490638.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009948.hg.1 | ENST00000490638.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009948.hg.1 | ENST00000490638.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009948.hg.1 | ENST00000490638.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009948.hg.1 | ENST00000490638.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009948.hg.1 | ENST00000490970.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009948.hg.1 | ENST00000490970.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009948.hg.1 | ENST00000490970.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009948.hg.1 | ENST00000490970.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009948.hg.1 | ENST00000490970.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009948.hg.1 | ENST00000490970.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009948.hg.1 | ENST00000490970.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009948.hg.1 | ENST00000490970.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009948.hg.1 | ENST00000490970.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009948.hg.1 | ENST00000490970.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009948.hg.1 | ENST00000490970.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009948.hg.1 | NM_032818.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009948.hg.1 | NM_032818.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009948.hg.1 | NM_032818.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009948.hg.1 | NM_032818.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009948.hg.1 | NM_032818.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009948.hg.1 | NM_032818.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009948.hg.1 | NM_032818.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009948.hg.1 | NM_032818.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009948.hg.1 | NM_032818.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009948.hg.1 | NM_032818.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009948.hg.1 | NM_032818.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009948.hg.1 | NM_032818.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009948.hg.1 | NM_032818.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009948.hg.1 | NM_032818.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009948.hg.1 | NM_032818.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009948.hg.1 | NM_032818.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009948.hg.1 | NM_032818.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009948.hg.1 | NM_032818.2 | Refseq |

ARHGEF39
Genomic Map