Assay Details
| Gene Symbol: CREB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001320793.1 | NP_001307722.1 | 3 - 4 | 508 | 84 | |
| NM_004379.4 | NP_004370.1 | 3 - 4 | 508 | 84 | ||
| XM_011510647.2 | XP_011508949.1 | 3 - 4 | 365 | 84 | ||
| XM_011510651.2 | XP_011508953.1 | 2 - 3 | 277 | 84 | ||
| XM_017003399.1 | XP_016858888.1 | 3 - 4 | 458 | 84 | ||
| XM_017003400.1 | XP_016858889.1 | 2 - 3 | 364 | 84 | ||
| GenBank mRNA | AK298740.1 | - | 3 - 4 | 330 | 84 | |
| AK315226.1 | - | 3 - 4 | 299 | 84 | ||
| BC095407.1 | - | 3 - 4 | 452 | 30344522 | 84 | |
| CR407617.1 | - | 2 - 3 | 257 | 84 | ||
| M27691.1 | - | 3 - 4 | 373 | 84 | ||
| X60003.1 | - | 2 - 3 | 257 | 84 | ||
More Information
Set Membership: |
|

CREB1
Genomic Map