Assay Details
| Gene Symbol: PLXNA4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020911.1 | NP_065962.1 | 32 | 9111 | 103 | |
| XM_005250686.4 | XP_005250743.1 | 32 | 9509 | 103 | ||
| XM_006716171.3 | XP_006716234.1 | 33 | 9602 | 103 | ||
| XM_017012779.1 | XP_016868268.1 | 31 | 8816 | 103 | ||
| GenBank mRNA | AB046770.3 | - | 30 | 7574 | 103 | |
| AK055487.1 | - | 1 | 538 | 103 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

PLXNA4
Genomic Map