Assay Details
| Gene Symbol: SEPHS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | U34044.1 | - | 1 - 2 | 184 | 72 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the U34044.1 transcripts.
Set Membership: |
|
Panther Classification:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | ENST00000327347.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | ENST00000327347.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | ENST00000327347.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | ENST00000327347.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | ENST00000327347.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | ENST00000327347.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | ENST00000327347.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | ENST00000327347.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | ENST00000327347.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | ENST00000327347.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | ENST00000327347.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | ENST00000327347.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | ENST00000327347.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | ENST00000327347.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | ENST00000327347.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | ENST00000378614.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | ENST00000378614.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | ENST00000378614.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | ENST00000378614.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | ENST00000378614.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | ENST00000378614.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | ENST00000378614.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | ENST00000378614.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | ENST00000378614.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | ENST00000378614.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | ENST00000378614.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | ENST00000378614.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | ENST00000378614.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | ENST00000413411.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | ENST00000413411.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | ENST00000413411.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | ENST00000413411.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | ENST00000413411.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | ENST00000413411.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | ENST00000425947.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | ENST00000425947.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | ENST00000425947.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | ENST00000425947.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | ENST00000494329.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | ENST00000494329.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | ENST00000494329.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | ENST00000494329.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | ENST00000545675.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | ENST00000545675.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | ENST00000545675.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | ENST00000545675.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | ENST00000545675.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | ENST00000545675.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | ENST00000545675.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | ENST00000545675.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | ENST00000545675.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | ENST00000545675.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | ENST00000545675.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | ENST00000545675.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | ENST00000545675.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | NM_001195602.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | NM_001195602.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | NM_001195602.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | NM_001195602.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | NM_001195602.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | NM_001195602.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | NM_001195602.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | NM_001195602.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | NM_001195602.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | NM_001195602.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | NM_001195602.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | NM_001195602.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | NM_001195602.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | NM_001195604.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | NM_001195604.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | NM_001195604.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | NM_001195604.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | NM_001195604.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | NM_001195604.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | NM_001195604.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | NM_001195604.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | NM_001195604.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | NM_001195604.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | NM_001195604.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | NM_001195604.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | NM_001195604.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | NM_001195604.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | NM_012247.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | NM_012247.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | NM_012247.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | NM_012247.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | NM_012247.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | NM_012247.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | NM_012247.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | NM_012247.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | NM_012247.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | NM_012247.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | NM_012247.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | NM_012247.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | NM_012247.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | NM_012247.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | NM_012247.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | ENST00000327347.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | ENST00000327347.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | ENST00000327347.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | ENST00000327347.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | ENST00000327347.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | ENST00000327347.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | ENST00000327347.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | ENST00000327347.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | ENST00000327347.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | ENST00000327347.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | ENST00000327347.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | ENST00000327347.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | ENST00000327347.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | ENST00000327347.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | ENST00000327347.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | ENST00000378614.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | ENST00000378614.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | ENST00000378614.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | ENST00000378614.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | ENST00000378614.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | ENST00000378614.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | ENST00000378614.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | ENST00000378614.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | ENST00000378614.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | ENST00000378614.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | ENST00000378614.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | ENST00000378614.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | ENST00000378614.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | ENST00000413411.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | ENST00000413411.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | ENST00000413411.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | ENST00000413411.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | ENST00000413411.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | ENST00000413411.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | ENST00000425947.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | ENST00000425947.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | ENST00000425947.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | ENST00000425947.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | ENST00000494329.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | ENST00000494329.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | ENST00000494329.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | ENST00000494329.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | ENST00000545675.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | ENST00000545675.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | ENST00000545675.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | ENST00000545675.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | ENST00000545675.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | ENST00000545675.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | ENST00000545675.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | ENST00000545675.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | ENST00000545675.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | ENST00000545675.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | ENST00000545675.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | ENST00000545675.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | ENST00000545675.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | NM_001195602.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | NM_001195602.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | NM_001195602.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | NM_001195602.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | NM_001195602.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | NM_001195602.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | NM_001195602.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | NM_001195602.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | NM_001195602.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | NM_001195602.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | NM_001195602.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | NM_001195602.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | NM_001195602.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | NM_001195604.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | NM_001195604.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | NM_001195604.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | NM_001195604.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | NM_001195604.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | NM_001195604.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | NM_001195604.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | NM_001195604.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | NM_001195604.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | NM_001195604.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | NM_001195604.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | NM_001195604.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | NM_001195604.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | NM_001195604.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | NM_012247.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | NM_012247.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | NM_012247.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | NM_012247.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | NM_012247.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | NM_012247.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | NM_012247.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | NM_012247.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | NM_012247.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | NM_012247.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | NM_012247.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | NM_012247.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | NM_012247.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | NM_012247.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | NM_012247.4 | Refseq |

SEPHS1
Genomic Map