Assay Details
| Gene Symbol: TRPC4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001135956.1 | NP_001129428.1 | 7 - 8 | 2121 | 131 | |
| GenBank mRNA | AF063825.1 | - | 7 - 8 | 2121 | 131 | |
More Information
Set Membership: |
|

TRPC4
Genomic Map