Assay Details
| Gene Symbol: LSAMP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001318915.1 | NP_001305844.1 | 5 - 6 | 1269 | 72 | |
| NM_002338.4 | NP_002329.2 | 5 - 6 | 1269 | 72 | ||
| XM_011512840.2 | XP_011511142.1 | 5 - 6 | 1273 | 72 | ||
| XM_017006383.1 | XP_016861872.1 | 5 - 6 | 1273 | 72 | ||
| XM_017006384.1 | XP_016861873.1 | 4 - 5 | 491 | 72 | ||
| GenBank mRNA | AK299851.1 | - | 1084 | 72 | ||
| AK315286.1 | - | 5 - 6 | 1208 | 72 | ||
| BC033803.1 | - | 5 - 6 | 1262 | 5188760 | 72 | |
| U41901.1 | - | 5 - 6 | 762 | 72 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC03001673.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR03030197.hg.1 | TC03001673.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03030197.hg.1 | ENST00000490035.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03030197.hg.1 | ENST00000498645.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03030197.hg.1 | ENST00000539563.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03030197.hg.1 | NM_002338.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03030196.hg.1 | TC03001673.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03030196.hg.1 | ENST00000490035.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03030196.hg.1 | ENST00000498645.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03030196.hg.1 | ENST00000539563.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03030196.hg.1 | NM_002338.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012073.hg.1 | ENST00000333617.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012073.hg.1 | ENST00000333617.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012073.hg.1 | ENST00000333617.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012073.hg.1 | ENST00000333617.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012073.hg.1 | ENST00000333617.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012073.hg.1 | ENST00000333617.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012073.hg.1 | ENST00000333617.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012073.hg.1 | ENST00000333617.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012073.hg.1 | ENST00000333617.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012073.hg.1 | ENST00000333617.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012073.hg.1 | ENST00000333617.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012073.hg.1 | ENST00000333617.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012073.hg.1 | ENST00000333617.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012073.hg.1 | ENST00000333617.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012073.hg.1 | ENST00000473171.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012073.hg.1 | ENST00000473171.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012073.hg.1 | ENST00000473171.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012073.hg.1 | ENST00000473171.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012073.hg.1 | ENST00000473171.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012073.hg.1 | ENST00000473171.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012073.hg.1 | ENST00000474851.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012073.hg.1 | ENST00000474851.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012073.hg.1 | ENST00000474851.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012073.hg.1 | ENST00000474851.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012073.hg.1 | ENST00000474851.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012073.hg.1 | ENST00000475403.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012073.hg.1 | ENST00000475403.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012073.hg.1 | ENST00000475403.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012073.hg.1 | ENST00000475403.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012073.hg.1 | ENST00000475403.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012073.hg.1 | ENST00000475403.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012073.hg.1 | ENST00000475403.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012073.hg.1 | ENST00000490035.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012073.hg.1 | ENST00000490035.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012073.hg.1 | ENST00000490035.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012073.hg.1 | ENST00000490035.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012073.hg.1 | ENST00000490035.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012073.hg.1 | ENST00000490035.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012073.hg.1 | ENST00000490035.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012073.hg.1 | ENST00000490035.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012073.hg.1 | ENST00000490035.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012073.hg.1 | ENST00000490035.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012073.hg.1 | ENST00000490035.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012073.hg.1 | ENST00000490035.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012073.hg.1 | ENST00000490035.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012073.hg.1 | ENST00000490035.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012073.hg.1 | ENST00000490035.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012073.hg.1 | ENST00000490035.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012073.hg.1 | ENST00000498645.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012073.hg.1 | ENST00000498645.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012073.hg.1 | ENST00000498645.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012073.hg.1 | ENST00000498645.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012073.hg.1 | ENST00000498645.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012073.hg.1 | ENST00000498645.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012073.hg.1 | ENST00000498645.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012073.hg.1 | ENST00000539563.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012073.hg.1 | ENST00000539563.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012073.hg.1 | ENST00000539563.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012073.hg.1 | ENST00000539563.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012073.hg.1 | ENST00000539563.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012073.hg.1 | ENST00000539563.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012073.hg.1 | ENST00000539563.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012073.hg.1 | ENST00000539563.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012073.hg.1 | ENST00000539563.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012073.hg.1 | ENST00000539563.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012073.hg.1 | ENST00000539563.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012073.hg.1 | ENST00000539563.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012073.hg.1 | NM_002338.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012073.hg.1 | NM_002338.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012073.hg.1 | NM_002338.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012073.hg.1 | NM_002338.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012073.hg.1 | NM_002338.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012073.hg.1 | NM_002338.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012073.hg.1 | NM_002338.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012073.hg.1 | NM_002338.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012073.hg.1 | NM_002338.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012073.hg.1 | NM_002338.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012073.hg.1 | NM_002338.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012073.hg.1 | NM_002338.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012073.hg.1 | NM_002338.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012073.hg.1 | NM_002338.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012073.hg.1 | NM_002338.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012073.hg.1 | NM_002338.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012073.hg.1 | ENST00000333617.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012073.hg.1 | ENST00000333617.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012073.hg.1 | ENST00000333617.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012073.hg.1 | ENST00000333617.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012073.hg.1 | ENST00000333617.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012073.hg.1 | ENST00000333617.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012073.hg.1 | ENST00000333617.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012073.hg.1 | ENST00000333617.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012073.hg.1 | ENST00000333617.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012073.hg.1 | ENST00000333617.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012073.hg.1 | ENST00000333617.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012073.hg.1 | ENST00000333617.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012073.hg.1 | ENST00000333617.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012073.hg.1 | ENST00000333617.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012073.hg.1 | ENST00000473171.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012073.hg.1 | ENST00000473171.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012073.hg.1 | ENST00000473171.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012073.hg.1 | ENST00000473171.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012073.hg.1 | ENST00000473171.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012073.hg.1 | ENST00000473171.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012073.hg.1 | ENST00000474851.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012073.hg.1 | ENST00000474851.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012073.hg.1 | ENST00000474851.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012073.hg.1 | ENST00000474851.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012073.hg.1 | ENST00000474851.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012073.hg.1 | ENST00000475403.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012073.hg.1 | ENST00000475403.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012073.hg.1 | ENST00000475403.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012073.hg.1 | ENST00000475403.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012073.hg.1 | ENST00000475403.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012073.hg.1 | ENST00000475403.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012073.hg.1 | ENST00000475403.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012073.hg.1 | ENST00000490035.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012073.hg.1 | ENST00000490035.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012073.hg.1 | ENST00000490035.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012073.hg.1 | ENST00000490035.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012073.hg.1 | ENST00000490035.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012073.hg.1 | ENST00000490035.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012073.hg.1 | ENST00000490035.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012073.hg.1 | ENST00000490035.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012073.hg.1 | ENST00000490035.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012073.hg.1 | ENST00000490035.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012073.hg.1 | ENST00000490035.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012073.hg.1 | ENST00000490035.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012073.hg.1 | ENST00000490035.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012073.hg.1 | ENST00000490035.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012073.hg.1 | ENST00000490035.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012073.hg.1 | ENST00000490035.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012073.hg.1 | ENST00000498645.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012073.hg.1 | ENST00000498645.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012073.hg.1 | ENST00000498645.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012073.hg.1 | ENST00000498645.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012073.hg.1 | ENST00000498645.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012073.hg.1 | ENST00000498645.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012073.hg.1 | ENST00000498645.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012073.hg.1 | ENST00000539563.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012073.hg.1 | ENST00000539563.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012073.hg.1 | ENST00000539563.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012073.hg.1 | ENST00000539563.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012073.hg.1 | ENST00000539563.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012073.hg.1 | ENST00000539563.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012073.hg.1 | ENST00000539563.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012073.hg.1 | ENST00000539563.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012073.hg.1 | ENST00000539563.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012073.hg.1 | ENST00000539563.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012073.hg.1 | ENST00000539563.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012073.hg.1 | ENST00000539563.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012073.hg.1 | NM_002338.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012073.hg.1 | NM_002338.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012073.hg.1 | NM_002338.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012073.hg.1 | NM_002338.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012073.hg.1 | NM_002338.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012073.hg.1 | NM_002338.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012073.hg.1 | NM_002338.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012073.hg.1 | NM_002338.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012073.hg.1 | NM_002338.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012073.hg.1 | NM_002338.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012073.hg.1 | NM_002338.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012073.hg.1 | NM_002338.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012073.hg.1 | NM_002338.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012073.hg.1 | NM_002338.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012073.hg.1 | NM_002338.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012073.hg.1 | NM_002338.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300189947.hg.1 | ENST00000333617.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300189947.hg.1 | ENST00000473171.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300189947.hg.1 | ENST00000475403.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300189947.hg.1 | ENST00000490035.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300189947.hg.1 | ENST00000498645.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300189947.hg.1 | ENST00000539563.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300189947.hg.1 | NM_002338.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300189947.hg.1 | ENST00000333617.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300189947.hg.1 | ENST00000473171.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300189947.hg.1 | ENST00000475403.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300189947.hg.1 | ENST00000490035.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300189947.hg.1 | ENST00000498645.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300189947.hg.1 | ENST00000539563.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300189947.hg.1 | NM_002338.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300189947.hg.1 | TC0300012073.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0300189948.hg.1 | ENST00000333617.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300189948.hg.1 | ENST00000473171.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300189948.hg.1 | ENST00000475403.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300189948.hg.1 | ENST00000490035.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300189948.hg.1 | ENST00000498645.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300189948.hg.1 | ENST00000539563.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300189948.hg.1 | NM_002338.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300189948.hg.1 | ENST00000333617.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300189948.hg.1 | ENST00000473171.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300189948.hg.1 | ENST00000475403.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300189948.hg.1 | ENST00000490035.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300189948.hg.1 | ENST00000498645.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300189948.hg.1 | ENST00000539563.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300189948.hg.1 | NM_002338.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300189948.hg.1 | TC0300012073.hg.1 | Clariom D TC |

LSAMP
Genomic Map